Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85267878-85268131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL10
Entrez Gene ID
8915
Gene Name
B-cell CLL/lymphoma 10
Gene Aliases
CARMEN, CIPER, CLAP, IMD37, c-E10, mE10
UniGene
Hs.193516
Gene Chromosome Location
Chr.1: 85265776-85276904 on Build GRCh38
Transcripts
NM_001320715.1 NM_003921.4 XM_011542397.2 XM_011542398.2 XM_011542399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205594 c.423delT p.S141fs*7 Chr.1: 85267906-85267906 on Build GRCh38
220637 c.407C>G p.S136* Chr.1: 85267922-85267922 on Build GRCh38
1602727 c.372T>G p.P124P Chr.1: 85267957-85267957 on Build GRCh38
6945522 c.421_422delAG p.S141fs*1 Chr.1: 85267907-85267908 on Build GRCh38
6207986 c.429C>T p.F143F Chr.1: 85267900-85267900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781284853 Chr.1: 85267905-85267905 on Build GRCh38
rs745885764 Chr.1: 85267927-85267927 on Build GRCh38
rs774261681 Chr.1: 85267933-85267933 on Build GRCh38
rs587776632 Chr.1: 85267902-85267903 on Build GRCh38
rs529023361 Chr.1: 85268077-85268077 on Build GRCh38
rs370245790 Chr.1: 85267937-85267937 on Build GRCh38
rs753115014 Chr.1: 85267979-85267979 on Build GRCh38
rs776283248 Chr.1: 85267964-85267964 on Build GRCh38
rs753358924 Chr.1: 85268082-85268082 on Build GRCh38
rs587776635 Chr.1: 85267919-85267919 on Build GRCh38
rs766089591 Chr.1: 85267963-85267963 on Build GRCh38
rs563408505 Chr.1: 85268050-85268050 on Build GRCh38
rs746009230 Chr.1: 85268022-85268022 on Build GRCh38
rs757479322 Chr.1: 85268002-85268002 on Build GRCh38
rs773197133 Chr.1: 85267966-85267967 on Build GRCh38
rs756178483 Chr.1: 85268027-85268027 on Build GRCh38
rs764388331 Chr.1: 85267986-85267986 on Build GRCh38
rs781336752 Chr.1: 85268019-85268019 on Build GRCh38
rs76553839 Chr.1: 85268055-85268059 on Build GRCh38
rs1034678876 Chr.1: 85267947-85267947 on Build GRCh38
rs587776636 Chr.1: 85267931-85267932 on Build GRCh38
rs760043303 Chr.1: 85268052-85268052 on Build GRCh38
rs956518784 Chr.1: 85268074-85268074 on Build GRCh38
rs762834577 Chr.1: 85268014-85268015 on Build GRCh38
rs587776638 Chr.1: 85267901-85267901 on Build GRCh38
rs374634792 Chr.1: 85267985-85267985 on Build GRCh38
rs759056975 Chr.1: 85267969-85267969 on Build GRCh38
rs367565183 Chr.1: 85267935-85267935 on Build GRCh38
rs185236679 Chr.1: 85268044-85268044 on Build GRCh38
rs199958663 Chr.1: 85268053-85268055 on Build GRCh38
rs748039228 Chr.1: 85267934-85267934 on Build GRCh38
rs77663629 Chr.1: 85267987-85267987 on Build GRCh38
rs766456443 Chr.1: 85268021-85268022 on Build GRCh38
rs190133949 Chr.1: 85268081-85268081 on Build GRCh38
rs1003194232 Chr.1: 85267939-85267939 on Build GRCh38
rs765912911 Chr.1: 85267978-85267978 on Build GRCh38
rs543106460 Chr.1: 85268046-85268046 on Build GRCh38
rs760264692 Chr.1: 85267941-85267941 on Build GRCh38
rs151312450 Chr.1: 85267919-85267919 on Build GRCh38

View Full Product Details