Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186151877-186152130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149A
Entrez Gene ID
25854
Gene Name
family with sequence similarity 149 member A
Gene Aliases
MST119, MSTP119
UniGene
Hs.744961
Gene Chromosome Location
Chr.4: 186104798-186172663 on Build GRCh38
Transcripts
NM_001006655.2 NM_015398.2 XM_005262911.1 XM_005262912.1 XM_005262913.1 XM_011531834.1 XM_011531835.2 XM_011531836.1 XM_017007996.1 XM_017007997.1 XM_017007998.1 XM_017007999.1 XM_017008000.1 XM_017008001.1 XM_017008002.1 XM_017008003.1 XM_017008004.1 XM_017008005.1 XM_017008006.1 XM_017008007.1 XM_017008008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842678 c.28C>G p.Q10E Chr.4: 186151987-186151987 on Build GRCh38
4842679 c.901C>G p.Q301E Chr.4: 186151987-186151987 on Build GRCh38
5928569 c.959+7C>T p.? Chr.4: 186152052-186152052 on Build GRCh38
7104097 c.888G>A p.G296G Chr.4: 186151974-186151974 on Build GRCh38
6895214 c.901C>T p.Q301* Chr.4: 186151987-186151987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745800363 Chr.4: 186151916-186151916 on Build GRCh38
rs773713434 Chr.4: 186152089-186152090 on Build GRCh38
rs868557399 Chr.4: 186151990-186151990 on Build GRCh38
rs770395092 Chr.4: 186151912-186151912 on Build GRCh38
rs749005261 Chr.4: 186151985-186151985 on Build GRCh38
rs756884346 Chr.4: 186152081-186152081 on Build GRCh38
rs766076273 Chr.4: 186151922-186151923 on Build GRCh38
rs778301511 Chr.4: 186152000-186152000 on Build GRCh38
rs1034317509 Chr.4: 186152008-186152008 on Build GRCh38
rs761106791 Chr.4: 186152052-186152052 on Build GRCh38
rs752034374 Chr.4: 186151945-186151945 on Build GRCh38
rs777545200 Chr.4: 186151981-186151981 on Build GRCh38
rs778421202 Chr.4: 186151952-186151952 on Build GRCh38
rs767941067 Chr.4: 186151961-186151961 on Build GRCh38
rs144466891 Chr.4: 186151975-186151975 on Build GRCh38
rs761984162 Chr.4: 186151929-186151929 on Build GRCh38
rs569913101 Chr.4: 186151922-186151922 on Build GRCh38
rs755451059 Chr.4: 186151953-186151953 on Build GRCh38
rs370157303 Chr.4: 186151935-186151935 on Build GRCh38
rs780010526 Chr.4: 186152006-186152006 on Build GRCh38
rs372317900 Chr.4: 186151936-186151936 on Build GRCh38
rs780975891 Chr.4: 186152095-186152095 on Build GRCh38
rs146640701 Chr.4: 186151998-186151998 on Build GRCh38
rs752125125 Chr.4: 186152054-186152054 on Build GRCh38
rs1002192300 Chr.4: 186152069-186152069 on Build GRCh38
rs375761611 Chr.4: 186151927-186151927 on Build GRCh38
rs768106559 Chr.4: 186152019-186152019 on Build GRCh38
rs759158928 Chr.4: 186152091-186152092 on Build GRCh38
rs377176675 Chr.4: 186152037-186152037 on Build GRCh38
rs757956296 Chr.4: 186152089-186152089 on Build GRCh38
rs765417973 Chr.4: 186151934-186151934 on Build GRCh38
rs756587438 Chr.4: 186151977-186151977 on Build GRCh38
rs779957273 Chr.4: 186152091-186152091 on Build GRCh38
rs768375964 Chr.4: 186152092-186152092 on Build GRCh38
rs367622396 Chr.4: 186152016-186152016 on Build GRCh38
rs749475482 Chr.4: 186152096-186152096 on Build GRCh38
rs753475001 Chr.4: 186152080-186152080 on Build GRCh38
rs372841067 Chr.4: 186152013-186152013 on Build GRCh38
rs369462778 Chr.4: 186152085-186152085 on Build GRCh38
rs374715651 Chr.4: 186151937-186151937 on Build GRCh38
rs749869735 Chr.4: 186152086-186152086 on Build GRCh38
rs369066270 Chr.4: 186152001-186152001 on Build GRCh38
rs763087733 Chr.4: 186152014-186152014 on Build GRCh38
rs764618067 Chr.4: 186152077-186152077 on Build GRCh38
rs555180542 Chr.4: 186152018-186152018 on Build GRCh38
rs770290197 Chr.4: 186151915-186151915 on Build GRCh38
rs192850325 Chr.4: 186151911-186151911 on Build GRCh38
rs775921104 Chr.4: 186152007-186152007 on Build GRCh38

View Full Product Details