Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 110875041-110875314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMD1
Entrez Gene ID
262
Gene Name
adenosylmethionine decarboxylase 1
Gene Aliases
ADOMETDC, AMD, SAMDC
UniGene
Hs.159118
Gene Chromosome Location
Chr.6: 110814621-110895713 on Build GRCh38
Transcripts
NM_001287214.1 NM_001287215.1 NM_001287216.1 NM_001634.5 NR_109768.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599062 c.31G>A p.E11K Chr.6: 110875136-110875136 on Build GRCh38
5552741 c.33G>A p.E11E Chr.6: 110875138-110875138 on Build GRCh38
4548895 c.45G>A p.E15E Chr.6: 110875150-110875150 on Build GRCh38
126549 c.109A>T p.R37* Chr.6: 110875214-110875214 on Build GRCh38
6448131 c.30C>T p.T10T Chr.6: 110875135-110875135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751122528 Chr.6: 110875132-110875132 on Build GRCh38
rs777727416 Chr.6: 110875163-110875163 on Build GRCh38
rs766944006 Chr.6: 110875244-110875244 on Build GRCh38
rs748942349 Chr.6: 110875215-110875216 on Build GRCh38
rs762027149 Chr.6: 110875219-110875219 on Build GRCh38
rs762221800 Chr.6: 110875083-110875083 on Build GRCh38
rs999410885 Chr.6: 110875255-110875255 on Build GRCh38
rs750718774 Chr.6: 110875090-110875090 on Build GRCh38
rs766756836 Chr.6: 110875118-110875118 on Build GRCh38
rs751998776 Chr.6: 110875228-110875228 on Build GRCh38
rs529884003 Chr.6: 110875225-110875225 on Build GRCh38
rs1012046781 Chr.6: 110875276-110875276 on Build GRCh38
rs776977473 Chr.6: 110875213-110875213 on Build GRCh38
rs755848950 Chr.6: 110875246-110875246 on Build GRCh38
rs776776300 Chr.6: 110875186-110875186 on Build GRCh38
rs900547327 Chr.6: 110875221-110875221 on Build GRCh38
rs752200598 Chr.6: 110875245-110875245 on Build GRCh38
rs770928759 Chr.6: 110875165-110875165 on Build GRCh38
rs759155969 Chr.6: 110875229-110875229 on Build GRCh38
rs754425919 Chr.6: 110875134-110875134 on Build GRCh38
rs367903991 Chr.6: 110875223-110875223 on Build GRCh38
rs765581668 Chr.6: 110875087-110875087 on Build GRCh38
rs140669549 Chr.6: 110875161-110875161 on Build GRCh38
rs749038400 Chr.6: 110875164-110875164 on Build GRCh38
rs753723963 Chr.6: 110875266-110875266 on Build GRCh38
rs777522934 Chr.6: 110875247-110875247 on Build GRCh38
rs768604319 Chr.6: 110875206-110875206 on Build GRCh38
rs368178349 Chr.6: 110875150-110875150 on Build GRCh38
rs758927681 Chr.6: 110875101-110875101 on Build GRCh38
rs780958784 Chr.6: 110875142-110875142 on Build GRCh38
rs199935766 Chr.6: 110875184-110875184 on Build GRCh38
rs374054924 Chr.6: 110875222-110875222 on Build GRCh38
rs1023807892 Chr.6: 110875294-110875294 on Build GRCh38

View Full Product Details