Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246851181-246851450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCTF1
Entrez Gene ID
25909
Gene Name
AT-hook containing transcription factor 1
Gene Aliases
ELYS, MST108, MSTP108, TMBS62
UniGene
Hs.300887
Gene Chromosome Location
Chr.1: 246839098-246931978 on Build GRCh38
Transcripts
NM_001323342.1 NM_001323343.1 NM_015446.4 NR_136586.1 XM_006711758.1 XM_011544156.2 XM_011544157.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5730133 c.4662G>A p.Q1554Q Chr.1: 246851344-246851344 on Build GRCh38
7354862 c.4723G>A p.E1575K Chr.1: 246851283-246851283 on Build GRCh38
5815563 c.4783A>T p.I1595L Chr.1: 246851223-246851223 on Build GRCh38
7354861 c.4750G>A p.E1584K Chr.1: 246851283-246851283 on Build GRCh38
6125487 c.4792G>T p.G1598C Chr.1: 246851214-246851214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754331331 Chr.1: 246851292-246851292 on Build GRCh38
rs757847395 Chr.1: 246851295-246851295 on Build GRCh38
rs750924356 Chr.1: 246851418-246851418 on Build GRCh38
rs372613414 Chr.1: 246851275-246851275 on Build GRCh38
rs140339459 Chr.1: 246851302-246851302 on Build GRCh38
rs138061071 Chr.1: 246851345-246851345 on Build GRCh38
rs750459242 Chr.1: 246851309-246851309 on Build GRCh38
rs773848576 Chr.1: 246851266-246851266 on Build GRCh38
rs375999683 Chr.1: 246851376-246851376 on Build GRCh38
rs758425751 Chr.1: 246851326-246851326 on Build GRCh38
rs773250325 Chr.1: 246851245-246851245 on Build GRCh38
rs966077470 Chr.1: 246851332-246851332 on Build GRCh38
rs749043981 Chr.1: 246851335-246851335 on Build GRCh38
rs770847163 Chr.1: 246851339-246851339 on Build GRCh38
rs747271989 Chr.1: 246851329-246851329 on Build GRCh38
rs201407224 Chr.1: 246851250-246851250 on Build GRCh38
rs373744452 Chr.1: 246851396-246851396 on Build GRCh38
rs768913266 Chr.1: 246851205-246851207 on Build GRCh38
rs149538294 Chr.1: 246851359-246851359 on Build GRCh38
rs775103193 Chr.1: 246851361-246851361 on Build GRCh38
rs765805897 Chr.1: 246851254-246851254 on Build GRCh38
rs775867832 Chr.1: 246851218-246851218 on Build GRCh38
rs759047411 Chr.1: 246851348-246851348 on Build GRCh38
rs766150171 Chr.1: 246851377-246851377 on Build GRCh38
rs1027296727 Chr.1: 246851296-246851296 on Build GRCh38
rs764492933 Chr.1: 246851285-246851285 on Build GRCh38
rs765721900 Chr.1: 246851417-246851417 on Build GRCh38
rs747899069 Chr.1: 246851227-246851227 on Build GRCh38
rs772269675 Chr.1: 246851212-246851212 on Build GRCh38
rs754132668 Chr.1: 246851394-246851394 on Build GRCh38
rs756440164 Chr.1: 246851278-246851278 on Build GRCh38
rs186771516 Chr.1: 246851330-246851330 on Build GRCh38
rs767153999 Chr.1: 246851273-246851273 on Build GRCh38
rs780379363 Chr.1: 246851328-246851328 on Build GRCh38
rs769738929 Chr.1: 246851239-246851239 on Build GRCh38
rs758986571 Chr.1: 246851267-246851267 on Build GRCh38

View Full Product Details