Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25729504-25729759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J
Entrez Gene ID
522
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6
Gene Aliases
ATP5, ATP5A, ATPM, CF6, F6
UniGene
Hs.246310
Gene Chromosome Location
Chr.21: 25724480-25735654 on Build GRCh38
Transcripts
NM_001003696.1 NM_001003697.1 NM_001003701.1 NM_001003703.1 NM_001320266.1 NM_001320267.1 NM_001685.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2841290 c.65G>T p.R22L Chr.21: 25729730-25729730 on Build GRCh38
4137198 c.161G>A p.R54Q Chr.21: 25729634-25729634 on Build GRCh38
3799814 c.138G>T p.K46N Chr.21: 25729657-25729657 on Build GRCh38
1153958 c.91G>A p.A31T Chr.21: 25729704-25729704 on Build GRCh38
3707990 c.37A>G p.I13V Chr.21: 25729758-25729758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3216073 Chr.21: 25729589-25729589 on Build GRCh38
rs397730336 Chr.21: 25729591-25729591 on Build GRCh38
rs771372468 Chr.21: 25729650-25729650 on Build GRCh38
rs748605598 Chr.21: 25729615-25729618 on Build GRCh38
rs1007985613 Chr.21: 25729695-25729695 on Build GRCh38
rs373840948 Chr.21: 25729666-25729666 on Build GRCh38
rs774671016 Chr.21: 25729654-25729654 on Build GRCh38
rs140201687 Chr.21: 25729731-25729731 on Build GRCh38
rs73338261 Chr.21: 25729703-25729703 on Build GRCh38
rs1010507611 Chr.21: 25729726-25729726 on Build GRCh38
rs752786714 Chr.21: 25729680-25729680 on Build GRCh38
rs1037185017 Chr.21: 25729611-25729611 on Build GRCh38
rs763191934 Chr.21: 25729635-25729635 on Build GRCh38
rs11539871 Chr.21: 25729732-25729732 on Build GRCh38
rs775176169 Chr.21: 25729609-25729609 on Build GRCh38
rs955957513 Chr.21: 25729723-25729723 on Build GRCh38
rs369621263 Chr.21: 25729730-25729730 on Build GRCh38
rs776490258 Chr.21: 25729631-25729631 on Build GRCh38
rs536059571 Chr.21: 25729685-25729685 on Build GRCh38
rs368172603 Chr.21: 25729697-25729697 on Build GRCh38
rs746509917 Chr.21: 25729727-25729727 on Build GRCh38
rs200421087 Chr.21: 25729715-25729715 on Build GRCh38
rs1019960803 Chr.21: 25729709-25729709 on Build GRCh38
rs898498345 Chr.21: 25729562-25729562 on Build GRCh38
rs764800829 Chr.21: 25729548-25729548 on Build GRCh38
rs567499162 Chr.21: 25729667-25729667 on Build GRCh38
rs758417035 Chr.21: 25729712-25729712 on Build GRCh38
rs764298477 Chr.21: 25729686-25729686 on Build GRCh38
rs149245672 Chr.21: 25729692-25729692 on Build GRCh38
rs771663153 Chr.21: 25729595-25729595 on Build GRCh38
rs746595654 Chr.21: 25729618-25729618 on Build GRCh38
rs931347634 Chr.21: 25729568-25729568 on Build GRCh38
rs200479849 Chr.21: 25729592-25729592 on Build GRCh38
rs1036997341 Chr.21: 25729542-25729542 on Build GRCh38
rs745868287 Chr.21: 25729586-25729586 on Build GRCh38
rs768407077 Chr.21: 25729624-25729624 on Build GRCh38
rs750292108 Chr.21: 25729707-25729707 on Build GRCh38

View Full Product Details