Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 98440998-98441247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM169B
Entrez Gene ID
283777
Gene Name
family with sequence similarity 169 member B
Gene Aliases
-
UniGene
Hs.668070
Gene Chromosome Location
Chr.15: 98437162-98514382 on Build GRCh38
Transcripts
NM_182562.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367136 c.451G>T p.G151C Chr.15: 98441079-98441079 on Build GRCh38
1375592 c.398G>A p.R133H Chr.15: 98441132-98441132 on Build GRCh38
4767947 c.390C>A p.D130E Chr.15: 98441140-98441140 on Build GRCh38
3887841 c.452G>A p.G151D Chr.15: 98441078-98441078 on Build GRCh38
3377706 c.401A>G p.H134R Chr.15: 98441129-98441129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000718810 Chr.15: 98441208-98441208 on Build GRCh38
rs747989617 Chr.15: 98441182-98441182 on Build GRCh38
rs755698112 Chr.15: 98441161-98441161 on Build GRCh38
rs534501126 Chr.15: 98441083-98441083 on Build GRCh38
rs768682656 Chr.15: 98441030-98441030 on Build GRCh38
rs772159781 Chr.15: 98441129-98441129 on Build GRCh38
rs776534742 Chr.15: 98441132-98441132 on Build GRCh38
rs761775106 Chr.15: 98441128-98441128 on Build GRCh38
rs762165545 Chr.15: 98441029-98441029 on Build GRCh38
rs76060825 Chr.15: 98441138-98441138 on Build GRCh38
rs754680341 Chr.15: 98441170-98441170 on Build GRCh38
rs753967197 Chr.15: 98441059-98441059 on Build GRCh38
rs567616878 Chr.15: 98441177-98441177 on Build GRCh38
rs942031578 Chr.15: 98441110-98441110 on Build GRCh38
rs147142052 Chr.15: 98441200-98441200 on Build GRCh38
rs774477515 Chr.15: 98441039-98441039 on Build GRCh38
rs779559628 Chr.15: 98441162-98441162 on Build GRCh38
rs773251446 Chr.15: 98441203-98441203 on Build GRCh38
rs769986835 Chr.15: 98441206-98441206 on Build GRCh38
rs72764391 Chr.15: 98441218-98441218 on Build GRCh38
rs201796033 Chr.15: 98441140-98441140 on Build GRCh38
rs73463586 Chr.15: 98441146-98441146 on Build GRCh38
rs749034620 Chr.15: 98441168-98441168 on Build GRCh38
rs775439911 Chr.15: 98441210-98441210 on Build GRCh38
rs200284374 Chr.15: 98441159-98441159 on Build GRCh38
rs749013237 Chr.15: 98441125-98441125 on Build GRCh38
rs745926964 Chr.15: 98441204-98441204 on Build GRCh38
rs149447808 Chr.15: 98441144-98441144 on Build GRCh38
rs562480555 Chr.15: 98441123-98441123 on Build GRCh38
rs372170534 Chr.15: 98441097-98441097 on Build GRCh38
rs965932698 Chr.15: 98441090-98441090 on Build GRCh38
rs75368525 Chr.15: 98441046-98441046 on Build GRCh38
rs545215241 Chr.15: 98441136-98441136 on Build GRCh38
rs200777104 Chr.15: 98441139-98441139 on Build GRCh38
rs201236619 Chr.15: 98441080-98441080 on Build GRCh38
rs979986425 Chr.15: 98441019-98441019 on Build GRCh38
rs745403056 Chr.15: 98441103-98441103 on Build GRCh38
rs117919150 Chr.15: 98441133-98441133 on Build GRCh38
rs778243872 Chr.15: 98441094-98441094 on Build GRCh38
rs758849987 Chr.15: 98441086-98441086 on Build GRCh38
rs769287001 Chr.15: 98441107-98441107 on Build GRCh38
rs992689976 Chr.15: 98441061-98441061 on Build GRCh38
rs780620011 Chr.15: 98441101-98441101 on Build GRCh38
rs759631036 Chr.15: 98441079-98441079 on Build GRCh38
rs555823411 Chr.15: 98441145-98441145 on Build GRCh38
rs766631242 Chr.15: 98441055-98441055 on Build GRCh38
rs774424137 Chr.15: 98441126-98441126 on Build GRCh38
rs762833234 Chr.15: 98441026-98441026 on Build GRCh38

View Full Product Details