Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117385585-117385850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871651 c.452C>T p.T151I Chr.7: 117385798-117385798 on Build GRCh38
3632268 c.540G>A p.E180E Chr.7: 117385710-117385710 on Build GRCh38
6606351 c.520G>A p.E174K Chr.7: 117385730-117385730 on Build GRCh38
300739 c.517G>A p.A173T Chr.7: 117385733-117385733 on Build GRCh38
1137618 c.463T>C p.Y155H Chr.7: 117385787-117385787 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746026562 Chr.7: 117385785-117385785 on Build GRCh38
rs556724584 Chr.7: 117385636-117385636 on Build GRCh38
rs771036618 Chr.7: 117385677-117385677 on Build GRCh38
rs753555024 Chr.7: 117385756-117385756 on Build GRCh38
rs759225056 Chr.7: 117385662-117385663 on Build GRCh38
rs370917484 Chr.7: 117385776-117385776 on Build GRCh38
rs374156881 Chr.7: 117385781-117385781 on Build GRCh38
rs138855550 Chr.7: 117385799-117385799 on Build GRCh38
rs374686580 Chr.7: 117385678-117385678 on Build GRCh38
rs778623890 Chr.7: 117385768-117385768 on Build GRCh38
rs753361798 Chr.7: 117385823-117385823 on Build GRCh38
rs142644843 Chr.7: 117385731-117385731 on Build GRCh38
rs760286066 Chr.7: 117385754-117385754 on Build GRCh38
rs199851030 Chr.7: 117385796-117385796 on Build GRCh38
rs373779367 Chr.7: 117385811-117385811 on Build GRCh38
rs888647019 Chr.7: 117385621-117385621 on Build GRCh38
rs749871762 Chr.7: 117385749-117385749 on Build GRCh38
rs377493865 Chr.7: 117385739-117385739 on Build GRCh38
rs202215395 Chr.7: 117385817-117385817 on Build GRCh38
rs754630010 Chr.7: 117385760-117385760 on Build GRCh38
rs769025569 Chr.7: 117385801-117385801 on Build GRCh38
rs770212168 Chr.7: 117385710-117385710 on Build GRCh38
rs773124338 Chr.7: 117385674-117385674 on Build GRCh38
rs774985013 Chr.7: 117385810-117385810 on Build GRCh38
rs752561261 Chr.7: 117385770-117385770 on Build GRCh38
rs747046585 Chr.7: 117385778-117385778 on Build GRCh38
rs781421889 Chr.7: 117385784-117385784 on Build GRCh38
rs772061186 Chr.7: 117385655-117385655 on Build GRCh38
rs776008592 Chr.7: 117385722-117385722 on Build GRCh38
rs766005625 Chr.7: 117385755-117385755 on Build GRCh38
rs764972373 Chr.7: 117385674-117385674 on Build GRCh38
rs1027996728 Chr.7: 117385682-117385682 on Build GRCh38
rs186384831 Chr.7: 117385790-117385790 on Build GRCh38
rs759925070 Chr.7: 117385659-117385659 on Build GRCh38
rs575161282 Chr.7: 117385639-117385639 on Build GRCh38
rs747150671 Chr.7: 117385676-117385676 on Build GRCh38
rs777814813 Chr.7: 117385777-117385777 on Build GRCh38
rs759971835 Chr.7: 117385698-117385698 on Build GRCh38
rs753770290 Chr.7: 117385805-117385805 on Build GRCh38

View Full Product Details