Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15010824-15011061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC105
Entrez Gene ID
126402
Gene Name
coiled-coil domain containing 105
Gene Aliases
-
UniGene
Hs.375985
Gene Chromosome Location
Chr.19: 15010727-15023271 on Build GRCh38
Transcripts
NM_173482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425672 c.46G>A p.V16I Chr.19: 15010871-15010871 on Build GRCh38
5639217 c.214C>A p.R72S Chr.19: 15011039-15011039 on Build GRCh38
4669651 c.59C>T p.A20V Chr.19: 15010884-15010884 on Build GRCh38
7278561 c.45C>T p.R15R Chr.19: 15010870-15010870 on Build GRCh38
5684648 c.208C>A p.P70T Chr.19: 15011033-15011033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779977866 Chr.19: 15011005-15011005 on Build GRCh38
rs77764676 Chr.19: 15010997-15010997 on Build GRCh38
rs951307558 Chr.19: 15010893-15010893 on Build GRCh38
rs756715181 Chr.19: 15010973-15010973 on Build GRCh38
rs776259940 Chr.19: 15010892-15010892 on Build GRCh38
rs768752309 Chr.19: 15011008-15011008 on Build GRCh38
rs778627656 Chr.19: 15010871-15010871 on Build GRCh38
rs773421461 Chr.19: 15011025-15011025 on Build GRCh38
rs774261249 Chr.19: 15011009-15011009 on Build GRCh38
rs1055960767 Chr.19: 15011006-15011006 on Build GRCh38
rs935423780 Chr.19: 15010846-15010846 on Build GRCh38
rs775614545 Chr.19: 15010917-15010917 on Build GRCh38
rs746595027 Chr.19: 15010889-15010889 on Build GRCh38
rs975723757 Chr.19: 15010951-15010951 on Build GRCh38
rs761899374 Chr.19: 15010934-15010934 on Build GRCh38
rs116867154 Chr.19: 15010848-15010848 on Build GRCh38
rs150456275 Chr.19: 15010933-15010933 on Build GRCh38
rs756543809 Chr.19: 15010869-15010869 on Build GRCh38
rs80121862 Chr.19: 15010890-15010890 on Build GRCh38
rs769705952 Chr.19: 15010902-15010902 on Build GRCh38
rs909726800 Chr.19: 15010996-15010996 on Build GRCh38
rs767960145 Chr.19: 15010941-15010941 on Build GRCh38
rs772183332 Chr.19: 15011020-15011020 on Build GRCh38
rs577782567 Chr.19: 15010878-15010879 on Build GRCh38
rs980046072 Chr.19: 15010995-15010995 on Build GRCh38
rs750727974 Chr.19: 15010942-15010942 on Build GRCh38
rs772720021 Chr.19: 15010943-15010943 on Build GRCh38
rs866314123 Chr.19: 15010878-15010878 on Build GRCh38
rs142952980 Chr.19: 15011012-15011012 on Build GRCh38
rs777584376 Chr.19: 15010886-15010886 on Build GRCh38
rs781099838 Chr.19: 15010994-15010994 on Build GRCh38
rs751267551 Chr.19: 15010988-15010988 on Build GRCh38
rs777380028 Chr.19: 15010982-15010982 on Build GRCh38
rs372938359 Chr.19: 15010978-15010978 on Build GRCh38
rs750966828 Chr.19: 15010864-15010864 on Build GRCh38
rs756837621 Chr.19: 15010991-15010991 on Build GRCh38
rs747660694 Chr.19: 15010872-15010872 on Build GRCh38
rs560857516 Chr.19: 15010844-15010844 on Build GRCh38
rs760962966 Chr.19: 15011040-15011040 on Build GRCh38
rs763022894 Chr.19: 15010918-15010918 on Build GRCh38
rs767722737 Chr.19: 15010854-15010854 on Build GRCh38
rs767002210 Chr.19: 15010974-15010974 on Build GRCh38
rs749173220 Chr.19: 15011007-15011007 on Build GRCh38
rs865932399 Chr.19: 15010898-15010898 on Build GRCh38
rs200591174 Chr.19: 15010880-15010880 on Build GRCh38
rs138297206 Chr.19: 15010981-15010981 on Build GRCh38
rs764101880 Chr.19: 15010923-15010923 on Build GRCh38

View Full Product Details