Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38914848-38915121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A1
Entrez Gene ID
92689
Gene Name
family with sequence similarity 114 member A1
Gene Aliases
Noxp20
UniGene
Hs.476517
Gene Chromosome Location
Chr.4: 38867733-38945744 on Build GRCh38
Transcripts
NM_138389.2 NR_033290.1 XM_005262672.2 XM_006714033.2 XM_017008838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6290408 c.916G>T p.E306* Chr.4: 38915044-38915044 on Build GRCh38
329345 c.874C>A p.L292M Chr.4: 38915002-38915002 on Build GRCh38
7249872 c.882T>C p.D294D Chr.4: 38915010-38915010 on Build GRCh38
3775822 c.854G>C p.R285T Chr.4: 38914982-38914982 on Build GRCh38
7221518 c.861G>A p.A287A Chr.4: 38914989-38914989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759885327 Chr.4: 38914998-38914998 on Build GRCh38
rs200754144 Chr.4: 38914872-38914872 on Build GRCh38
rs750031226 Chr.4: 38914893-38914893 on Build GRCh38
rs778109875 Chr.4: 38914971-38914971 on Build GRCh38
rs780000661 Chr.4: 38915074-38915074 on Build GRCh38
rs200864225 Chr.4: 38914890-38914890 on Build GRCh38
rs754130990 Chr.4: 38914996-38914996 on Build GRCh38
rs371512230 Chr.4: 38914999-38914999 on Build GRCh38
rs199646586 Chr.4: 38915064-38915064 on Build GRCh38
rs370743217 Chr.4: 38914886-38914886 on Build GRCh38
rs372301008 Chr.4: 38914987-38914987 on Build GRCh38
rs367645644 Chr.4: 38914871-38914871 on Build GRCh38
rs748291760 Chr.4: 38914993-38914993 on Build GRCh38
rs766612147 Chr.4: 38914922-38914922 on Build GRCh38
rs993650917 Chr.4: 38915057-38915057 on Build GRCh38
rs867912315 Chr.4: 38915019-38915019 on Build GRCh38
rs752836149 Chr.4: 38914967-38914967 on Build GRCh38
rs778965484 Chr.4: 38914964-38914964 on Build GRCh38
rs1008540813 Chr.4: 38915000-38915000 on Build GRCh38
rs368885217 Chr.4: 38915090-38915090 on Build GRCh38
rs768649787 Chr.4: 38914986-38914986 on Build GRCh38
rs183070525 Chr.4: 38914981-38914981 on Build GRCh38
rs200183888 Chr.4: 38914980-38914980 on Build GRCh38
rs769458516 Chr.4: 38914870-38914870 on Build GRCh38
rs753119108 Chr.4: 38915015-38915015 on Build GRCh38
rs754363682 Chr.4: 38914952-38914952 on Build GRCh38
rs768559895 Chr.4: 38915091-38915091 on Build GRCh38
rs761378449 Chr.4: 38914888-38914888 on Build GRCh38
rs149833714 Chr.4: 38914995-38914995 on Build GRCh38
rs779867184 Chr.4: 38914943-38914945 on Build GRCh38
rs757367796 Chr.4: 38915071-38915071 on Build GRCh38
rs868480455 Chr.4: 38914991-38914993 on Build GRCh38
rs769664563 Chr.4: 38914974-38914974 on Build GRCh38
rs755418170 Chr.4: 38914963-38914963 on Build GRCh38
rs770599161 Chr.4: 38914973-38914973 on Build GRCh38
rs758391021 Chr.4: 38914968-38914968 on Build GRCh38
rs143144596 Chr.4: 38915065-38915065 on Build GRCh38
rs562870312 Chr.4: 38914879-38914879 on Build GRCh38
rs4261994 Chr.4: 38915010-38915010 on Build GRCh38
rs772948767 Chr.4: 38914991-38914991 on Build GRCh38
rs761140777 Chr.4: 38914909-38914909 on Build GRCh38
rs758548831 Chr.4: 38915043-38915043 on Build GRCh38
rs960007701 Chr.4: 38914976-38914976 on Build GRCh38
rs897597616 Chr.4: 38915016-38915016 on Build GRCh38
rs761445177 Chr.4: 38914988-38914988 on Build GRCh38
rs760195122 Chr.4: 38914993-38914993 on Build GRCh38
rs117679360 Chr.4: 38914989-38914989 on Build GRCh38

View Full Product Details