Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14909158-14909383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLRE1C
Entrez Gene ID
64421
Gene Name
DNA cross-link repair 1C
Gene Aliases
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
UniGene
Hs.655932
Gene Chromosome Location
Chr.10: 14897853-14954432 on Build GRCh38
Transcripts
NM_001033855.2 NM_001033857.2 NM_001033858.2 NM_001289076.1 NM_001289077.1 NM_001289078.1 NM_001289079.1 NM_022487.3 NR_110297.1 XM_006717491.3 XM_011519616.1 XM_011519617.1 XM_011519619.1 XM_017016556.1 XM_017016557.1 XM_017016558.1 XM_017016559.1 XM_017016560.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012985 c.868G>A p.V290I Chr.10: 14909274-14909274 on Build GRCh38
1203141 c.833T>G p.F278C Chr.10: 14909309-14909309 on Build GRCh38
5999220 c.913G>A p.A305T Chr.10: 14909229-14909229 on Build GRCh38
1203144 c.1186C>A p.P396T Chr.10: 14909301-14909301 on Build GRCh38
6569201 c.1157-5C>T p.? Chr.10: 14909335-14909335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926152401 Chr.10: 14909283-14909283 on Build GRCh38
rs772982087 Chr.10: 14909264-14909264 on Build GRCh38
rs921819288 Chr.10: 14909184-14909184 on Build GRCh38
rs752292924 Chr.10: 14909302-14909302 on Build GRCh38
rs755447150 Chr.10: 14909220-14909221 on Build GRCh38
rs759039414 Chr.10: 14909292-14909292 on Build GRCh38
rs532892332 Chr.10: 14909263-14909263 on Build GRCh38
rs113870881 Chr.10: 14909203-14909203 on Build GRCh38
rs1043126231 Chr.10: 14909269-14909269 on Build GRCh38
rs748479870 Chr.10: 14909346-14909347 on Build GRCh38
rs768647014 Chr.10: 14909248-14909248 on Build GRCh38
rs749319265 Chr.10: 14909247-14909247 on Build GRCh38
rs971914398 Chr.10: 14909258-14909258 on Build GRCh38
rs1023122829 Chr.10: 14909253-14909253 on Build GRCh38
rs764820306 Chr.10: 14909295-14909295 on Build GRCh38
rs557790274 Chr.10: 14909318-14909318 on Build GRCh38
rs746284396 Chr.10: 14909219-14909219 on Build GRCh38
rs765990122 Chr.10: 14909284-14909284 on Build GRCh38
rs954715514 Chr.10: 14909191-14909191 on Build GRCh38
rs115081573 Chr.10: 14909200-14909200 on Build GRCh38
rs1013537366 Chr.10: 14909238-14909238 on Build GRCh38
rs752137206 Chr.10: 14909202-14909202 on Build GRCh38
rs114279611 Chr.10: 14909288-14909288 on Build GRCh38
rs140194645 Chr.10: 14909211-14909211 on Build GRCh38
rs569397454 Chr.10: 14909268-14909268 on Build GRCh38
rs779415949 Chr.10: 14909317-14909318 on Build GRCh38
rs753332017 Chr.10: 14909181-14909181 on Build GRCh38
rs780280112 Chr.10: 14909328-14909328 on Build GRCh38
rs770043960 Chr.10: 14909226-14909226 on Build GRCh38
rs774222216 Chr.10: 14909249-14909249 on Build GRCh38
rs780148041 Chr.10: 14909243-14909243 on Build GRCh38
rs750741791 Chr.10: 14909319-14909319 on Build GRCh38
rs761710395 Chr.10: 14909254-14909254 on Build GRCh38
rs61755347 Chr.10: 14909313-14909313 on Build GRCh38
rs148509969 Chr.10: 14909195-14909195 on Build GRCh38

View Full Product Details