Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36598866-36599123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS11
Entrez Gene ID
79154
Gene Name
dehydrogenase/reductase 11
Gene Aliases
ARPG836, SDR24C1, spDHRS11
UniGene
Hs.462859
Gene Chromosome Location
Chr.17: 36591791-36600804 on Build GRCh38
Transcripts
NM_024308.3 XM_005257658.2 XM_011525233.2

SNP Detail

SNP ID Chromosome Location
rs968169781 Chr.17: 36599034-36599034 on Build GRCh38
rs550229634 Chr.17: 36598935-36598935 on Build GRCh38
rs780816334 Chr.17: 36599050-36599050 on Build GRCh38
rs757572328 Chr.17: 36598923-36598923 on Build GRCh38
rs774883923 Chr.17: 36599001-36599001 on Build GRCh38
rs750548733 Chr.17: 36598934-36598934 on Build GRCh38
rs767660664 Chr.17: 36598928-36598928 on Build GRCh38
rs566647119 Chr.17: 36598985-36598985 on Build GRCh38
rs186860451 Chr.17: 36598902-36598902 on Build GRCh38
rs764591361 Chr.17: 36598915-36598915 on Build GRCh38
rs749758859 Chr.17: 36599069-36599069 on Build GRCh38
rs140626590 Chr.17: 36598940-36598940 on Build GRCh38
rs752633188 Chr.17: 36599022-36599022 on Build GRCh38
rs996061794 Chr.17: 36598991-36598991 on Build GRCh38
rs750681184 Chr.17: 36599017-36599017 on Build GRCh38
rs777568391 Chr.17: 36598956-36598956 on Build GRCh38
rs746625289 Chr.17: 36598966-36598966 on Build GRCh38
rs535830379 Chr.17: 36598949-36598949 on Build GRCh38
rs769287307 Chr.17: 36599088-36599088 on Build GRCh38
rs751369244 Chr.17: 36599043-36599043 on Build GRCh38
rs374687848 Chr.17: 36599006-36599006 on Build GRCh38
rs200684120 Chr.17: 36599011-36599011 on Build GRCh38
rs772607977 Chr.17: 36599099-36599099 on Build GRCh38
rs770574452 Chr.17: 36598980-36598980 on Build GRCh38
rs370398386 Chr.17: 36598984-36598984 on Build GRCh38
rs879068579 Chr.17: 36599002-36599002 on Build GRCh38
rs145372121 Chr.17: 36599049-36599049 on Build GRCh38
rs759126695 Chr.17: 36598946-36598947 on Build GRCh38
rs758364882 Chr.17: 36599039-36599039 on Build GRCh38
rs762294067 Chr.17: 36599088-36599089 on Build GRCh38
rs766421237 Chr.17: 36599021-36599021 on Build GRCh38
rs762055507 Chr.17: 36599008-36599008 on Build GRCh38
rs775918205 Chr.17: 36598904-36598904 on Build GRCh38
rs777478465 Chr.17: 36599042-36599042 on Build GRCh38
rs763369739 Chr.17: 36598907-36598907 on Build GRCh38
rs553233261 Chr.17: 36598950-36598950 on Build GRCh38
rs762267832 Chr.17: 36599098-36599098 on Build GRCh38
rs765787069 Chr.17: 36598903-36598903 on Build GRCh38
rs377050445 Chr.17: 36598917-36598917 on Build GRCh38
rs769323176 Chr.17: 36599059-36599059 on Build GRCh38
rs769026210 Chr.17: 36598947-36598947 on Build GRCh38
rs745545059 Chr.17: 36599054-36599054 on Build GRCh38
rs763416991 Chr.17: 36598992-36598992 on Build GRCh38
rs367726729 Chr.17: 36599089-36599089 on Build GRCh38
rs995337160 Chr.17: 36598889-36598889 on Build GRCh38
rs773971041 Chr.17: 36598906-36598906 on Build GRCh38
rs764501467 Chr.17: 36598995-36598995 on Build GRCh38
rs889905225 Chr.17: 36598993-36598993 on Build GRCh38
rs371943922 Chr.17: 36599100-36599100 on Build GRCh38
rs760780032 Chr.17: 36599018-36599018 on Build GRCh38
rs779449405 Chr.17: 36599061-36599061 on Build GRCh38
rs776967857 Chr.17: 36599081-36599081 on Build GRCh38
rs150477741 Chr.17: 36598944-36598944 on Build GRCh38
rs776060583 Chr.17: 36598981-36598981 on Build GRCh38

View Full Product Details