Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12999270-12999507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP192
Entrez Gene ID
55125
Gene Name
centrosomal protein 192
Gene Aliases
PPP1R62
UniGene
Hs.100914
Gene Chromosome Location
Chr.18: 12991307-13125053 on Build GRCh38
Transcripts
NM_032142.3 XM_005258107.3 XM_005258109.3 XM_006722326.3 XM_006722327.3 XM_006722330.3 XM_011525673.2 XM_017025803.1 XM_017025804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387820 c.13C>T p.R5* Chr.18: 12999437-12999437 on Build GRCh38
5528571 c.51C>T p.T17T Chr.18: 12999475-12999475 on Build GRCh38
7360879 c.37C>T p.P13S Chr.18: 12999461-12999461 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757372725 Chr.18: 12999387-12999387 on Build GRCh38
rs897379224 Chr.18: 12999335-12999335 on Build GRCh38
rs553449095 Chr.18: 12999353-12999353 on Build GRCh38
rs1024828407 Chr.18: 12999458-12999458 on Build GRCh38
rs776888796 Chr.18: 12999306-12999306 on Build GRCh38
rs748072561 Chr.18: 12999451-12999451 on Build GRCh38
rs572021521 Chr.18: 12999389-12999389 on Build GRCh38
rs1037312884 Chr.18: 12999330-12999330 on Build GRCh38
rs180793650 Chr.18: 12999444-12999444 on Build GRCh38
rs746120392 Chr.18: 12999437-12999437 on Build GRCh38
rs771835959 Chr.18: 12999461-12999461 on Build GRCh38
rs998579864 Chr.18: 12999380-12999380 on Build GRCh38
rs759914101 Chr.18: 12999434-12999434 on Build GRCh38
rs151161485 Chr.18: 12999466-12999466 on Build GRCh38
rs985594523 Chr.18: 12999308-12999309 on Build GRCh38
rs931366394 Chr.18: 12999336-12999336 on Build GRCh38
rs904708452 Chr.18: 12999315-12999315 on Build GRCh38
rs1012315557 Chr.18: 12999457-12999457 on Build GRCh38
rs1031714514 Chr.18: 12999326-12999326 on Build GRCh38
rs746997670 Chr.18: 12999382-12999382 on Build GRCh38
rs768408870 Chr.18: 12999438-12999438 on Build GRCh38
rs999013560 Chr.18: 12999316-12999316 on Build GRCh38
rs538926286 Chr.18: 12999309-12999309 on Build GRCh38

View Full Product Details