Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69336476-69336749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG8
Entrez Gene ID
84342
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
CDG2H, DOR1
UniGene
Hs.130849
Gene Chromosome Location
Chr.16: 69328621-69339623 on Build GRCh38
Transcripts
NM_032382.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160109 c.478C>T p.Q160* Chr.16: 69336612-69336612 on Build GRCh38
7187377 c.540C>T p.Y180Y Chr.16: 69336550-69336550 on Build GRCh38
5851318 c.397G>A p.E133K Chr.16: 69336693-69336693 on Build GRCh38
7098647 c.523C>G p.L175V Chr.16: 69336567-69336567 on Build GRCh38
3730548 c.492C>A p.T164T Chr.16: 69336598-69336598 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770925117 Chr.16: 69336655-69336655 on Build GRCh38
rs764627647 Chr.16: 69336599-69336599 on Build GRCh38
rs762499740 Chr.16: 69336674-69336674 on Build GRCh38
rs536433624 Chr.16: 69336699-69336699 on Build GRCh38
rs567137330 Chr.16: 69336546-69336546 on Build GRCh38
rs764884654 Chr.16: 69336590-69336590 on Build GRCh38
rs139342248 Chr.16: 69336705-69336705 on Build GRCh38
rs764165010 Chr.16: 69336700-69336700 on Build GRCh38
rs199726114 Chr.16: 69336516-69336516 on Build GRCh38
rs762521780 Chr.16: 69336596-69336596 on Build GRCh38
rs187905134 Chr.16: 69336565-69336565 on Build GRCh38
rs746103938 Chr.16: 69336557-69336557 on Build GRCh38
rs752183309 Chr.16: 69336542-69336542 on Build GRCh38
rs746971487 Chr.16: 69336653-69336653 on Build GRCh38
rs780005639 Chr.16: 69336567-69336567 on Build GRCh38
rs369750195 Chr.16: 69336593-69336593 on Build GRCh38
rs756306527 Chr.16: 69336612-69336612 on Build GRCh38
rs781443187 Chr.16: 69336550-69336550 on Build GRCh38
rs757713000 Chr.16: 69336605-69336605 on Build GRCh38
rs771846065 Chr.16: 69336688-69336690 on Build GRCh38
rs375728590 Chr.16: 69336671-69336671 on Build GRCh38
rs770868214 Chr.16: 69336580-69336580 on Build GRCh38
rs760538440 Chr.16: 69336499-69336499 on Build GRCh38
rs780312851 Chr.16: 69336614-69336614 on Build GRCh38
rs141717174 Chr.16: 69336579-69336579 on Build GRCh38
rs906744517 Chr.16: 69336725-69336727 on Build GRCh38
rs748442815 Chr.16: 69336608-69336608 on Build GRCh38
rs773250434 Chr.16: 69336577-69336577 on Build GRCh38
rs775179371 Chr.16: 69336672-69336672 on Build GRCh38
rs751032320 Chr.16: 69336677-69336677 on Build GRCh38
rs966313565 Chr.16: 69336647-69336647 on Build GRCh38
rs968235814 Chr.16: 69336665-69336665 on Build GRCh38
rs562484375 Chr.16: 69336497-69336497 on Build GRCh38
rs764684169 Chr.16: 69336536-69336536 on Build GRCh38
rs974974225 Chr.16: 69336648-69336648 on Build GRCh38
rs201091389 Chr.16: 69336589-69336589 on Build GRCh38
rs146248068 Chr.16: 69336704-69336704 on Build GRCh38
rs573017898 Chr.16: 69336706-69336706 on Build GRCh38
rs568491704 Chr.16: 69336685-69336685 on Build GRCh38
rs767956895 Chr.16: 69336684-69336684 on Build GRCh38
rs372270200 Chr.16: 69336664-69336664 on Build GRCh38
rs375227294 Chr.16: 69336568-69336568 on Build GRCh38
rs754606895 Chr.16: 69336532-69336532 on Build GRCh38
rs781571851 Chr.16: 69336606-69336606 on Build GRCh38
rs766556659 Chr.16: 69336694-69336694 on Build GRCh38
rs552924056 Chr.16: 69336581-69336581 on Build GRCh38
rs776301238 Chr.16: 69336662-69336662 on Build GRCh38
rs756467489 Chr.16: 69336693-69336693 on Build GRCh38
rs201787629 Chr.16: 69336675-69336675 on Build GRCh38
rs138647246 Chr.16: 69336512-69336512 on Build GRCh38
rs757320339 Chr.16: 69336713-69336713 on Build GRCh38
rs777969581 Chr.16: 69336718-69336718 on Build GRCh38
rs753514529 Chr.16: 69336529-69336529 on Build GRCh38
rs369001100 Chr.16: 69336680-69336680 on Build GRCh38
rs201289118 Chr.16: 69336543-69336543 on Build GRCh38
rs753893688 Chr.16: 69336710-69336710 on Build GRCh38
rs757588056 Chr.16: 69336549-69336549 on Build GRCh38
rs548526704 Chr.16: 69336545-69336545 on Build GRCh38
rs759115422 Chr.16: 69336584-69336584 on Build GRCh38
rs200225054 Chr.16: 69336639-69336639 on Build GRCh38
rs768831779 Chr.16: 69336634-69336634 on Build GRCh38
rs991797535 Chr.16: 69336709-69336709 on Build GRCh38
rs749363867 Chr.16: 69336626-69336626 on Build GRCh38
rs543723269 Chr.16: 69336650-69336650 on Build GRCh38

View Full Product Details