Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 443305-443576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM101B
Entrez Gene ID
359845
Gene Name
family with sequence similarity 101 member B
Gene Aliases
CFM1
UniGene
Hs.345588
Gene Chromosome Location
Chr.17: 439978-445940 on Build GRCh38
Transcripts
NM_182705.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
705689 c.268C>G p.P90A Chr.17: 443331-443331 on Build GRCh38
3179515 c.259C>T p.R87C Chr.17: 443340-443340 on Build GRCh38
3179519 c.227G>A p.R76H Chr.17: 443372-443372 on Build GRCh38
7096857 c.144C>A p.F48L Chr.17: 443455-443455 on Build GRCh38
7225378 c.116T>C p.L39S Chr.17: 443483-443483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782771305 Chr.17: 443430-443430 on Build GRCh38
rs781865189 Chr.17: 443433-443433 on Build GRCh38
rs369481196 Chr.17: 443518-443518 on Build GRCh38
rs370609692 Chr.17: 443358-443358 on Build GRCh38
rs782285828 Chr.17: 443470-443470 on Build GRCh38
rs963800067 Chr.17: 443391-443391 on Build GRCh38
rs781985104 Chr.17: 443402-443402 on Build GRCh38
rs935784493 Chr.17: 443472-443472 on Build GRCh38
rs374226782 Chr.17: 443340-443340 on Build GRCh38
rs782216464 Chr.17: 443393-443393 on Build GRCh38
rs782782619 Chr.17: 443500-443500 on Build GRCh38
rs541436356 Chr.17: 443519-443519 on Build GRCh38
rs782031541 Chr.17: 443419-443419 on Build GRCh38
rs782097171 Chr.17: 443345-443345 on Build GRCh38
rs532003276 Chr.17: 443556-443556 on Build GRCh38
rs782767576 Chr.17: 443346-443346 on Build GRCh38
rs782610327 Chr.17: 443452-443452 on Build GRCh38
rs372687153 Chr.17: 443509-443509 on Build GRCh38
rs781839209 Chr.17: 443508-443508 on Build GRCh38
rs368788591 Chr.17: 443401-443401 on Build GRCh38
rs782131752 Chr.17: 443404-443404 on Build GRCh38
rs571620470 Chr.17: 443329-443329 on Build GRCh38
rs8069166 Chr.17: 443347-443347 on Build GRCh38
rs912160216 Chr.17: 443428-443428 on Build GRCh38
rs782792365 Chr.17: 443413-443413 on Build GRCh38
rs562629922 Chr.17: 443504-443504 on Build GRCh38
rs781889947 Chr.17: 443510-443510 on Build GRCh38
rs781967796 Chr.17: 443325-443325 on Build GRCh38
rs782220939 Chr.17: 443368-443368 on Build GRCh38
rs78050935 Chr.17: 443469-443469 on Build GRCh38
rs782057242 Chr.17: 443338-443338 on Build GRCh38
rs782050755 Chr.17: 443421-443421 on Build GRCh38
rs782480143 Chr.17: 443451-443451 on Build GRCh38
rs782472948 Chr.17: 443423-443423 on Build GRCh38
rs782465699 Chr.17: 443360-443360 on Build GRCh38
rs782713598 Chr.17: 443339-443339 on Build GRCh38
rs138567082 Chr.17: 443550-443550 on Build GRCh38
rs368165658 Chr.17: 443367-443367 on Build GRCh38
rs117842563 Chr.17: 443384-443384 on Build GRCh38
rs371785121 Chr.17: 443541-443541 on Build GRCh38
rs76404864 Chr.17: 443483-443483 on Build GRCh38
rs953352313 Chr.17: 443527-443527 on Build GRCh38
rs375207031 Chr.17: 443517-443517 on Build GRCh38
rs550861769 Chr.17: 443372-443372 on Build GRCh38
rs372868305 Chr.17: 443377-443377 on Build GRCh38
rs782249233 Chr.17: 443466-443466 on Build GRCh38
rs921978372 Chr.17: 443407-443407 on Build GRCh38
rs781983088 Chr.17: 443489-443489 on Build GRCh38
rs181976961 Chr.17: 443467-443467 on Build GRCh38
rs782244999 Chr.17: 443528-443528 on Build GRCh38
rs782388184 Chr.17: 443540-443540 on Build GRCh38
rs782557368 Chr.17: 443445-443445 on Build GRCh38
rs533993075 Chr.17: 443473-443473 on Build GRCh38
rs782127225 Chr.17: 443490-443490 on Build GRCh38
rs782575478 Chr.17: 443446-443446 on Build GRCh38
rs113505750 Chr.17: 443520-443520 on Build GRCh38
rs185634910 Chr.17: 443501-443501 on Build GRCh38
rs201027561 Chr.17: 443359-443359 on Build GRCh38
rs782532789 Chr.17: 443351-443351 on Build GRCh38
rs782359007 Chr.17: 443485-443485 on Build GRCh38
rs782050400 Chr.17: 443544-443544 on Build GRCh38
rs782620316 Chr.17: 443526-443526 on Build GRCh38
rs782411833 Chr.17: 443326-443326 on Build GRCh38
rs536237828 Chr.17: 443378-443378 on Build GRCh38
rs782206933 Chr.17: 443448-443448 on Build GRCh38
rs371491730 Chr.17: 443479-443479 on Build GRCh38
rs894394971 Chr.17: 443352-443352 on Build GRCh38
rs782646185 Chr.17: 443373-443373 on Build GRCh38

View Full Product Details