Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3639894-3640147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART5
Entrez Gene ID
116969
Gene Name
ADP-ribosyltransferase 5
Gene Aliases
ARTC5
UniGene
Hs.125680
Gene Chromosome Location
Chr.11: 3638503-3642392 on Build GRCh38
Transcripts
NM_001079536.1 NM_001297668.1 NM_053017.4 NR_123732.1 XM_005252779.3 XM_006718136.3 XM_011519879.2 XM_017017171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259867 c.341T>C p.V114A Chr.11: 3640088-3640088 on Build GRCh38
3447075 c.529C>T p.R177C Chr.11: 3639900-3639900 on Build GRCh38
926746 c.433C>T p.R145* Chr.11: 3639996-3639996 on Build GRCh38
3869122 c.458G>A p.G153E Chr.11: 3639971-3639971 on Build GRCh38
3447079 c.369C>T p.L123L Chr.11: 3640060-3640060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61746063 Chr.11: 3639963-3639963 on Build GRCh38
rs191937078 Chr.11: 3640072-3640072 on Build GRCh38
rs200406523 Chr.11: 3640078-3640078 on Build GRCh38
rs749039512 Chr.11: 3639950-3639950 on Build GRCh38
rs770291984 Chr.11: 3639982-3639982 on Build GRCh38
rs750561021 Chr.11: 3640004-3640004 on Build GRCh38
rs754739993 Chr.11: 3639951-3639951 on Build GRCh38
rs774546425 Chr.11: 3639995-3639995 on Build GRCh38
rs751867051 Chr.11: 3639931-3639931 on Build GRCh38
rs775919547 Chr.11: 3639983-3639983 on Build GRCh38
rs769133129 Chr.11: 3639989-3639989 on Build GRCh38
rs147444671 Chr.11: 3640014-3640014 on Build GRCh38
rs752787959 Chr.11: 3640029-3640029 on Build GRCh38
rs772424596 Chr.11: 3640067-3640067 on Build GRCh38
rs757415100 Chr.11: 3639932-3639932 on Build GRCh38
rs749840441 Chr.11: 3640045-3640045 on Build GRCh38
rs746486084 Chr.11: 3639972-3639972 on Build GRCh38
rs746537106 Chr.11: 3640039-3640039 on Build GRCh38
rs776431527 Chr.11: 3640038-3640052 on Build GRCh38
rs762042736 Chr.11: 3640056-3640056 on Build GRCh38
rs574265854 Chr.11: 3640007-3640007 on Build GRCh38
rs758082696 Chr.11: 3640020-3640020 on Build GRCh38
rs1009474793 Chr.11: 3639947-3639947 on Build GRCh38
rs773506388 Chr.11: 3640068-3640068 on Build GRCh38
rs373165519 Chr.11: 3640090-3640090 on Build GRCh38
rs61747703 Chr.11: 3640089-3640089 on Build GRCh38
rs779219965 Chr.11: 3640120-3640120 on Build GRCh38
rs544698668 Chr.11: 3640047-3640047 on Build GRCh38
rs772733690 Chr.11: 3639971-3639971 on Build GRCh38
rs762141846 Chr.11: 3639996-3639996 on Build GRCh38
rs770488380 Chr.11: 3640041-3640041 on Build GRCh38
rs763357904 Chr.11: 3639986-3639986 on Build GRCh38
rs760609896 Chr.11: 3639938-3639939 on Build GRCh38
rs754710318 Chr.11: 3640058-3640058 on Build GRCh38
rs748533200 Chr.11: 3640121-3640121 on Build GRCh38
rs938674699 Chr.11: 3640021-3640021 on Build GRCh38
rs777531507 Chr.11: 3640087-3640087 on Build GRCh38
rs1049504292 Chr.11: 3640081-3640081 on Build GRCh38
rs767799360 Chr.11: 3640001-3640001 on Build GRCh38
rs138160202 Chr.11: 3640082-3640082 on Build GRCh38
rs767744521 Chr.11: 3639933-3639933 on Build GRCh38
rs753868670 Chr.11: 3640011-3640011 on Build GRCh38
rs770653508 Chr.11: 3639957-3639962 on Build GRCh38
rs777343930 Chr.11: 3640035-3640035 on Build GRCh38
rs758132085 Chr.11: 3640086-3640086 on Build GRCh38
rs376205015 Chr.11: 3640111-3640111 on Build GRCh38
rs572893155 Chr.11: 3639942-3639942 on Build GRCh38
rs780564029 Chr.11: 3640044-3640044 on Build GRCh38
rs560195676 Chr.11: 3640085-3640085 on Build GRCh38
rs143327826 Chr.11: 3640013-3640013 on Build GRCh38
rs752703581 Chr.11: 3640077-3640077 on Build GRCh38
rs890328056 Chr.11: 3640099-3640099 on Build GRCh38
rs766428143 Chr.11: 3640008-3640008 on Build GRCh38
rs777713113 Chr.11: 3640108-3640108 on Build GRCh38
rs549358265 Chr.11: 3640123-3640123 on Build GRCh38
rs774801191 Chr.11: 3640051-3640051 on Build GRCh38
rs1019865067 Chr.11: 3639945-3639945 on Build GRCh38
rs780801593 Chr.11: 3640098-3640098 on Build GRCh38
rs142518955 Chr.11: 3639961-3639961 on Build GRCh38
rs763312413 Chr.11: 3639912-3639912 on Build GRCh38
rs186753401 Chr.11: 3639937-3639937 on Build GRCh38
rs764236648 Chr.11: 3639916-3639916 on Build GRCh38
rs377530388 Chr.11: 3639941-3639941 on Build GRCh38

View Full Product Details