Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38406871-38407126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98C
Entrez Gene ID
147965
Gene Name
family with sequence similarity 98 member C
Gene Aliases
-
UniGene
Hs.355162
Gene Chromosome Location
Chr.19: 38403124-38409088 on Build GRCh38
Transcripts
NM_174905.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7289361 c.862C>T p.L288F Chr.19: 38407021-38407021 on Build GRCh38
995967 c.786A>G p.P262P Chr.19: 38406945-38406945 on Build GRCh38
4855215 c.760G>A p.E254K Chr.19: 38406919-38406919 on Build GRCh38
1564448 c.860G>A p.C287Y Chr.19: 38407019-38407019 on Build GRCh38
3933022 c.866T>C p.V289A Chr.19: 38407025-38407025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200965223 Chr.19: 38407034-38407034 on Build GRCh38
rs768768718 Chr.19: 38407048-38407048 on Build GRCh38
rs751871392 Chr.19: 38406902-38406902 on Build GRCh38
rs372902479 Chr.19: 38406990-38406990 on Build GRCh38
rs780005583 Chr.19: 38406971-38406971 on Build GRCh38
rs564478351 Chr.19: 38406921-38406921 on Build GRCh38
rs769880116 Chr.19: 38406965-38406965 on Build GRCh38
rs765846466 Chr.19: 38407016-38407016 on Build GRCh38
rs759095515 Chr.19: 38407090-38407090 on Build GRCh38
rs371060977 Chr.19: 38407039-38407039 on Build GRCh38
rs186152772 Chr.19: 38407063-38407063 on Build GRCh38
rs1001446188 Chr.19: 38406934-38406934 on Build GRCh38
rs201689266 Chr.19: 38406924-38406924 on Build GRCh38
rs767148861 Chr.19: 38406893-38406893 on Build GRCh38
rs372453291 Chr.19: 38407023-38407023 on Build GRCh38
rs768570723 Chr.19: 38406976-38406976 on Build GRCh38
rs765741519 Chr.19: 38407013-38407013 on Build GRCh38
rs753300754 Chr.19: 38407017-38407017 on Build GRCh38
rs369314567 Chr.19: 38406956-38406956 on Build GRCh38
rs761561177 Chr.19: 38406991-38406991 on Build GRCh38
rs180979305 Chr.19: 38406949-38406949 on Build GRCh38
rs760274852 Chr.19: 38407009-38407009 on Build GRCh38
rs111816359 Chr.19: 38407024-38407024 on Build GRCh38
rs375869844 Chr.19: 38406993-38406993 on Build GRCh38
rs760315125 Chr.19: 38407064-38407064 on Build GRCh38
rs781443555 Chr.19: 38407028-38407028 on Build GRCh38
rs764863217 Chr.19: 38407099-38407099 on Build GRCh38
rs1014023200 Chr.19: 38406906-38406906 on Build GRCh38
rs752074044 Chr.19: 38407106-38407106 on Build GRCh38
rs750079593 Chr.19: 38406894-38406894 on Build GRCh38
rs949526293 Chr.19: 38407058-38407058 on Build GRCh38
rs758977176 Chr.19: 38406935-38406935 on Build GRCh38
rs760814820 Chr.19: 38406897-38406903 on Build GRCh38
rs757737681 Chr.19: 38406955-38406955 on Build GRCh38
rs781455000 Chr.19: 38406960-38406960 on Build GRCh38
rs777253562 Chr.19: 38406969-38406975 on Build GRCh38
rs879101940 Chr.19: 38407038-38407038 on Build GRCh38
rs770476417 Chr.19: 38407073-38407073 on Build GRCh38
rs749368929 Chr.19: 38407045-38407045 on Build GRCh38
rs563007712 Chr.19: 38407052-38407052 on Build GRCh38
rs753205605 Chr.19: 38406922-38406922 on Build GRCh38
rs201037487 Chr.19: 38407003-38407003 on Build GRCh38
rs905194379 Chr.19: 38407074-38407074 on Build GRCh38
rs757437214 Chr.19: 38407036-38407036 on Build GRCh38
rs552370895 Chr.19: 38407088-38407088 on Build GRCh38
rs748103028 Chr.19: 38407049-38407049 on Build GRCh38
rs201902099 Chr.19: 38407018-38407018 on Build GRCh38
rs746102252 Chr.19: 38406962-38406962 on Build GRCh38
rs1020470432 Chr.19: 38406977-38406977 on Build GRCh38
rs766738820 Chr.19: 38406947-38406947 on Build GRCh38
rs190729541 Chr.19: 38406896-38406896 on Build GRCh38
rs76769969 Chr.19: 38406908-38406908 on Build GRCh38
rs751959453 Chr.19: 38406950-38406950 on Build GRCh38
rs751237965 Chr.19: 38407021-38407031 on Build GRCh38
rs866869564 Chr.19: 38406943-38406943 on Build GRCh38
rs772869983 Chr.19: 38407007-38407007 on Build GRCh38
rs759978373 Chr.19: 38407004-38407004 on Build GRCh38
rs957207795 Chr.19: 38406953-38406953 on Build GRCh38
rs375336018 Chr.19: 38407044-38407044 on Build GRCh38
rs749194507 Chr.19: 38406972-38406972 on Build GRCh38
rs764693689 Chr.19: 38407021-38407021 on Build GRCh38

View Full Product Details