Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29400478-29400731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN2
Entrez Gene ID
1124
Gene Name
chimerin 2
Gene Aliases
ARHGAP3, BCH, CHN2-3, RHOGAP3
UniGene
Hs.654611
Gene Chromosome Location
Chr.7: 29146547-29514335 on Build GRCh38
Transcripts
NM_001293069.1 NM_001293070.1 NM_001293071.1 NM_001293072.1 NM_004067.3 XM_011515105.2 XM_011515106.2 XM_011515107.2 XM_017011721.1 XM_017011722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471852 c.560A>C p.K187T Chr.7: 29400587-29400587 on Build GRCh38
5720899 c.648C>T p.A216A Chr.7: 29400675-29400675 on Build GRCh38
3637802 c.659C>T p.S220L Chr.7: 29400686-29400686 on Build GRCh38
5953740 c.369C>T p.D123D Chr.7: 29400582-29400582 on Build GRCh38
4751858 c.446C>T p.T149I Chr.7: 29400698-29400698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024611827 Chr.7: 29400665-29400665 on Build GRCh38
rs758485827 Chr.7: 29400624-29400624 on Build GRCh38
rs200309206 Chr.7: 29400599-29400599 on Build GRCh38
rs1046010867 Chr.7: 29400538-29400538 on Build GRCh38
rs754195099 Chr.7: 29400603-29400603 on Build GRCh38
rs777886911 Chr.7: 29400634-29400634 on Build GRCh38
rs963532872 Chr.7: 29400682-29400682 on Build GRCh38
rs113586121 Chr.7: 29400617-29400617 on Build GRCh38
rs755348140 Chr.7: 29400604-29400604 on Build GRCh38
rs200681447 Chr.7: 29400661-29400661 on Build GRCh38
rs557065952 Chr.7: 29400510-29400510 on Build GRCh38
rs747086829 Chr.7: 29400635-29400635 on Build GRCh38
rs762930046 Chr.7: 29400677-29400677 on Build GRCh38
rs757031409 Chr.7: 29400586-29400586 on Build GRCh38
rs779311792 Chr.7: 29400616-29400616 on Build GRCh38
rs34045356 Chr.7: 29400579-29400579 on Build GRCh38
rs1016127535 Chr.7: 29400560-29400560 on Build GRCh38
rs759340406 Chr.7: 29400537-29400537 on Build GRCh38
rs765132257 Chr.7: 29400558-29400558 on Build GRCh38
rs544256613 Chr.7: 29400702-29400702 on Build GRCh38
rs139046995 Chr.7: 29400582-29400582 on Build GRCh38
rs149886903 Chr.7: 29400648-29400648 on Build GRCh38
rs577212527 Chr.7: 29400675-29400675 on Build GRCh38
rs751272925 Chr.7: 29400583-29400583 on Build GRCh38
rs140592115 Chr.7: 29400536-29400536 on Build GRCh38
rs772092920 Chr.7: 29400511-29400511 on Build GRCh38
rs186911567 Chr.7: 29400618-29400618 on Build GRCh38
rs762580332 Chr.7: 29400580-29400580 on Build GRCh38
rs532994878 Chr.7: 29400514-29400514 on Build GRCh38
rs190512543 Chr.7: 29400515-29400515 on Build GRCh38
rs771128115 Chr.7: 29400534-29400534 on Build GRCh38
rs747174668 Chr.7: 29400522-29400522 on Build GRCh38
rs775296935 Chr.7: 29400676-29400676 on Build GRCh38
rs768705079 Chr.7: 29400697-29400697 on Build GRCh38
rs990792626 Chr.7: 29400643-29400643 on Build GRCh38
rs774076136 Chr.7: 29400700-29400700 on Build GRCh38

View Full Product Details