Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202377304-202377565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR2
Entrez Gene ID
659
Gene Name
bone morphogenetic protein receptor type 2
Gene Aliases
BMPR-II, BMPR3, BMR2, BRK-3, POVD1, PPH1, T-ALK
UniGene
Hs.471119
Gene Chromosome Location
Chr.2: 202376310-202567751 on Build GRCh38
Transcripts
NM_001204.6 XM_011511687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3576403 c.23C>T p.P8L Chr.2: 202377497-202377497 on Build GRCh38
4090291 c.46T>C p.W16R Chr.2: 202377520-202377520 on Build GRCh38
4854503 c.71C>A p.A24E Chr.2: 202377545-202377545 on Build GRCh38
1404571 c.27G>A p.W9* Chr.2: 202377501-202377501 on Build GRCh38
2907987 c.48G>A p.W16* Chr.2: 202377522-202377522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377020025 Chr.2: 202377544-202377544 on Build GRCh38
rs567729546 Chr.2: 202377435-202377435 on Build GRCh38
rs140659948 Chr.2: 202377542-202377542 on Build GRCh38
rs755516081 Chr.2: 202377448-202377448 on Build GRCh38
rs758635863 Chr.2: 202377457-202377457 on Build GRCh38
rs779617809 Chr.2: 202377449-202377449 on Build GRCh38
rs748855275 Chr.2: 202377481-202377481 on Build GRCh38
rs759723251 Chr.2: 202377531-202377531 on Build GRCh38
rs370382371 Chr.2: 202377467-202377467 on Build GRCh38
rs376097435 Chr.2: 202377463-202377463 on Build GRCh38
rs201940669 Chr.2: 202377455-202377455 on Build GRCh38
rs965825599 Chr.2: 202377380-202377380 on Build GRCh38
rs766753997 Chr.2: 202377523-202377523 on Build GRCh38
rs886039219 Chr.2: 202377490-202377490 on Build GRCh38
rs774089439 Chr.2: 202377493-202377493 on Build GRCh38
rs981756684 Chr.2: 202377367-202377367 on Build GRCh38
rs766524931 Chr.2: 202377430-202377430 on Build GRCh38
rs141716313 Chr.2: 202377541-202377541 on Build GRCh38
rs745474189 Chr.2: 202377464-202377464 on Build GRCh38
rs911336473 Chr.2: 202377337-202377337 on Build GRCh38
rs549281044 Chr.2: 202377483-202377483 on Build GRCh38
rs765376013 Chr.2: 202377534-202377534 on Build GRCh38
rs906306607 Chr.2: 202377507-202377507 on Build GRCh38
rs767166903 Chr.2: 202377505-202377505 on Build GRCh38
rs1048629816 Chr.2: 202377429-202377429 on Build GRCh38
rs765364075 Chr.2: 202377405-202377405 on Build GRCh38
rs933516159 Chr.2: 202377495-202377495 on Build GRCh38
rs918573678 Chr.2: 202377400-202377400 on Build GRCh38
rs760946823 Chr.2: 202377514-202377514 on Build GRCh38
rs483352902 Chr.2: 202377518-202377518 on Build GRCh38
rs527863342 Chr.2: 202377329-202377329 on Build GRCh38
rs373783006 Chr.2: 202377509-202377509 on Build GRCh38
rs969913869 Chr.2: 202377354-202377354 on Build GRCh38
rs988842901 Chr.2: 202377427-202377427 on Build GRCh38
rs1022755331 Chr.2: 202377336-202377336 on Build GRCh38
rs761823322 Chr.2: 202377503-202377503 on Build GRCh38
rs922087863 Chr.2: 202377485-202377485 on Build GRCh38
rs760123053 Chr.2: 202377439-202377439 on Build GRCh38
rs147936167 Chr.2: 202377382-202377382 on Build GRCh38
rs149973695 Chr.2: 202377537-202377537 on Build GRCh38
rs1004254626 Chr.2: 202377540-202377540 on Build GRCh38
rs370120266 Chr.2: 202377545-202377545 on Build GRCh38
rs778496865 Chr.2: 202377443-202377443 on Build GRCh38
rs947457883 Chr.2: 202377441-202377441 on Build GRCh38
rs774960253 Chr.2: 202377471-202377471 on Build GRCh38

View Full Product Details