Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 30710120-30710361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPRIN2
Entrez Gene ID
65981
Gene Name
caprin family member 2
Gene Aliases
C1QDC1, EEG-1, EEG1, RNG140
UniGene
Hs.234355
Gene Chromosome Location
Chr.12: 30709552-30754994 on Build GRCh38
Transcripts
NM_001002259.2 NM_001206856.2 NM_001319842.1 NM_001319843.1 NM_001319844.1 NM_001319845.1 NM_001319846.1 NM_023925.4 NM_032156.4 XM_006719138.3 XM_006719139.3 XM_006719140.3 XM_006719142.3 XM_006719144.3 XM_006719145.3 XM_006719146.3 XM_006719147.3 XM_017019851.1 XM_017019852.1 XM_017019853.1 XM_017019854.1 XM_017019855.1 XM_017019856.1 XM_017019857.1 XM_017019858.1 XM_017019859.1 XM_017019860.1 XM_017019861.1 XM_017019862.1 XM_017019863.1 XM_017019865.1 XM_017019866.1 XM_017019867.1 XM_017019868.1 XM_017019869.1 XM_017019870.1 XM_017019871.1 XM_017019872.1 XM_017019873.1 XM_017019874.1 XM_017019875.1 XM_017019876.1 XM_017019877.1 XM_017019878.1 XM_017019879.1 XM_017019880.1 XM_017019881.1 XM_017019882.1 XM_017019883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2205127 c.3079G>A p.G1027R Chr.12: 30710207-30710207 on Build GRCh38
4669197 c.2944G>A p.A982T Chr.12: 30710342-30710342 on Build GRCh38
54384 c.3159G>A p.K1053K Chr.12: 30710127-30710127 on Build GRCh38
4669196 c.3110T>G p.F1037C Chr.12: 30710176-30710176 on Build GRCh38
2205126 c.3080G>A p.G1027E Chr.12: 30710206-30710206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776471356 Chr.12: 30710273-30710273 on Build GRCh38
rs776145639 Chr.12: 30710232-30710232 on Build GRCh38
rs756917174 Chr.12: 30710325-30710325 on Build GRCh38
rs765096929 Chr.12: 30710307-30710307 on Build GRCh38
rs372208056 Chr.12: 30710337-30710337 on Build GRCh38
rs951655638 Chr.12: 30710258-30710258 on Build GRCh38
rs965803693 Chr.12: 30710331-30710331 on Build GRCh38
rs35671069 Chr.12: 30710273-30710274 on Build GRCh38
rs150951809 Chr.12: 30710321-30710321 on Build GRCh38
rs202247225 Chr.12: 30710328-30710328 on Build GRCh38
rs7137533 Chr.12: 30710151-30710151 on Build GRCh38
rs777431817 Chr.12: 30710148-30710148 on Build GRCh38
rs768642984 Chr.12: 30710246-30710246 on Build GRCh38
rs140741615 Chr.12: 30710336-30710336 on Build GRCh38
rs200055967 Chr.12: 30710317-30710317 on Build GRCh38
rs759470076 Chr.12: 30710304-30710304 on Build GRCh38
rs770489821 Chr.12: 30710211-30710211 on Build GRCh38
rs111761506 Chr.12: 30710322-30710322 on Build GRCh38
rs746362702 Chr.12: 30710201-30710201 on Build GRCh38
rs780659467 Chr.12: 30710340-30710340 on Build GRCh38
rs957278438 Chr.12: 30710297-30710297 on Build GRCh38
rs900613675 Chr.12: 30710174-30710174 on Build GRCh38
rs368158820 Chr.12: 30710254-30710254 on Build GRCh38
rs570016104 Chr.12: 30710308-30710308 on Build GRCh38
rs766177185 Chr.12: 30710262-30710262 on Build GRCh38
rs752649925 Chr.12: 30710311-30710311 on Build GRCh38
rs781457861 Chr.12: 30710196-30710196 on Build GRCh38
rs774594835 Chr.12: 30710252-30710252 on Build GRCh38
rs774768463 Chr.12: 30710192-30710192 on Build GRCh38
rs746668837 Chr.12: 30710150-30710150 on Build GRCh38
rs142547870 Chr.12: 30710315-30710315 on Build GRCh38
rs375715809 Chr.12: 30710242-30710242 on Build GRCh38

View Full Product Details