Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4901043-4901316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf107
Entrez Gene ID
100130311
Gene Name
chromosome 17 open reading frame 107
Gene Aliases
-
UniGene
Hs.579243
Gene Chromosome Location
Chr.17: 4899653-4902932 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6081535 c.688G>T p.V230F Chr.17: 4901104-4901104 on Build GRCh38
5055573 c.687C>T p.D229D Chr.17: 4901105-4901105 on Build GRCh38
5936282 c.713G>A p.R238Q Chr.17: 4901079-4901079 on Build GRCh38
6628801 c.622G>A p.D208N Chr.17: 4901170-4901170 on Build GRCh38
1384468 c.699G>A p.S233S Chr.17: 4901093-4901093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949381816 Chr.17: 4901245-4901245 on Build GRCh38
rs781166271 Chr.17: 4901140-4901140 on Build GRCh38
rs769424853 Chr.17: 4901083-4901083 on Build GRCh38
rs762103017 Chr.17: 4901196-4901196 on Build GRCh38
rs201434993 Chr.17: 4901082-4901082 on Build GRCh38
rs761086547 Chr.17: 4901194-4901194 on Build GRCh38
rs931818729 Chr.17: 4901201-4901201 on Build GRCh38
rs776203354 Chr.17: 4901193-4901193 on Build GRCh38
rs1045728830 Chr.17: 4901251-4901251 on Build GRCh38
rs748765369 Chr.17: 4901087-4901087 on Build GRCh38
rs770306808 Chr.17: 4901159-4901159 on Build GRCh38
rs778485449 Chr.17: 4901214-4901214 on Build GRCh38
rs754148818 Chr.17: 4901127-4901127 on Build GRCh38
rs764599384 Chr.17: 4901195-4901195 on Build GRCh38
rs773916039 Chr.17: 4901160-4901160 on Build GRCh38
rs760118625 Chr.17: 4901097-4901097 on Build GRCh38
rs760565441 Chr.17: 4901188-4901188 on Build GRCh38
rs767704727 Chr.17: 4901112-4901112 on Build GRCh38
rs757240025 Chr.17: 4901131-4901131 on Build GRCh38
rs764709459 Chr.17: 4901142-4901143 on Build GRCh38
rs765175027 Chr.17: 4901200-4901200 on Build GRCh38
rs1023841193 Chr.17: 4901119-4901119 on Build GRCh38
rs777150987 Chr.17: 4901228-4901228 on Build GRCh38
rs374236994 Chr.17: 4901242-4901242 on Build GRCh38
rs957078131 Chr.17: 4901161-4901161 on Build GRCh38
rs746300839 Chr.17: 4901163-4901163 on Build GRCh38
rs940761317 Chr.17: 4901221-4901221 on Build GRCh38
rs748961544 Chr.17: 4901149-4901149 on Build GRCh38
rs994082504 Chr.17: 4901075-4901076 on Build GRCh38
rs950829559 Chr.17: 4901225-4901225 on Build GRCh38
rs1014252253 Chr.17: 4901099-4901099 on Build GRCh38
rs757313173 Chr.17: 4901204-4901205 on Build GRCh38
rs769079236 Chr.17: 4901223-4901223 on Build GRCh38
rs763066225 Chr.17: 4901236-4901236 on Build GRCh38
rs141183965 Chr.17: 4901121-4901121 on Build GRCh38
rs564044217 Chr.17: 4901141-4901141 on Build GRCh38
rs746035699 Chr.17: 4901236-4901237 on Build GRCh38
rs753828284 Chr.17: 4901172-4901178 on Build GRCh38
rs776008550 Chr.17: 4901118-4901118 on Build GRCh38
rs770523947 Chr.17: 4901088-4901088 on Build GRCh38
rs28999110 Chr.17: 4901071-4901071 on Build GRCh38
rs886053126 Chr.17: 4901069-4901069 on Build GRCh38
rs145522662 Chr.17: 4901086-4901086 on Build GRCh38
rs1036405246 Chr.17: 4901237-4901237 on Build GRCh38
rs765394012 Chr.17: 4901137-4901137 on Build GRCh38
rs202127846 Chr.17: 4901204-4901204 on Build GRCh38
rs531123327 Chr.17: 4901122-4901122 on Build GRCh38
rs745376445 Chr.17: 4901216-4901216 on Build GRCh38
rs375146171 Chr.17: 4901093-4901093 on Build GRCh38
rs376001778 Chr.17: 4901139-4901139 on Build GRCh38
rs746993242 Chr.17: 4901077-4901077 on Build GRCh38
rs778873861 Chr.17: 4901232-4901235 on Build GRCh38
rs1032496823 Chr.17: 4901282-4901282 on Build GRCh38
rs771744770 Chr.17: 4901218-4901218 on Build GRCh38
rs780681661 Chr.17: 4901220-4901220 on Build GRCh38
rs1049141147 Chr.17: 4901072-4901072 on Build GRCh38
rs762144942 Chr.17: 4901232-4901232 on Build GRCh38
rs748823182 Chr.17: 4901208-4901208 on Build GRCh38
rs912870458 Chr.17: 4901183-4901183 on Build GRCh38
rs751388919 Chr.17: 4901138-4901138 on Build GRCh38
rs563436065 Chr.17: 4901295-4901295 on Build GRCh38
rs577641512 Chr.17: 4901146-4901146 on Build GRCh38
rs767412498 Chr.17: 4901202-4901202 on Build GRCh38
rs756946862 Chr.17: 4901209-4901209 on Build GRCh38
rs755943507 Chr.17: 4901142-4901142 on Build GRCh38
rs1001048941 Chr.17: 4901270-4901270 on Build GRCh38
rs768299579 Chr.17: 4901078-4901078 on Build GRCh38
rs772616743 Chr.17: 4901170-4901170 on Build GRCh38
rs905423038 Chr.17: 4901261-4901261 on Build GRCh38

View Full Product Details