Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 95875881-95876152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC6L2
Entrez Gene ID
375748
Gene Name
ERCC excision repair 6 like 2
Gene Aliases
BMFS2, C9orf102, RAD26L, SR278
UniGene
Hs.432364
Gene Chromosome Location
Chr.9: 95875618-96041092 on Build GRCh38
Transcripts
NM_001010895.2 NM_020207.4 XM_011518641.2 XM_011518644.2 XM_011518645.2 XM_011518646.2 XM_011518647.1 XM_017014707.1 NR_023389.1 NR_023390.1

Gene Symbol
LINC00476
Entrez Gene ID
100128782
Gene Name
long intergenic non-protein coding RNA 476
Gene Aliases
C9orf130, NAG11, NAG12
UniGene
Hs.494549
Gene Chromosome Location
Chr.9: 95806088-95875977 on Build GRCh38
Transcripts
NM_001010895.2 NM_020207.4 XM_011518641.2 XM_011518644.2 XM_011518645.2 XM_011518646.2 XM_011518647.1 XM_017014707.1 NR_023389.1 NR_023390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278622 c.62C>G p.A21G Chr.9: 95876067-95876067 on Build GRCh38
5422538 c.79G>T p.D27Y Chr.9: 95876084-95876084 on Build GRCh38
5638324 c.41C>G p.P14R Chr.9: 95876046-95876046 on Build GRCh38
456305 c.1A>G p.M1V Chr.9: 95876006-95876006 on Build GRCh38
4501503 c.58C>T p.R20C Chr.9: 95876063-95876063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746362971 Chr.9: 95876036-95876036 on Build GRCh38
rs951631275 Chr.9: 95875961-95875961 on Build GRCh38
rs758868728 Chr.9: 95876038-95876038 on Build GRCh38
rs752897090 Chr.9: 95875971-95875971 on Build GRCh38
rs758889766 Chr.9: 95876067-95876067 on Build GRCh38
rs78293324 Chr.9: 95876031-95876031 on Build GRCh38
rs1019676408 Chr.9: 95876068-95876068 on Build GRCh38
rs759779322 Chr.9: 95876108-95876109 on Build GRCh38
rs1041556138 Chr.9: 95876009-95876009 on Build GRCh38
rs202222262 Chr.9: 95876026-95876026 on Build GRCh38
rs899953370 Chr.9: 95876056-95876056 on Build GRCh38
rs201393222 Chr.9: 95876027-95876027 on Build GRCh38
rs750367352 Chr.9: 95876017-95876017 on Build GRCh38
rs776425022 Chr.9: 95876045-95876045 on Build GRCh38
rs761678165 Chr.9: 95876014-95876014 on Build GRCh38
rs760749677 Chr.9: 95876055-95876055 on Build GRCh38
rs749050155 Chr.9: 95876094-95876094 on Build GRCh38
rs543576960 Chr.9: 95876082-95876082 on Build GRCh38
rs768453979 Chr.9: 95876004-95876004 on Build GRCh38
rs999464531 Chr.9: 95875903-95875903 on Build GRCh38
rs690528 Chr.9: 95876006-95876006 on Build GRCh38
rs75475951 Chr.9: 95875974-95875974 on Build GRCh38
rs370033874 Chr.9: 95876104-95876104 on Build GRCh38
rs917768404 Chr.9: 95876000-95876000 on Build GRCh38
rs912240845 Chr.9: 95875972-95875972 on Build GRCh38
rs780716399 Chr.9: 95876073-95876073 on Build GRCh38
rs35504182 Chr.9: 95876059-95876060 on Build GRCh38
rs756372606 Chr.9: 95875973-95875973 on Build GRCh38
rs1008286958 Chr.9: 95875912-95875912 on Build GRCh38
rs982713997 Chr.9: 95876072-95876072 on Build GRCh38
rs748315283 Chr.9: 95876035-95876035 on Build GRCh38
rs746429736 Chr.9: 95875987-95875987 on Build GRCh38
rs759529546 Chr.9: 95876105-95876105 on Build GRCh38
rs750783247 Chr.9: 95876066-95876066 on Build GRCh38
rs558663065 Chr.9: 95876133-95876133 on Build GRCh38
rs779481462 Chr.9: 95875982-95875982 on Build GRCh38
rs764135092 Chr.9: 95876106-95876106 on Build GRCh38
rs1012339660 Chr.9: 95875939-95875939 on Build GRCh38
rs772020691 Chr.9: 95876003-95876004 on Build GRCh38
rs780904686 Chr.9: 95876081-95876081 on Build GRCh38
rs746450201 Chr.9: 95876076-95876076 on Build GRCh38
rs576071831 Chr.9: 95876077-95876077 on Build GRCh38
rs371970239 Chr.9: 95876030-95876030 on Build GRCh38
rs141071823 Chr.9: 95875960-95875960 on Build GRCh38
rs56108623 Chr.9: 95876024-95876024 on Build GRCh38
rs781189850 Chr.9: 95875963-95875963 on Build GRCh38
rs765604584 Chr.9: 95876062-95876062 on Build GRCh38
rs986866020 Chr.9: 95875970-95875970 on Build GRCh38
rs766330761 Chr.9: 95876053-95876053 on Build GRCh38
rs760372624 Chr.9: 95875997-95875997 on Build GRCh38
rs761797983 Chr.9: 95875958-95875958 on Build GRCh38
rs139286981 Chr.9: 95876059-95876059 on Build GRCh38
rs1056506872 Chr.9: 95875908-95875908 on Build GRCh38
rs771284265 Chr.9: 95875980-95875980 on Build GRCh38
rs371959202 Chr.9: 95876113-95876113 on Build GRCh38
rs768362145 Chr.9: 95876042-95876042 on Build GRCh38
rs1024183803 Chr.9: 95875940-95875940 on Build GRCh38
rs865819226 Chr.9: 95876060-95876060 on Build GRCh38
rs769611647 Chr.9: 95876049-95876049 on Build GRCh38
rs773014434 Chr.9: 95876091-95876091 on Build GRCh38
rs767605945 Chr.9: 95876025-95876025 on Build GRCh38
rs548324740 Chr.9: 95875992-95875992 on Build GRCh38
rs112412135 Chr.9: 95875922-95875922 on Build GRCh38
rs532130675 Chr.9: 95875956-95875956 on Build GRCh38
rs10986795 Chr.9: 95876007-95876007 on Build GRCh38
rs962888238 Chr.9: 95875953-95875953 on Build GRCh38
rs778926161 Chr.9: 95876057-95876057 on Build GRCh38
rs151308487 Chr.9: 95876046-95876046 on Build GRCh38
rs866086136 Chr.9: 95876019-95876019 on Build GRCh38
rs371776172 Chr.9: 95876050-95876050 on Build GRCh38
rs775186765 Chr.9: 95875996-95875996 on Build GRCh38
rs35881432 Chr.9: 95876114-95876115 on Build GRCh38
rs762054429 Chr.9: 95876125-95876125 on Build GRCh38
rs747907138 Chr.9: 95875985-95875987 on Build GRCh38
rs769366535 Chr.9: 95876084-95876084 on Build GRCh38
rs926399972 Chr.9: 95876023-95876023 on Build GRCh38
rs1021312142 Chr.9: 95876127-95876127 on Build GRCh38
rs995728267 Chr.9: 95875955-95875955 on Build GRCh38
rs951299785 Chr.9: 95876063-95876063 on Build GRCh38
rs566324412 Chr.9: 95876040-95876040 on Build GRCh38

View Full Product Details