Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175181228-175181501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5G3
Entrez Gene ID
518
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Gene Aliases
P3
UniGene
Hs.429
Gene Chromosome Location
Chr.2: 175176258-175181762 on Build GRCh38
Transcripts
NM_001002258.4 NM_001190329.1 NM_001689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3720920 c.14C>T p.A5V Chr.2: 175181380-175181380 on Build GRCh38
4979482 c.33C>T p.P11P Chr.2: 175181361-175181361 on Build GRCh38
5626778 c.6C>T p.F2F Chr.2: 175181388-175181388 on Build GRCh38
4086890 c.31C>G p.P11A Chr.2: 175181363-175181363 on Build GRCh38
4086889 c.36T>A p.S12S Chr.2: 175181358-175181358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111790501 Chr.2: 175181424-175181424 on Build GRCh38
rs760618392 Chr.2: 175181375-175181375 on Build GRCh38
rs368052450 Chr.2: 175181305-175181305 on Build GRCh38
rs200580954 Chr.2: 175181364-175181364 on Build GRCh38
rs754405934 Chr.2: 175181408-175181408 on Build GRCh38
rs750114777 Chr.2: 175181399-175181399 on Build GRCh38
rs777174231 Chr.2: 175181440-175181440 on Build GRCh38
rs758040136 Chr.2: 175181401-175181401 on Build GRCh38
rs757950195 Chr.2: 175181332-175181332 on Build GRCh38
rs41270205 Chr.2: 175181439-175181439 on Build GRCh38
rs940882579 Chr.2: 175181263-175181263 on Build GRCh38
rs191911488 Chr.2: 175181457-175181457 on Build GRCh38
rs746985380 Chr.2: 175181339-175181339 on Build GRCh38
rs762279119 Chr.2: 175181443-175181443 on Build GRCh38
rs772347621 Chr.2: 175181428-175181428 on Build GRCh38
rs202118535 Chr.2: 175181376-175181376 on Build GRCh38
rs939031501 Chr.2: 175181475-175181475 on Build GRCh38
rs933328782 Chr.2: 175181250-175181250 on Build GRCh38
rs906765319 Chr.2: 175181472-175181472 on Build GRCh38
rs569629231 Chr.2: 175181427-175181427 on Build GRCh38
rs760570453 Chr.2: 175181344-175181344 on Build GRCh38
rs774429139 Chr.2: 175181257-175181257 on Build GRCh38
rs527399417 Chr.2: 175181249-175181249 on Build GRCh38
rs940746869 Chr.2: 175181454-175181454 on Build GRCh38
rs368357116 Chr.2: 175181410-175181410 on Build GRCh38
rs770212339 Chr.2: 175181444-175181444 on Build GRCh38
rs975770297 Chr.2: 175181361-175181361 on Build GRCh38
rs767762226 Chr.2: 175181291-175181291 on Build GRCh38
rs1005376467 Chr.2: 175181482-175181482 on Build GRCh38
rs1052071226 Chr.2: 175181480-175181480 on Build GRCh38
rs200125762 Chr.2: 175181411-175181411 on Build GRCh38
rs753175334 Chr.2: 175181460-175181460 on Build GRCh38
rs140931538 Chr.2: 175181363-175181363 on Build GRCh38
rs551029493 Chr.2: 175181403-175181403 on Build GRCh38
rs142182201 Chr.2: 175181362-175181362 on Build GRCh38
rs906223502 Chr.2: 175181301-175181301 on Build GRCh38
rs777080129 Chr.2: 175181381-175181381 on Build GRCh38
rs116934836 Chr.2: 175181275-175181275 on Build GRCh38
rs779012670 Chr.2: 175181468-175181468 on Build GRCh38
rs1045316363 Chr.2: 175181294-175181294 on Build GRCh38
rs563242795 Chr.2: 175181438-175181438 on Build GRCh38
rs73039765 Chr.2: 175181279-175181279 on Build GRCh38
rs747989377 Chr.2: 175181355-175181355 on Build GRCh38
rs551727050 Chr.2: 175181258-175181258 on Build GRCh38
rs757547399 Chr.2: 175181328-175181328 on Build GRCh38
rs761243040 Chr.2: 175181307-175181307 on Build GRCh38
rs375608255 Chr.2: 175181402-175181402 on Build GRCh38
rs775629707 Chr.2: 175181373-175181373 on Build GRCh38
rs764428783 Chr.2: 175181313-175181313 on Build GRCh38
rs146495548 Chr.2: 175181465-175181465 on Build GRCh38
rs192662247 Chr.2: 175181292-175181292 on Build GRCh38
rs750026487 Chr.2: 175181330-175181330 on Build GRCh38
rs1040747511 Chr.2: 175181463-175181463 on Build GRCh38
rs777708939 Chr.2: 175181413-175181413 on Build GRCh38
rs185401588 Chr.2: 175181306-175181306 on Build GRCh38
rs370456305 Chr.2: 175181359-175181359 on Build GRCh38
rs375263899 Chr.2: 175181317-175181317 on Build GRCh38
rs372802971 Chr.2: 175181352-175181352 on Build GRCh38
rs751085612 Chr.2: 175181405-175181405 on Build GRCh38
rs1052311549 Chr.2: 175181251-175181251 on Build GRCh38
rs762185123 Chr.2: 175181388-175181388 on Build GRCh38
rs1042985 Chr.2: 175181400-175181400 on Build GRCh38
rs764897675 Chr.2: 175181329-175181329 on Build GRCh38
rs994034815 Chr.2: 175181296-175181296 on Build GRCh38
rs779583781 Chr.2: 175181334-175181334 on Build GRCh38
rs1010757349 Chr.2: 175181343-175181343 on Build GRCh38

View Full Product Details