Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100081476-100081705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPN1
Entrez Gene ID
1369
Gene Name
carboxypeptidase N subunit 1
Gene Aliases
CPN, SCPN
UniGene
Hs.2246
Gene Chromosome Location
Chr.10: 100042308-100081885 on Build GRCh38
Transcripts
NM_001308.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3433511 c.98G>A p.R33Q Chr.10: 100081528-100081528 on Build GRCh38
23428 c.141G>A p.R47R Chr.10: 100081485-100081485 on Build GRCh38
5721571 c.42C>T p.L14L Chr.10: 100081584-100081584 on Build GRCh38
220234 c.139C>T p.R47W Chr.10: 100081487-100081487 on Build GRCh38
1702899 c.121G>A p.E41K Chr.10: 100081505-100081505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762992078 Chr.10: 100081658-100081658 on Build GRCh38
rs955117066 Chr.10: 100081632-100081632 on Build GRCh38
rs893376063 Chr.10: 100081680-100081680 on Build GRCh38
rs111400589 Chr.10: 100081612-100081612 on Build GRCh38
rs746251575 Chr.10: 100081571-100081571 on Build GRCh38
rs770270851 Chr.10: 100081572-100081572 on Build GRCh38
rs773064523 Chr.10: 100081536-100081536 on Build GRCh38
rs752640687 Chr.10: 100081652-100081652 on Build GRCh38
rs568940128 Chr.10: 100081671-100081671 on Build GRCh38
rs758527116 Chr.10: 100081555-100081555 on Build GRCh38
rs781573757 Chr.10: 100081563-100081563 on Build GRCh38
rs922192921 Chr.10: 100081616-100081616 on Build GRCh38
rs899596582 Chr.10: 100081633-100081633 on Build GRCh38
rs751664367 Chr.10: 100081667-100081667 on Build GRCh38
rs549281938 Chr.10: 100081512-100081512 on Build GRCh38
rs757536670 Chr.10: 100081592-100081592 on Build GRCh38
rs766323805 Chr.10: 100081541-100081541 on Build GRCh38
rs370527794 Chr.10: 100081567-100081567 on Build GRCh38
rs553176799 Chr.10: 100081580-100081580 on Build GRCh38
rs781275676 Chr.10: 100081675-100081675 on Build GRCh38
rs765273218 Chr.10: 100081567-100081567 on Build GRCh38
rs751706282 Chr.10: 100081525-100081525 on Build GRCh38
rs771747138 Chr.10: 100081584-100081584 on Build GRCh38
rs748205584 Chr.10: 100081499-100081499 on Build GRCh38
rs771355307 Chr.10: 100081515-100081515 on Build GRCh38
rs747648440 Chr.10: 100081581-100081581 on Build GRCh38
rs776976696 Chr.10: 100081516-100081516 on Build GRCh38
rs757598430 Chr.10: 100081560-100081560 on Build GRCh38
rs776417344 Chr.10: 100081608-100081608 on Build GRCh38
rs200920833 Chr.10: 100081640-100081640 on Build GRCh38
rs752752245 Chr.10: 100081675-100081678 on Build GRCh38
rs538591971 Chr.10: 100081619-100081619 on Build GRCh38
rs562025469 Chr.10: 100081578-100081578 on Build GRCh38
rs760354309 Chr.10: 100081602-100081602 on Build GRCh38
rs567825978 Chr.10: 100081529-100081529 on Build GRCh38
rs754901371 Chr.10: 100081549-100081549 on Build GRCh38
rs765087377 Chr.10: 100081551-100081551 on Build GRCh38
rs758547188 Chr.10: 100081500-100081500 on Build GRCh38
rs770737401 Chr.10: 100081605-100081605 on Build GRCh38
rs760515524 Chr.10: 100081538-100081538 on Build GRCh38
rs866703720 Chr.10: 100081495-100081495 on Build GRCh38
rs751823708 Chr.10: 100081556-100081556 on Build GRCh38
rs777979903 Chr.10: 100081511-100081511 on Build GRCh38
rs779001062 Chr.10: 100081497-100081497 on Build GRCh38
rs764210605 Chr.10: 100081659-100081659 on Build GRCh38
rs200035303 Chr.10: 100081533-100081533 on Build GRCh38
rs201817147 Chr.10: 100081546-100081546 on Build GRCh38
rs1039448436 Chr.10: 100081537-100081537 on Build GRCh38
rs754989179 Chr.10: 100081496-100081496 on Build GRCh38
rs373785594 Chr.10: 100081553-100081553 on Build GRCh38
rs746438789 Chr.10: 100081523-100081523 on Build GRCh38

View Full Product Details