Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10948511-10948766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf127
Entrez Gene ID
148345
Gene Name
chromosome 1 open reading frame 127
Gene Aliases
-
UniGene
Hs.127026
Gene Chromosome Location
Chr.1: 10946473-10982037 on Build GRCh38
Transcripts
NM_001170754.1 XM_017000373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5843000 c.1579C>T p.R527* Chr.1: 10948556-10948556 on Build GRCh38
3470915 c.1123G>A p.E375K Chr.1: 10948511-10948511 on Build GRCh38
5843001 c.1078C>T p.R360* Chr.1: 10948556-10948556 on Build GRCh38
3470928 c.1405G>A p.E469K Chr.1: 10948730-10948730 on Build GRCh38
3470924 c.1423C>T p.H475Y Chr.1: 10948712-10948712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755373283 Chr.1: 10948567-10948567 on Build GRCh38
rs201751216 Chr.1: 10948570-10948570 on Build GRCh38
rs777772710 Chr.1: 10948642-10948642 on Build GRCh38
rs775550746 Chr.1: 10948605-10948605 on Build GRCh38
rs74772255 Chr.1: 10948742-10948742 on Build GRCh38
rs761380061 Chr.1: 10948661-10948661 on Build GRCh38
rs746066280 Chr.1: 10948702-10948702 on Build GRCh38
rs1281016 Chr.1: 10948537-10948537 on Build GRCh38
rs776223426 Chr.1: 10948592-10948592 on Build GRCh38
rs1017485318 Chr.1: 10948623-10948623 on Build GRCh38
rs749096620 Chr.1: 10948706-10948706 on Build GRCh38
rs1002695540 Chr.1: 10948697-10948697 on Build GRCh38
rs17036032 Chr.1: 10948638-10948638 on Build GRCh38
rs371473808 Chr.1: 10948684-10948684 on Build GRCh38
rs768635702 Chr.1: 10948713-10948713 on Build GRCh38
rs757970948 Chr.1: 10948550-10948550 on Build GRCh38
rs753556392 Chr.1: 10948738-10948738 on Build GRCh38
rs779386863 Chr.1: 10948569-10948569 on Build GRCh38
rs552406765 Chr.1: 10948649-10948649 on Build GRCh38
rs200187203 Chr.1: 10948590-10948590 on Build GRCh38
rs773929314 Chr.1: 10948660-10948660 on Build GRCh38
rs149489353 Chr.1: 10948643-10948643 on Build GRCh38
rs772115676 Chr.1: 10948575-10948575 on Build GRCh38
rs139797750 Chr.1: 10948549-10948549 on Build GRCh38
rs763654600 Chr.1: 10948607-10948607 on Build GRCh38
rs112710664 Chr.1: 10948665-10948665 on Build GRCh38
rs145775173 Chr.1: 10948576-10948576 on Build GRCh38
rs777328651 Chr.1: 10948693-10948693 on Build GRCh38
rs569028775 Chr.1: 10948715-10948715 on Build GRCh38
rs753074905 Chr.1: 10948683-10948683 on Build GRCh38
rs373780786 Chr.1: 10948577-10948577 on Build GRCh38
rs564155660 Chr.1: 10948656-10948656 on Build GRCh38
rs758927695 Chr.1: 10948639-10948639 on Build GRCh38
rs148950438 Chr.1: 10948600-10948600 on Build GRCh38
rs764494881 Chr.1: 10948744-10948744 on Build GRCh38
rs747186057 Chr.1: 10948588-10948588 on Build GRCh38
rs200134961 Chr.1: 10948621-10948621 on Build GRCh38
rs780231005 Chr.1: 10948557-10948557 on Build GRCh38
rs772627997 Chr.1: 10948729-10948729 on Build GRCh38
rs139817492 Chr.1: 10948651-10948651 on Build GRCh38
rs774331508 Chr.1: 10948717-10948717 on Build GRCh38
rs763748868 Chr.1: 10948552-10948552 on Build GRCh38
rs115282418 Chr.1: 10948721-10948721 on Build GRCh38
rs568588607 Chr.1: 10948692-10948692 on Build GRCh38
rs766025626 Chr.1: 10948735-10948735 on Build GRCh38
rs774769731 Chr.1: 10948555-10948556 on Build GRCh38
rs558047600 Chr.1: 10948731-10948731 on Build GRCh38
rs75518308 Chr.1: 10948628-10948628 on Build GRCh38
rs766992937 Chr.1: 10948663-10948663 on Build GRCh38
rs768725521 Chr.1: 10948658-10948658 on Build GRCh38
rs770985364 Chr.1: 10948648-10948648 on Build GRCh38
rs759373363 Chr.1: 10948601-10948601 on Build GRCh38
rs765538661 Chr.1: 10948680-10948680 on Build GRCh38
rs759334727 Chr.1: 10948536-10948536 on Build GRCh38
rs146743970 Chr.1: 10948556-10948556 on Build GRCh38
rs760285108 Chr.1: 10948668-10948668 on Build GRCh38
rs775452968 Chr.1: 10948654-10948654 on Build GRCh38
rs368505079 Chr.1: 10948637-10948637 on Build GRCh38
rs749674655 Chr.1: 10948566-10948566 on Build GRCh38
rs763027861 Chr.1: 10948606-10948606 on Build GRCh38
rs150987350 Chr.1: 10948705-10948705 on Build GRCh38
rs145944258 Chr.1: 10948541-10948541 on Build GRCh38
rs142322789 Chr.1: 10948626-10948626 on Build GRCh38
rs914338327 Chr.1: 10948617-10948617 on Build GRCh38
rs200317838 Chr.1: 10948627-10948627 on Build GRCh38
rs765001510 Chr.1: 10948604-10948604 on Build GRCh38
rs535570756 Chr.1: 10948696-10948696 on Build GRCh38

View Full Product Details