Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34202045-34202298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
UniGene
Hs.194610
Gene Chromosome Location
Chr.1: 34166883-34219131 on Build GRCh38
Transcripts
NM_001134734.1 NM_032884.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143852 c.1370A>C p.N457T Chr.1: 34202183-34202183 on Build GRCh38
7422504 c.835C>G p.P279A Chr.1: 34202218-34202218 on Build GRCh38
1290190 c.750G>A p.G250G Chr.1: 34202133-34202133 on Build GRCh38
5578168 c.1334C>T p.P445L Chr.1: 34202147-34202147 on Build GRCh38
7358332 c.833C>T p.P278L Chr.1: 34202216-34202216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371336971 Chr.1: 34202202-34202202 on Build GRCh38
rs140823731 Chr.1: 34202171-34202171 on Build GRCh38
rs772138055 Chr.1: 34202165-34202165 on Build GRCh38
rs755371890 Chr.1: 34202094-34202094 on Build GRCh38
rs17556981 Chr.1: 34202125-34202125 on Build GRCh38
rs759896568 Chr.1: 34202162-34202163 on Build GRCh38
rs375505288 Chr.1: 34202119-34202119 on Build GRCh38
rs754997639 Chr.1: 34202260-34202260 on Build GRCh38
rs370193573 Chr.1: 34202153-34202153 on Build GRCh38
rs778757099 Chr.1: 34202254-34202254 on Build GRCh38
rs777086117 Chr.1: 34202169-34202169 on Build GRCh38
rs777515823 Chr.1: 34202159-34202159 on Build GRCh38
rs774712117 Chr.1: 34202160-34202183 on Build GRCh38
rs757921925 Chr.1: 34202122-34202122 on Build GRCh38
rs780665898 Chr.1: 34202176-34202176 on Build GRCh38
rs773231180 Chr.1: 34202152-34202153 on Build GRCh38
rs144339572 Chr.1: 34202100-34202100 on Build GRCh38
rs145574381 Chr.1: 34202201-34202201 on Build GRCh38
rs772188975 Chr.1: 34202181-34202181 on Build GRCh38
rs564773544 Chr.1: 34202232-34202232 on Build GRCh38
rs772544721 Chr.1: 34202167-34202177 on Build GRCh38
rs80015626 Chr.1: 34202183-34202183 on Build GRCh38
rs749952400 Chr.1: 34202121-34202121 on Build GRCh38
rs368292790 Chr.1: 34202120-34202120 on Build GRCh38
rs771047582 Chr.1: 34202157-34202158 on Build GRCh38
rs757218388 Chr.1: 34202249-34202249 on Build GRCh38
rs377265826 Chr.1: 34202127-34202127 on Build GRCh38
rs752291290 Chr.1: 34202236-34202236 on Build GRCh38
rs114850350 Chr.1: 34202244-34202244 on Build GRCh38
rs759549703 Chr.1: 34202089-34202089 on Build GRCh38
rs762015951 Chr.1: 34202170-34202170 on Build GRCh38
rs746009445 Chr.1: 34202164-34202164 on Build GRCh38
rs775731027 Chr.1: 34202167-34202167 on Build GRCh38
rs768928931 Chr.1: 34202187-34202187 on Build GRCh38
rs752553098 Chr.1: 34202178-34202178 on Build GRCh38
rs12746310 Chr.1: 34202262-34202262 on Build GRCh38
rs139397885 Chr.1: 34202135-34202135 on Build GRCh38
rs762617203 Chr.1: 34202173-34202173 on Build GRCh38
rs747845445 Chr.1: 34202150-34202150 on Build GRCh38
rs116349842 Chr.1: 34202233-34202233 on Build GRCh38
rs747435742 Chr.1: 34202084-34202084 on Build GRCh38
rs776657242 Chr.1: 34202193-34202193 on Build GRCh38
rs754855956 Chr.1: 34202134-34202134 on Build GRCh38
rs761099051 Chr.1: 34202099-34202099 on Build GRCh38
rs531268354 Chr.1: 34202218-34202218 on Build GRCh38
rs150896772 Chr.1: 34202123-34202123 on Build GRCh38
rs767743604 Chr.1: 34202091-34202091 on Build GRCh38
rs769657697 Chr.1: 34202148-34202149 on Build GRCh38
rs763528554 Chr.1: 34202221-34202221 on Build GRCh38
rs768712920 Chr.1: 34202168-34202168 on Build GRCh38
rs755982441 Chr.1: 34202240-34202240 on Build GRCh38
rs140728068 Chr.1: 34202231-34202231 on Build GRCh38
rs747103106 Chr.1: 34202259-34202259 on Build GRCh38
rs766941019 Chr.1: 34202227-34202227 on Build GRCh38
rs750952809 Chr.1: 34202172-34202172 on Build GRCh38
rs777521537 Chr.1: 34202243-34202243 on Build GRCh38
rs61741817 Chr.1: 34202088-34202088 on Build GRCh38
rs777837038 Chr.1: 34202180-34202180 on Build GRCh38
rs770963089 Chr.1: 34202070-34202070 on Build GRCh38
rs755851680 Chr.1: 34202179-34202179 on Build GRCh38
rs749278694 Chr.1: 34202157-34202183 on Build GRCh38
rs770238251 Chr.1: 34202268-34202268 on Build GRCh38
rs751301476 Chr.1: 34202174-34202174 on Build GRCh38
rs754698347 Chr.1: 34202175-34202175 on Build GRCh38

View Full Product Details