Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35728672-35728925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL5
Entrez Gene ID
80831
Gene Name
apolipoprotein L5
Gene Aliases
APOL-V, APOLV
UniGene
Hs.326561
Gene Chromosome Location
Chr.22: 35708424-35730279 on Build GRCh38
Transcripts
NM_030642.1 XM_006724321.3 XM_017028945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1033782 c.1167G>T p.Q389H Chr.22: 35728763-35728763 on Build GRCh38
5900838 c.1249C>T p.P417S Chr.22: 35728845-35728845 on Build GRCh38
1161424 c.1144C>T p.P382S Chr.22: 35728740-35728740 on Build GRCh38
7428081 c.1217C>G p.S406C Chr.22: 35728813-35728813 on Build GRCh38
5526814 c.1272G>A p.G424G Chr.22: 35728868-35728868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774540854 Chr.22: 35728747-35728747 on Build GRCh38
rs752353710 Chr.22: 35728880-35728880 on Build GRCh38
rs761942950 Chr.22: 35728834-35728834 on Build GRCh38
rs752157146 Chr.22: 35728796-35728796 on Build GRCh38
rs768827112 Chr.22: 35728733-35728733 on Build GRCh38
rs867564567 Chr.22: 35728823-35728823 on Build GRCh38
rs753644173 Chr.22: 35728781-35728781 on Build GRCh38
rs778576283 Chr.22: 35728746-35728746 on Build GRCh38
rs766268094 Chr.22: 35728868-35728868 on Build GRCh38
rs759275735 Chr.22: 35728878-35728878 on Build GRCh38
rs760656500 Chr.22: 35728858-35728858 on Build GRCh38
rs75305023 Chr.22: 35728843-35728843 on Build GRCh38
rs757370353 Chr.22: 35728740-35728740 on Build GRCh38
rs890841017 Chr.22: 35728793-35728793 on Build GRCh38
rs781681932 Chr.22: 35728717-35728717 on Build GRCh38
rs778391053 Chr.22: 35728711-35728711 on Build GRCh38
rs755423266 Chr.22: 35728904-35728904 on Build GRCh38
rs779255435 Chr.22: 35728831-35728832 on Build GRCh38
rs148906900 Chr.22: 35728849-35728849 on Build GRCh38
rs746397810 Chr.22: 35728806-35728806 on Build GRCh38
rs145495755 Chr.22: 35728728-35728728 on Build GRCh38
rs767511524 Chr.22: 35728757-35728757 on Build GRCh38
rs747699892 Chr.22: 35728712-35728712 on Build GRCh38
rs148034663 Chr.22: 35728891-35728891 on Build GRCh38
rs758801359 Chr.22: 35728893-35728893 on Build GRCh38
rs753544492 Chr.22: 35728706-35728706 on Build GRCh38
rs778591084 Chr.22: 35728791-35728791 on Build GRCh38
rs753648908 Chr.22: 35728869-35728869 on Build GRCh38
rs147690523 Chr.22: 35728749-35728749 on Build GRCh38
rs201193967 Chr.22: 35728832-35728832 on Build GRCh38
rs757843541 Chr.22: 35728889-35728889 on Build GRCh38
rs950220870 Chr.22: 35728709-35728709 on Build GRCh38
rs757937833 Chr.22: 35728799-35728799 on Build GRCh38
rs142361344 Chr.22: 35728763-35728763 on Build GRCh38
rs201846595 Chr.22: 35728786-35728786 on Build GRCh38
rs370966999 Chr.22: 35728800-35728800 on Build GRCh38
rs2076673 Chr.22: 35728813-35728813 on Build GRCh38
rs752841163 Chr.22: 35728840-35728841 on Build GRCh38
rs766183368 Chr.22: 35728771-35728771 on Build GRCh38
rs770196932 Chr.22: 35728726-35728726 on Build GRCh38
rs903449763 Chr.22: 35728745-35728745 on Build GRCh38
rs780458091 Chr.22: 35728821-35728821 on Build GRCh38
rs200999318 Chr.22: 35728769-35728769 on Build GRCh38
rs777960363 Chr.22: 35728868-35728869 on Build GRCh38
rs772292828 Chr.22: 35728848-35728848 on Build GRCh38
rs200371039 Chr.22: 35728831-35728831 on Build GRCh38
rs764832192 Chr.22: 35728879-35728879 on Build GRCh38
rs771410087 Chr.22: 35728713-35728713 on Build GRCh38
rs143581523 Chr.22: 35728890-35728890 on Build GRCh38
rs754644924 Chr.22: 35728707-35728707 on Build GRCh38
rs763219644 Chr.22: 35728729-35728729 on Build GRCh38
rs749613751 Chr.22: 35728899-35728899 on Build GRCh38
rs398102316 Chr.22: 35728846-35728846 on Build GRCh38
rs746193988 Chr.22: 35728723-35728723 on Build GRCh38
rs749653865 Chr.22: 35728826-35728826 on Build GRCh38

View Full Product Details