Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8059687-8059942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3AR1
Entrez Gene ID
719
Gene Name
complement component 3a receptor 1
Gene Aliases
AZ3B, C3AR, HNFAG09
UniGene
Hs.504656
Gene Chromosome Location
Chr.12: 8058323-8066359 on Build GRCh38
Transcripts
NM_001326475.1 NM_001326477.1 NM_004054.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5896162 c.441G>A p.W147* Chr.12: 8059745-8059745 on Build GRCh38
7282579 c.255G>T p.Q85H Chr.12: 8059931-8059931 on Build GRCh38
309718 c.460T>C p.C154R Chr.12: 8059726-8059726 on Build GRCh38
4481651 c.252C>T p.L84L Chr.12: 8059934-8059934 on Build GRCh38
4496034 c.466C>G p.P156A Chr.12: 8059720-8059720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775336929 Chr.12: 8059767-8059767 on Build GRCh38
rs762488984 Chr.12: 8059783-8059783 on Build GRCh38
rs755422554 Chr.12: 8059712-8059712 on Build GRCh38
rs553595145 Chr.12: 8059830-8059831 on Build GRCh38
rs768427252 Chr.12: 8059771-8059771 on Build GRCh38
rs140119047 Chr.12: 8059916-8059916 on Build GRCh38
rs566712249 Chr.12: 8059871-8059871 on Build GRCh38
rs1024893657 Chr.12: 8059754-8059754 on Build GRCh38
rs199741793 Chr.12: 8059870-8059870 on Build GRCh38
rs938770801 Chr.12: 8059816-8059816 on Build GRCh38
rs968601984 Chr.12: 8059811-8059811 on Build GRCh38
rs768480392 Chr.12: 8059861-8059861 on Build GRCh38
rs746946191 Chr.12: 8059860-8059860 on Build GRCh38
rs869312973 Chr.12: 8059829-8059830 on Build GRCh38
rs138306314 Chr.12: 8059846-8059846 on Build GRCh38
rs780026701 Chr.12: 8059851-8059851 on Build GRCh38
rs1010451100 Chr.12: 8059738-8059738 on Build GRCh38
rs776413209 Chr.12: 8059775-8059775 on Build GRCh38
rs772027352 Chr.12: 8059762-8059762 on Build GRCh38
rs976554031 Chr.12: 8059727-8059727 on Build GRCh38
rs146137267 Chr.12: 8059915-8059915 on Build GRCh38
rs977673489 Chr.12: 8059863-8059863 on Build GRCh38
rs762606886 Chr.12: 8059900-8059900 on Build GRCh38
rs576903115 Chr.12: 8059833-8059833 on Build GRCh38
rs769366782 Chr.12: 8059872-8059872 on Build GRCh38
rs761184871 Chr.12: 8059809-8059809 on Build GRCh38
rs776176515 Chr.12: 8059907-8059907 on Build GRCh38
rs761341417 Chr.12: 8059776-8059776 on Build GRCh38
rs772856114 Chr.12: 8059878-8059878 on Build GRCh38
rs746821514 Chr.12: 8059770-8059770 on Build GRCh38
rs772623748 Chr.12: 8059781-8059781 on Build GRCh38
rs761924608 Chr.12: 8059729-8059729 on Build GRCh38
rs11567812 Chr.12: 8059721-8059721 on Build GRCh38
rs965135147 Chr.12: 8059725-8059725 on Build GRCh38
rs533727474 Chr.12: 8059761-8059761 on Build GRCh38
rs984437173 Chr.12: 8059791-8059791 on Build GRCh38
rs778196171 Chr.12: 8059728-8059728 on Build GRCh38
rs141615526 Chr.12: 8059735-8059735 on Build GRCh38
rs142478022 Chr.12: 8059715-8059715 on Build GRCh38
rs932513196 Chr.12: 8059753-8059753 on Build GRCh38
rs61736560 Chr.12: 8059904-8059904 on Build GRCh38
rs907502587 Chr.12: 8059814-8059814 on Build GRCh38
rs765189903 Chr.12: 8059845-8059845 on Build GRCh38
rs750645866 Chr.12: 8059921-8059921 on Build GRCh38
rs745911291 Chr.12: 8059757-8059757 on Build GRCh38
rs754135233 Chr.12: 8059821-8059821 on Build GRCh38
rs760997627 Chr.12: 8059827-8059827 on Build GRCh38
rs776909737 Chr.12: 8059747-8059747 on Build GRCh38
rs773999327 Chr.12: 8059828-8059828 on Build GRCh38
rs754185116 Chr.12: 8059910-8059910 on Build GRCh38
rs764519677 Chr.12: 8059820-8059820 on Build GRCh38
rs61736561 Chr.12: 8059786-8059786 on Build GRCh38
rs202184221 Chr.12: 8059909-8059909 on Build GRCh38
rs756745490 Chr.12: 8059785-8059785 on Build GRCh38
rs753145243 Chr.12: 8059718-8059718 on Build GRCh38
rs780827622 Chr.12: 8059864-8059864 on Build GRCh38
rs956502987 Chr.12: 8059733-8059733 on Build GRCh38
rs758711287 Chr.12: 8059711-8059711 on Build GRCh38
rs11567806 Chr.12: 8059779-8059779 on Build GRCh38
rs1050232636 Chr.12: 8059897-8059897 on Build GRCh38

View Full Product Details