Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 173358613-173358886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA7
Entrez Gene ID
83879
Gene Name
cell division cycle associated 7
Gene Aliases
ICF3, JPO1
UniGene
Hs.470654
Gene Chromosome Location
Chr.2: 173354017-173368992 on Build GRCh38
Transcripts
NM_031942.4 NM_145810.2 XM_011511957.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6337467 c.79T>C p.S27P Chr.2: 173358769-173358769 on Build GRCh38
4952319 c.145A>T p.T49S Chr.2: 173358835-173358835 on Build GRCh38
1683880 c.45_47delGAA p.K15delK Chr.2: 173358735-173358737 on Build GRCh38
1752109 c.141A>G p.A47A Chr.2: 173358831-173358831 on Build GRCh38
6010392 c.108C>T p.D36D Chr.2: 173358798-173358798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142079460 Chr.2: 173358783-173358783 on Build GRCh38
rs964226690 Chr.2: 173358791-173358791 on Build GRCh38
rs758866134 Chr.2: 173358836-173358836 on Build GRCh38
rs764552233 Chr.2: 173358668-173358668 on Build GRCh38
rs779015155 Chr.2: 173358715-173358715 on Build GRCh38
rs751164512 Chr.2: 173358806-173358806 on Build GRCh38
rs933931768 Chr.2: 173358844-173358844 on Build GRCh38
rs762628426 Chr.2: 173358801-173358801 on Build GRCh38
rs759103879 Chr.2: 173358816-173358816 on Build GRCh38
rs532245371 Chr.2: 173358789-173358789 on Build GRCh38
rs770979057 Chr.2: 173358730-173358732 on Build GRCh38
rs765866818 Chr.2: 173358700-173358700 on Build GRCh38
rs12989017 Chr.2: 173358744-173358744 on Build GRCh38
rs749407514 Chr.2: 173358667-173358667 on Build GRCh38
rs769206471 Chr.2: 173358772-173358772 on Build GRCh38
rs113082218 Chr.2: 173358747-173358747 on Build GRCh38
rs146641750 Chr.2: 173358763-173358763 on Build GRCh38
rs781254734 Chr.2: 173358709-173358709 on Build GRCh38
rs777114913 Chr.2: 173358782-173358782 on Build GRCh38
rs777581752 Chr.2: 173358841-173358841 on Build GRCh38
rs1024987115 Chr.2: 173358661-173358661 on Build GRCh38
rs778918232 Chr.2: 173358741-173358744 on Build GRCh38
rs757863320 Chr.2: 173358698-173358698 on Build GRCh38
rs189447098 Chr.2: 173358683-173358683 on Build GRCh38
rs746053753 Chr.2: 173358720-173358720 on Build GRCh38
rs894676218 Chr.2: 173358656-173358656 on Build GRCh38
rs1013130291 Chr.2: 173358658-173358658 on Build GRCh38
rs770141916 Chr.2: 173358788-173358788 on Build GRCh38
rs752945397 Chr.2: 173358663-173358664 on Build GRCh38
rs777965222 Chr.2: 173358667-173358671 on Build GRCh38
rs777027005 Chr.2: 173358675-173358675 on Build GRCh38
rs1003229238 Chr.2: 173358686-173358686 on Build GRCh38
rs755933689 Chr.2: 173358711-173358711 on Build GRCh38
rs781623087 Chr.2: 173358663-173358666 on Build GRCh38
rs918503928 Chr.2: 173358640-173358641 on Build GRCh38
rs752597450 Chr.2: 173358710-173358710 on Build GRCh38
rs752614678 Chr.2: 173358823-173358823 on Build GRCh38
rs751581799 Chr.2: 173358704-173358704 on Build GRCh38
rs750685249 Chr.2: 173358760-173358760 on Build GRCh38
rs113611019 Chr.2: 173358695-173358695 on Build GRCh38
rs753545956 Chr.2: 173358851-173358851 on Build GRCh38
rs140240952 Chr.2: 173358803-173358803 on Build GRCh38
rs758694596 Chr.2: 173358862-173358862 on Build GRCh38
rs931281365 Chr.2: 173358641-173358641 on Build GRCh38
rs552244926 Chr.2: 173358706-173358706 on Build GRCh38
rs139753635 Chr.2: 173358735-173358735 on Build GRCh38

View Full Product Details