Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57806451-57806676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
UniGene
Hs.584854
Gene Chromosome Location
Chr.12: 57797372-57818704 on Build GRCh38
Transcripts
NM_006576.3 XM_017018710.1 XM_017018711.1 XM_017018712.1 XM_017018713.1 XM_017018714.1 XM_017018715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2156263 c.1574C>G p.T525S Chr.12: 57806457-57806457 on Build GRCh38
2156262 c.1573A>T p.T525S Chr.12: 57806458-57806458 on Build GRCh38
2156252 c.1568C>A p.S523Y Chr.12: 57806463-57806463 on Build GRCh38
1363448 c.1565A>C p.K522T Chr.12: 57806466-57806466 on Build GRCh38
1989944 c.1575C>A p.T525T Chr.12: 57806456-57806456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048351035 Chr.12: 57806534-57806534 on Build GRCh38
rs983632064 Chr.12: 57806520-57806520 on Build GRCh38
rs750487437 Chr.12: 57806509-57806509 on Build GRCh38
rs779545561 Chr.12: 57806552-57806552 on Build GRCh38
rs762043345 Chr.12: 57806494-57806494 on Build GRCh38
rs771805994 Chr.12: 57806551-57806551 on Build GRCh38
rs776512313 Chr.12: 57806579-57806579 on Build GRCh38
rs1035573250 Chr.12: 57806644-57806644 on Build GRCh38
rs552650655 Chr.12: 57806526-57806526 on Build GRCh38
rs376322015 Chr.12: 57806576-57806576 on Build GRCh38
rs886646616 Chr.12: 57806566-57806566 on Build GRCh38
rs746892386 Chr.12: 57806570-57806570 on Build GRCh38
rs769802237 Chr.12: 57806584-57806584 on Build GRCh38
rs367900033 Chr.12: 57806512-57806512 on Build GRCh38
rs1052236272 Chr.12: 57806609-57806609 on Build GRCh38
rs771540578 Chr.12: 57806538-57806538 on Build GRCh38
rs368889532 Chr.12: 57806587-57806587 on Build GRCh38
rs756548425 Chr.12: 57806542-57806542 on Build GRCh38
rs150473391 Chr.12: 57806501-57806501 on Build GRCh38
rs762907011 Chr.12: 57806588-57806588 on Build GRCh38
rs1023322969 Chr.12: 57806643-57806643 on Build GRCh38
rs187759826 Chr.12: 57806617-57806617 on Build GRCh38
rs768285344 Chr.12: 57806582-57806583 on Build GRCh38
rs777062181 Chr.12: 57806608-57806608 on Build GRCh38
rs778427686 Chr.12: 57806544-57806544 on Build GRCh38
rs113363824 Chr.12: 57806627-57806627 on Build GRCh38
rs771131780 Chr.12: 57806543-57806544 on Build GRCh38
rs372000272 Chr.12: 57806539-57806539 on Build GRCh38
rs766484991 Chr.12: 57806513-57806513 on Build GRCh38
rs755293608 Chr.12: 57806529-57806529 on Build GRCh38
rs745314355 Chr.12: 57806548-57806548 on Build GRCh38

View Full Product Details