Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136996318-136996543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC3
Entrez Gene ID
9022
Gene Name
chloride intracellular channel 3
Gene Aliases
-
UniGene
Hs.64746
Gene Chromosome Location
Chr.9: 136994608-136997929 on Build GRCh38
Transcripts
NM_004669.2 XM_017015281.1 XM_017015282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461271 c.6G>A p.A2A Chr.9: 136996538-136996538 on Build GRCh38
7187460 c.22C>G p.L8V Chr.9: 136996522-136996522 on Build GRCh38
4518561 c.6_7GG>AA p.E3K Chr.9: 136996537-136996538 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765902448 Chr.9: 136996502-136996502 on Build GRCh38
rs777315086 Chr.9: 136996524-136996524 on Build GRCh38
rs969400293 Chr.9: 136996451-136996451 on Build GRCh38
rs377313643 Chr.9: 136996467-136996467 on Build GRCh38
rs754185672 Chr.9: 136996495-136996495 on Build GRCh38
rs753242320 Chr.9: 136996510-136996510 on Build GRCh38
rs545096923 Chr.9: 136996421-136996421 on Build GRCh38
rs1048631090 Chr.9: 136996438-136996438 on Build GRCh38
rs747585856 Chr.9: 136996463-136996463 on Build GRCh38
rs560740663 Chr.9: 136996485-136996485 on Build GRCh38
rs1024087395 Chr.9: 136996429-136996429 on Build GRCh38
rs897275629 Chr.9: 136996429-136996429 on Build GRCh38
rs564792973 Chr.9: 136996450-136996450 on Build GRCh38
rs569271517 Chr.9: 136996385-136996385 on Build GRCh38
rs908834 Chr.9: 136996466-136996466 on Build GRCh38
rs994243067 Chr.9: 136996432-136996432 on Build GRCh38
rs2292922 Chr.9: 136996489-136996489 on Build GRCh38
rs200825202 Chr.9: 136996490-136996490 on Build GRCh38
rs376163123 Chr.9: 136996481-136996481 on Build GRCh38
rs757885849 Chr.9: 136996511-136996511 on Build GRCh38
rs951639300 Chr.9: 136996476-136996476 on Build GRCh38
rs957977456 Chr.9: 136996408-136996408 on Build GRCh38
rs572829758 Chr.9: 136996392-136996392 on Build GRCh38
rs944057731 Chr.9: 136996412-136996412 on Build GRCh38
rs199613987 Chr.9: 136996482-136996482 on Build GRCh38
rs560125561 Chr.9: 136996501-136996501 on Build GRCh38
rs999323369 Chr.9: 136996338-136996338 on Build GRCh38

View Full Product Details