Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19188087-19188350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP3
Entrez Gene ID
8048
Gene Name
cysteine and glycine rich protein 3
Gene Aliases
CLP, CMD1M, CMH12, CRP3, LMO4, MLP
UniGene
Hs.83577
Gene Chromosome Location
Chr.11: 19182030-19210571 on Build GRCh38
Transcripts
NM_003476.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1353124 c.213C>T p.I71I Chr.11: 19188204-19188204 on Build GRCh38
1353125 c.141G>A p.T47T Chr.11: 19188276-19188276 on Build GRCh38
5626580 c.270G>A p.L90L Chr.11: 19188147-19188147 on Build GRCh38
1718533 c.256G>A p.E86K Chr.11: 19188161-19188161 on Build GRCh38
2093296 c.191G>A p.R64H Chr.11: 19188226-19188226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112884796 Chr.11: 19188297-19188297 on Build GRCh38
rs777211110 Chr.11: 19188209-19188209 on Build GRCh38
rs145300736 Chr.11: 19188269-19188269 on Build GRCh38
rs894467251 Chr.11: 19188284-19188284 on Build GRCh38
rs759455306 Chr.11: 19188256-19188256 on Build GRCh38
rs727504436 Chr.11: 19188145-19188145 on Build GRCh38
rs539656337 Chr.11: 19188212-19188212 on Build GRCh38
rs748417030 Chr.11: 19188301-19188301 on Build GRCh38
rs137852764 Chr.11: 19188211-19188211 on Build GRCh38
rs982949558 Chr.11: 19188158-19188158 on Build GRCh38
rs137852765 Chr.11: 19188281-19188281 on Build GRCh38
rs1020735963 Chr.11: 19188261-19188261 on Build GRCh38
rs769003538 Chr.11: 19188242-19188242 on Build GRCh38
rs367827746 Chr.11: 19188152-19188152 on Build GRCh38
rs1047686575 Chr.11: 19188186-19188186 on Build GRCh38
rs45476991 Chr.11: 19188204-19188204 on Build GRCh38
rs139805841 Chr.11: 19188268-19188268 on Build GRCh38
rs920763927 Chr.11: 19188299-19188299 on Build GRCh38
rs34075817 Chr.11: 19188234-19188234 on Build GRCh38
rs755784426 Chr.11: 19188205-19188205 on Build GRCh38
rs773218235 Chr.11: 19188310-19188310 on Build GRCh38
rs45552933 Chr.11: 19188203-19188203 on Build GRCh38
rs752452689 Chr.11: 19188264-19188264 on Build GRCh38
rs772117382 Chr.11: 19188201-19188201 on Build GRCh38
rs760832923 Chr.11: 19188148-19188148 on Build GRCh38
rs777327517 Chr.11: 19188166-19188166 on Build GRCh38
rs752338221 Chr.11: 19188162-19188162 on Build GRCh38
rs769986102 Chr.11: 19188303-19188303 on Build GRCh38
rs374764059 Chr.11: 19188248-19188248 on Build GRCh38
rs796360127 Chr.11: 19188304-19188304 on Build GRCh38
rs557643642 Chr.11: 19188216-19188216 on Build GRCh38
rs963128995 Chr.11: 19188184-19188184 on Build GRCh38
rs774366633 Chr.11: 19188147-19188147 on Build GRCh38
rs767360228 Chr.11: 19188249-19188249 on Build GRCh38
rs765287458 Chr.11: 19188131-19188131 on Build GRCh38
rs779421059 Chr.11: 19188219-19188219 on Build GRCh38
rs772138289 Chr.11: 19188187-19188187 on Build GRCh38
rs758256351 Chr.11: 19188153-19188153 on Build GRCh38
rs375014380 Chr.11: 19188226-19188226 on Build GRCh38
rs768767452 Chr.11: 19188189-19188189 on Build GRCh38
rs747363563 Chr.11: 19188279-19188279 on Build GRCh38
rs773476497 Chr.11: 19188156-19188156 on Build GRCh38
rs758842207 Chr.11: 19188276-19188276 on Build GRCh38
rs397516854 Chr.11: 19188220-19188220 on Build GRCh38
rs7124801 Chr.11: 19188267-19188267 on Build GRCh38
rs751348376 Chr.11: 19188201-19188201 on Build GRCh38
rs182798086 Chr.11: 19188159-19188159 on Build GRCh38
rs368392588 Chr.11: 19188227-19188227 on Build GRCh38
rs767787661 Chr.11: 19188135-19188135 on Build GRCh38
rs771444930 Chr.11: 19188177-19188177 on Build GRCh38
rs886041190 Chr.11: 19188293-19188294 on Build GRCh38
rs779184695 Chr.11: 19188179-19188179 on Build GRCh38
rs112848043 Chr.11: 19188255-19188255 on Build GRCh38
rs267606753 Chr.11: 19188253-19188253 on Build GRCh38
rs576346189 Chr.11: 19188270-19188270 on Build GRCh38
rs199793773 Chr.11: 19188181-19188181 on Build GRCh38
rs866941870 Chr.11: 19188236-19188236 on Build GRCh38
rs104894205 Chr.11: 19188286-19188286 on Build GRCh38
rs746207 Chr.11: 19188188-19188188 on Build GRCh38
rs755614464 Chr.11: 19188165-19188165 on Build GRCh38
rs397516852 Chr.11: 19188198-19188198 on Build GRCh38
rs767326021 Chr.11: 19188262-19188262 on Build GRCh38
rs759699327 Chr.11: 19188125-19188125 on Build GRCh38
rs780917755 Chr.11: 19188161-19188161 on Build GRCh38
rs770041119 Chr.11: 19188251-19188251 on Build GRCh38
rs397516853 Chr.11: 19188265-19188265 on Build GRCh38
rs104894204 Chr.11: 19188245-19188245 on Build GRCh38
rs762915899 Chr.11: 19188195-19188195 on Build GRCh38
rs776862812 Chr.11: 19188194-19188194 on Build GRCh38
rs281865416 Chr.11: 19188253-19188257 on Build GRCh38
rs397516851 Chr.11: 19188277-19188277 on Build GRCh38

View Full Product Details