Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 51930921-51931190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA10
Entrez Gene ID
56934
Gene Name
carbonic anhydrase 10
Gene Aliases
CA-RPX, CARPX, HUCEP-15
UniGene
Hs.463466
Gene Chromosome Location
Chr.17: 51630313-52160017 on Build GRCh38
Transcripts
NM_001082533.1 NM_001082534.1 NM_020178.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3958550 c.293G>T p.R98M Chr.17: 51930994-51930994 on Build GRCh38
4859402 c.246G>T p.M82I Chr.17: 51931041-51931041 on Build GRCh38
3889965 c.207G>A p.G69G Chr.17: 51931080-51931080 on Build GRCh38
4858881 c.290G>T p.G97V Chr.17: 51930997-51930997 on Build GRCh38
6617080 c.278T>C p.I93T Chr.17: 51931009-51931009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768447818 Chr.17: 51930959-51930959 on Build GRCh38
rs746462691 Chr.17: 51930951-51930951 on Build GRCh38
rs866323451 Chr.17: 51931120-51931120 on Build GRCh38
rs150982272 Chr.17: 51931075-51931075 on Build GRCh38
rs916246853 Chr.17: 51930997-51930997 on Build GRCh38
rs760815614 Chr.17: 51931169-51931169 on Build GRCh38
rs147775605 Chr.17: 51931005-51931005 on Build GRCh38
rs770874732 Chr.17: 51931140-51931140 on Build GRCh38
rs759648220 Chr.17: 51931146-51931146 on Build GRCh38
rs200191508 Chr.17: 51931152-51931152 on Build GRCh38
rs769454483 Chr.17: 51931000-51931000 on Build GRCh38
rs373462624 Chr.17: 51931019-51931019 on Build GRCh38
rs148839711 Chr.17: 51931012-51931012 on Build GRCh38
rs779515865 Chr.17: 51931065-51931065 on Build GRCh38
rs867369783 Chr.17: 51931110-51931110 on Build GRCh38
rs267604958 Chr.17: 51931041-51931041 on Build GRCh38
rs377394275 Chr.17: 51931057-51931057 on Build GRCh38
rs775852360 Chr.17: 51931153-51931153 on Build GRCh38
rs767808441 Chr.17: 51931013-51931013 on Build GRCh38
rs375676352 Chr.17: 51931142-51931142 on Build GRCh38
rs200959325 Chr.17: 51930950-51930950 on Build GRCh38
rs369235836 Chr.17: 51931116-51931116 on Build GRCh38
rs748016552 Chr.17: 51930987-51930987 on Build GRCh38
rs780948449 Chr.17: 51931076-51931076 on Build GRCh38
rs372980426 Chr.17: 51931125-51931125 on Build GRCh38
rs138454442 Chr.17: 51931046-51931046 on Build GRCh38
rs556166013 Chr.17: 51931002-51931002 on Build GRCh38
rs764365763 Chr.17: 51931044-51931044 on Build GRCh38
rs144171943 Chr.17: 51931003-51931003 on Build GRCh38
rs1042246688 Chr.17: 51931030-51931030 on Build GRCh38
rs371674596 Chr.17: 51930960-51930960 on Build GRCh38
rs375717497 Chr.17: 51931018-51931018 on Build GRCh38
rs935060725 Chr.17: 51931050-51931050 on Build GRCh38
rs769305570 Chr.17: 51931006-51931006 on Build GRCh38
rs1012255413 Chr.17: 51931069-51931069 on Build GRCh38
rs201633277 Chr.17: 51931035-51931035 on Build GRCh38
rs749022886 Chr.17: 51931136-51931136 on Build GRCh38
rs143498531 Chr.17: 51931068-51931068 on Build GRCh38
rs747743270 Chr.17: 51931113-51931113 on Build GRCh38

View Full Product Details