Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200807187-200807434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
UniGene
Hs.23585
Gene Chromosome Location
Chr.1: 200738905-200860707 on Build GRCh38
Transcripts
NM_001297707.1 NM_001297708.1 NM_203459.2 XM_005245041.3 XM_017000799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3976816 c.400-2A>G p.? Chr.1: 200807374-200807374 on Build GRCh38
5483699 c.411G>A p.L137L Chr.1: 200807387-200807387 on Build GRCh38
7290084 c.431T>C p.M144T Chr.1: 200807407-200807407 on Build GRCh38
901988 c.402T>C p.S134S Chr.1: 200807378-200807378 on Build GRCh38
3976815 c.400-2A>G p.? Chr.1: 200807374-200807374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888613343 Chr.1: 200807275-200807275 on Build GRCh38
rs745548085 Chr.1: 200807366-200807366 on Build GRCh38
rs758271769 Chr.1: 200807340-200807340 on Build GRCh38
rs183164262 Chr.1: 200807317-200807317 on Build GRCh38
rs769381640 Chr.1: 200807367-200807367 on Build GRCh38
rs200778331 Chr.1: 200807368-200807368 on Build GRCh38
rs750467755 Chr.1: 200807327-200807327 on Build GRCh38
rs780126552 Chr.1: 200807335-200807335 on Build GRCh38
rs767889233 Chr.1: 200807228-200807228 on Build GRCh38
rs556798899 Chr.1: 200807339-200807339 on Build GRCh38
rs756339866 Chr.1: 200807401-200807401 on Build GRCh38
rs541714051 Chr.1: 200807328-200807328 on Build GRCh38
rs774768517 Chr.1: 200807379-200807379 on Build GRCh38
rs79456176 Chr.1: 200807372-200807372 on Build GRCh38
rs201642655 Chr.1: 200807360-200807360 on Build GRCh38
rs762120282 Chr.1: 200807390-200807390 on Build GRCh38
rs777618456 Chr.1: 200807343-200807343 on Build GRCh38
rs754308936 Chr.1: 200807352-200807355 on Build GRCh38
rs746836477 Chr.1: 200807350-200807350 on Build GRCh38
rs765613601 Chr.1: 200807355-200807355 on Build GRCh38
rs767895767 Chr.1: 200807391-200807391 on Build GRCh38
rs553072504 Chr.1: 200807304-200807304 on Build GRCh38
rs750592470 Chr.1: 200807396-200807396 on Build GRCh38
rs993178430 Chr.1: 200807351-200807351 on Build GRCh38
rs1004349085 Chr.1: 200807285-200807285 on Build GRCh38
rs910454067 Chr.1: 200807258-200807258 on Build GRCh38

View Full Product Details