Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86395241-86395514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC81
Entrez Gene ID
60494
Gene Name
coiled-coil domain containing 81
Gene Aliases
-
UniGene
Hs.144913
Gene Chromosome Location
Chr.11: 86374736-86423109 on Build GRCh38
Transcripts
NM_001156474.1 NM_021827.4 XM_005274161.1 XM_005274162.3 XM_017018105.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1605317 c.558G>T p.R186S Chr.11: 86395336-86395336 on Build GRCh38
4037228 c.299T>C p.V100A Chr.11: 86395347-86395347 on Build GRCh38
7173935 c.320G>A p.R107K Chr.11: 86395368-86395368 on Build GRCh38
7173934 c.590G>A p.R197K Chr.11: 86395368-86395368 on Build GRCh38
4037227 c.569T>C p.V190A Chr.11: 86395347-86395347 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771789221 Chr.11: 86395324-86395324 on Build GRCh38
rs772783052 Chr.11: 86395325-86395325 on Build GRCh38
rs771813240 Chr.11: 86395369-86395369 on Build GRCh38
rs962851661 Chr.11: 86395310-86395310 on Build GRCh38
rs909419974 Chr.11: 86395267-86395267 on Build GRCh38
rs757599239 Chr.11: 86395349-86395349 on Build GRCh38
rs758211040 Chr.11: 86395434-86395434 on Build GRCh38
rs764660941 Chr.11: 86395385-86395385 on Build GRCh38
rs377360756 Chr.11: 86395404-86395404 on Build GRCh38
rs200850297 Chr.11: 86395284-86395284 on Build GRCh38
rs199619761 Chr.11: 86395335-86395335 on Build GRCh38
rs778801750 Chr.11: 86395429-86395429 on Build GRCh38
rs752000386 Chr.11: 86395347-86395347 on Build GRCh38
rs768373717 Chr.11: 86395298-86395298 on Build GRCh38
rs562183450 Chr.11: 86395375-86395375 on Build GRCh38
rs927542073 Chr.11: 86395319-86395319 on Build GRCh38
rs374647566 Chr.11: 86395329-86395329 on Build GRCh38
rs770411800 Chr.11: 86395374-86395374 on Build GRCh38
rs760616440 Chr.11: 86395353-86395353 on Build GRCh38
rs73517949 Chr.11: 86395371-86395371 on Build GRCh38
rs369807010 Chr.11: 86395282-86395282 on Build GRCh38
rs762221484 Chr.11: 86395396-86395396 on Build GRCh38
rs780001823 Chr.11: 86395354-86395354 on Build GRCh38
rs201607444 Chr.11: 86395393-86395393 on Build GRCh38
rs377628889 Chr.11: 86395427-86395427 on Build GRCh38
rs772908422 Chr.11: 86395370-86395370 on Build GRCh38
rs754931533 Chr.11: 86395358-86395358 on Build GRCh38
rs752538603 Chr.11: 86395411-86395411 on Build GRCh38
rs1043180434 Chr.11: 86395452-86395452 on Build GRCh38
rs762864821 Chr.11: 86395418-86395418 on Build GRCh38
rs1043857008 Chr.11: 86395477-86395477 on Build GRCh38
rs778640139 Chr.11: 86395360-86395360 on Build GRCh38
rs749289321 Chr.11: 86395297-86395297 on Build GRCh38
rs764499459 Chr.11: 86395345-86395345 on Build GRCh38
rs764031242 Chr.11: 86395487-86395487 on Build GRCh38
rs777436667 Chr.11: 86395444-86395444 on Build GRCh38
rs765226842 Chr.11: 86395377-86395377 on Build GRCh38
rs748095018 Chr.11: 86395432-86395432 on Build GRCh38
rs147436959 Chr.11: 86395378-86395378 on Build GRCh38
rs539788278 Chr.11: 86395405-86395405 on Build GRCh38
rs781451099 Chr.11: 86395350-86395350 on Build GRCh38
rs763563308 Chr.11: 86395339-86395339 on Build GRCh38
rs753255217 Chr.11: 86395331-86395331 on Build GRCh38
rs761407331 Chr.11: 86395315-86395315 on Build GRCh38
rs946524593 Chr.11: 86395471-86395471 on Build GRCh38
rs369882372 Chr.11: 86395327-86395327 on Build GRCh38
rs568934403 Chr.11: 86395355-86395355 on Build GRCh38
rs141376644 Chr.11: 86395287-86395287 on Build GRCh38
rs529341057 Chr.11: 86395446-86395447 on Build GRCh38
rs746618488 Chr.11: 86395447-86395447 on Build GRCh38
rs763510233 Chr.11: 86395381-86395381 on Build GRCh38
rs760212492 Chr.11: 86395328-86395328 on Build GRCh38
rs756302629 Chr.11: 86395406-86395406 on Build GRCh38
rs764027357 Chr.11: 86395398-86395398 on Build GRCh38

View Full Product Details