Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83299019-83299284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD3
Entrez Gene ID
257019
Gene Name
FERM domain containing 3
Gene Aliases
4.1O, EPB41L4O, EPB41LO, P410
UniGene
Hs.127535
Gene Chromosome Location
Chr.9: 83242990-83560208 on Build GRCh38
Transcripts
NM_001244959.1 NM_001244960.1 NM_001244961.1 NM_174938.5 XM_017014588.1 XM_017014589.1 XM_017014590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110581 c.632A>C p.Q211P Chr.9: 83299169-83299169 on Build GRCh38
1110579 c.682C>T p.R228* Chr.9: 83299119-83299119 on Build GRCh38
6183846 c.377C>G p.S126* Chr.9: 83299154-83299154 on Build GRCh38
5571213 c.643G>A p.V215I Chr.9: 83299158-83299158 on Build GRCh38
6666909 c.666delT p.F222fs*44 Chr.9: 83299135-83299135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761877197 Chr.9: 83299184-83299184 on Build GRCh38
rs773878690 Chr.9: 83299193-83299193 on Build GRCh38
rs774834059 Chr.9: 83299074-83299074 on Build GRCh38
rs774712672 Chr.9: 83299070-83299070 on Build GRCh38
rs777309279 Chr.9: 83299174-83299174 on Build GRCh38
rs759952321 Chr.9: 83299075-83299075 on Build GRCh38
rs750626841 Chr.9: 83299221-83299221 on Build GRCh38
rs771078003 Chr.9: 83299074-83299074 on Build GRCh38
rs768069610 Chr.9: 83299085-83299085 on Build GRCh38
rs928307994 Chr.9: 83299244-83299244 on Build GRCh38
rs748109321 Chr.9: 83299156-83299156 on Build GRCh38
rs150556834 Chr.9: 83299180-83299180 on Build GRCh38
rs752605019 Chr.9: 83299230-83299230 on Build GRCh38
rs899791213 Chr.9: 83299064-83299064 on Build GRCh38
rs776939619 Chr.9: 83299194-83299194 on Build GRCh38
rs142582268 Chr.9: 83299150-83299150 on Build GRCh38
rs747015951 Chr.9: 83299069-83299069 on Build GRCh38
rs761305213 Chr.9: 83299205-83299205 on Build GRCh38
rs368061183 Chr.9: 83299207-83299207 on Build GRCh38
rs776098844 Chr.9: 83299201-83299201 on Build GRCh38
rs572303503 Chr.9: 83299222-83299222 on Build GRCh38
rs761938334 Chr.9: 83299217-83299217 on Build GRCh38
rs779701415 Chr.9: 83299141-83299141 on Build GRCh38
rs756098484 Chr.9: 83299233-83299233 on Build GRCh38
rs551143107 Chr.9: 83299103-83299103 on Build GRCh38
rs895548781 Chr.9: 83299247-83299247 on Build GRCh38
rs139594830 Chr.9: 83299186-83299186 on Build GRCh38
rs767058585 Chr.9: 83299153-83299153 on Build GRCh38
rs757548840 Chr.9: 83299140-83299140 on Build GRCh38
rs776831942 Chr.9: 83299209-83299209 on Build GRCh38
rs368454551 Chr.9: 83299087-83299087 on Build GRCh38
rs370741927 Chr.9: 83299119-83299119 on Build GRCh38
rs769259390 Chr.9: 83299166-83299166 on Build GRCh38
rs746183365 Chr.9: 83299194-83299194 on Build GRCh38
rs375118684 Chr.9: 83299073-83299073 on Build GRCh38
rs772587451 Chr.9: 83299198-83299198 on Build GRCh38
rs754083167 Chr.9: 83299133-83299133 on Build GRCh38
rs762703661 Chr.9: 83299071-83299071 on Build GRCh38
rs569434487 Chr.9: 83299144-83299144 on Build GRCh38
rs554456430 Chr.9: 83299220-83299220 on Build GRCh38

View Full Product Details