Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 451218-451477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_011534748.2 XM_011534749.1 XM_017013313.1 XM_017013314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6357353 c.494A>G p.N165S Chr.8: 451287-451287 on Build GRCh38
750377 c.507C>G p.I169M Chr.8: 451300-451300 on Build GRCh38
3900135 c.624C>T p.A208A Chr.8: 451417-451417 on Build GRCh38
486466 c.603C>T p.C201C Chr.8: 451396-451396 on Build GRCh38
6113402 c.517C>G p.L173V Chr.8: 451310-451310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933954912 Chr.8: 451313-451313 on Build GRCh38
rs757215791 Chr.8: 451432-451432 on Build GRCh38
rs752239783 Chr.8: 451256-451256 on Build GRCh38
rs375152662 Chr.8: 451250-451250 on Build GRCh38
rs373138185 Chr.8: 451334-451334 on Build GRCh38
rs747562890 Chr.8: 451426-451426 on Build GRCh38
rs200670984 Chr.8: 451445-451445 on Build GRCh38
rs111290397 Chr.8: 451371-451371 on Build GRCh38
rs368217727 Chr.8: 451251-451251 on Build GRCh38
rs763639285 Chr.8: 451329-451329 on Build GRCh38
rs756934535 Chr.8: 451335-451335 on Build GRCh38
rs867652617 Chr.8: 451378-451378 on Build GRCh38
rs201151069 Chr.8: 451302-451302 on Build GRCh38
rs755783388 Chr.8: 451407-451407 on Build GRCh38
rs200119344 Chr.8: 451423-451423 on Build GRCh38
rs150976957 Chr.8: 451439-451439 on Build GRCh38
rs761364341 Chr.8: 451415-451415 on Build GRCh38
rs558624439 Chr.8: 451362-451362 on Build GRCh38
rs747124599 Chr.8: 451375-451375 on Build GRCh38
rs145775965 Chr.8: 451350-451350 on Build GRCh38
rs368705954 Chr.8: 451417-451417 on Build GRCh38
rs765135269 Chr.8: 451316-451316 on Build GRCh38
rs770181228 Chr.8: 451437-451437 on Build GRCh38
rs144227759 Chr.8: 451398-451398 on Build GRCh38
rs148503218 Chr.8: 451383-451383 on Build GRCh38
rs144159684 Chr.8: 451436-451436 on Build GRCh38
rs775530412 Chr.8: 451321-451321 on Build GRCh38
rs200227738 Chr.8: 451258-451258 on Build GRCh38
rs773998501 Chr.8: 451403-451403 on Build GRCh38
rs113945607 Chr.8: 451429-451429 on Build GRCh38
rs760924947 Chr.8: 451312-451312 on Build GRCh38
rs374460400 Chr.8: 451409-451409 on Build GRCh38
rs146754998 Chr.8: 451400-451400 on Build GRCh38
rs200806337 Chr.8: 451414-451414 on Build GRCh38
rs769141075 Chr.8: 451290-451290 on Build GRCh38
rs753620470 Chr.8: 451248-451248 on Build GRCh38
rs775475294 Chr.8: 451394-451394 on Build GRCh38
rs1004702055 Chr.8: 451319-451319 on Build GRCh38
rs777874042 Chr.8: 451366-451366 on Build GRCh38
rs149790925 Chr.8: 451330-451330 on Build GRCh38
rs759460552 Chr.8: 451315-451315 on Build GRCh38
rs767136429 Chr.8: 451343-451343 on Build GRCh38
rs760463462 Chr.8: 451248-451253 on Build GRCh38
rs568931568 Chr.8: 451267-451267 on Build GRCh38
rs777180855 Chr.8: 451266-451266 on Build GRCh38
rs776934257 Chr.8: 451385-451385 on Build GRCh38
rs769666062 Chr.8: 451393-451393 on Build GRCh38
rs147288048 Chr.8: 451304-451304 on Build GRCh38
rs140368463 Chr.8: 451433-451433 on Build GRCh38
rs537982331 Chr.8: 451279-451279 on Build GRCh38
rs558370110 Chr.8: 451294-451294 on Build GRCh38
rs767062092 Chr.8: 451405-451405 on Build GRCh38
rs746091563 Chr.8: 451387-451387 on Build GRCh38
rs768450042 Chr.8: 451308-451308 on Build GRCh38
rs1023512139 Chr.8: 451354-451354 on Build GRCh38
rs756909108 Chr.8: 451283-451283 on Build GRCh38
rs773183517 Chr.8: 451307-451307 on Build GRCh38
rs749722619 Chr.8: 451287-451287 on Build GRCh38
rs779337773 Chr.8: 451293-451293 on Build GRCh38
rs139896334 Chr.8: 451314-451314 on Build GRCh38
rs78003365 Chr.8: 451274-451274 on Build GRCh38
rs763007601 Chr.8: 451326-451326 on Build GRCh38
rs764009229 Chr.8: 451399-451399 on Build GRCh38
rs762952735 Chr.8: 451395-451395 on Build GRCh38
rs750092416 Chr.8: 451344-451344 on Build GRCh38

View Full Product Details