Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3586639-3586892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC3
Entrez Gene ID
126326
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
C19orf64, DFNB15, DFNB72, DFNB95
UniGene
Hs.266873
Gene Chromosome Location
Chr.19: 3585531-3593541 on Build GRCh38
Transcripts
NM_133261.2 XM_005259492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1392431 c.472G>A p.E158K Chr.19: 3586874-3586874 on Build GRCh38
4686978 c.484G>A p.D162N Chr.19: 3586886-3586886 on Build GRCh38
4413286 c.435C>G p.I145M Chr.19: 3586837-3586837 on Build GRCh38
7098555 c.421G>A p.E141K Chr.19: 3586823-3586823 on Build GRCh38
6224643 c.387C>T p.N129N Chr.19: 3586656-3586656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749877432 Chr.19: 3586796-3586796 on Build GRCh38
rs941263972 Chr.19: 3586759-3586759 on Build GRCh38
rs777929510 Chr.19: 3586713-3586713 on Build GRCh38
rs780839716 Chr.19: 3586824-3586824 on Build GRCh38
rs371678561 Chr.19: 3586679-3586679 on Build GRCh38
rs887498875 Chr.19: 3586783-3586783 on Build GRCh38
rs764731510 Chr.19: 3586795-3586795 on Build GRCh38
rs147646886 Chr.19: 3586735-3586736 on Build GRCh38
rs774607609 Chr.19: 3586730-3586730 on Build GRCh38
rs876657692 Chr.19: 3586681-3586681 on Build GRCh38
rs751423501 Chr.19: 3586811-3586811 on Build GRCh38
rs776322867 Chr.19: 3586774-3586774 on Build GRCh38
rs929822321 Chr.19: 3586734-3586734 on Build GRCh38
rs766301789 Chr.19: 3586803-3586803 on Build GRCh38
rs749099704 Chr.19: 3586783-3586783 on Build GRCh38
rs553397537 Chr.19: 3586753-3586753 on Build GRCh38
rs947095066 Chr.19: 3586760-3586760 on Build GRCh38
rs755770901 Chr.19: 3586728-3586728 on Build GRCh38
rs748361788 Chr.19: 3586701-3586701 on Build GRCh38
rs958425987 Chr.19: 3586714-3586714 on Build GRCh38
rs983071681 Chr.19: 3586716-3586716 on Build GRCh38
rs370334898 Chr.19: 3586857-3586857 on Build GRCh38
rs778018411 Chr.19: 3586835-3586835 on Build GRCh38
rs1010472144 Chr.19: 3586668-3586668 on Build GRCh38
rs370952334 Chr.19: 3586735-3586735 on Build GRCh38
rs761468917 Chr.19: 3586863-3586863 on Build GRCh38
rs909821701 Chr.19: 3586758-3586758 on Build GRCh38
rs749823033 Chr.19: 3586715-3586715 on Build GRCh38
rs746771891 Chr.19: 3586691-3586691 on Build GRCh38
rs756856501 Chr.19: 3586695-3586695 on Build GRCh38
rs779520578 Chr.19: 3586847-3586847 on Build GRCh38
rs772300090 Chr.19: 3586731-3586731 on Build GRCh38
rs761450311 Chr.19: 3586776-3586776 on Build GRCh38
rs772555181 Chr.19: 3586860-3586860 on Build GRCh38
rs775765891 Chr.19: 3586862-3586862 on Build GRCh38
rs779727957 Chr.19: 3586690-3586690 on Build GRCh38
rs758266699 Chr.19: 3586689-3586689 on Build GRCh38
rs754854408 Chr.19: 3586823-3586823 on Build GRCh38
rs750427587 Chr.19: 3586687-3586687 on Build GRCh38
rs749337255 Chr.19: 3586841-3586841 on Build GRCh38
rs753038931 Chr.19: 3586827-3586827 on Build GRCh38
rs531054314 Chr.19: 3586727-3586728 on Build GRCh38
rs760264727 Chr.19: 3586664-3586664 on Build GRCh38
rs536816169 Chr.19: 3586684-3586684 on Build GRCh38
rs557145939 Chr.19: 3586705-3586705 on Build GRCh38
rs781520349 Chr.19: 3586700-3586700 on Build GRCh38
rs141293401 Chr.19: 3586842-3586842 on Build GRCh38
rs756342494 Chr.19: 3586833-3586833 on Build GRCh38
rs764713230 Chr.19: 3586686-3586686 on Build GRCh38
rs756809309 Chr.19: 3586685-3586685 on Build GRCh38
rs567549706 Chr.19: 3586727-3586727 on Build GRCh38
rs755189458 Chr.19: 3586692-3586692 on Build GRCh38
rs769390047 Chr.19: 3586865-3586865 on Build GRCh38
rs762901930 Chr.19: 3586798-3586798 on Build GRCh38
rs900520717 Chr.19: 3586828-3586828 on Build GRCh38

View Full Product Details