Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6977345-6977616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NR_135131.1 NM_005768.5

Gene Symbol
LPCAT3
Entrez Gene ID
10162
Gene Name
lysophosphatidylcholine acyltransferase 3
Gene Aliases
C3F, LPCAT, LPLAT 5, LPSAT, MBOAT5, OACT5, nessy
UniGene
Hs.655248
Gene Chromosome Location
Chr.12: 6976185-7018538 on Build GRCh38
Transcripts
NR_135131.1 NM_005768.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234923 c.1179G>A p.V393V Chr.12: 6977607-6977607 on Build GRCh38
3396089 c.1282C>T p.H428Y Chr.12: 6977432-6977432 on Build GRCh38
160729 c.1257C>G p.F419L Chr.12: 6977457-6977457 on Build GRCh38
5492916 c.1193C>T p.A398V Chr.12: 6977521-6977521 on Build GRCh38
4044591 c.1307T>C p.M436T Chr.12: 6977407-6977407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782536773 Chr.12: 6977400-6977400 on Build GRCh38
rs782605348 Chr.12: 6977442-6977442 on Build GRCh38
rs782767044 Chr.12: 6977379-6977379 on Build GRCh38
rs782562293 Chr.12: 6977578-6977578 on Build GRCh38
rs782195366 Chr.12: 6977463-6977463 on Build GRCh38
rs771879699 Chr.12: 6977558-6977558 on Build GRCh38
rs781898267 Chr.12: 6977553-6977553 on Build GRCh38
rs782434418 Chr.12: 6977476-6977476 on Build GRCh38
rs782459858 Chr.12: 6977494-6977494 on Build GRCh38
rs782796529 Chr.12: 6977511-6977511 on Build GRCh38
rs782383008 Chr.12: 6977487-6977487 on Build GRCh38
rs782462102 Chr.12: 6977533-6977533 on Build GRCh38
rs782597754 Chr.12: 6977585-6977585 on Build GRCh38
rs781852721 Chr.12: 6977436-6977436 on Build GRCh38
rs782468847 Chr.12: 6977579-6977579 on Build GRCh38
rs1035421832 Chr.12: 6977497-6977497 on Build GRCh38
rs1002601281 Chr.12: 6977449-6977449 on Build GRCh38
rs782642541 Chr.12: 6977564-6977564 on Build GRCh38
rs782178630 Chr.12: 6977586-6977586 on Build GRCh38
rs782685832 Chr.12: 6977540-6977540 on Build GRCh38
rs1042565726 Chr.12: 6977420-6977420 on Build GRCh38
rs142552475 Chr.12: 6977367-6977367 on Build GRCh38
rs111742644 Chr.12: 6977369-6977369 on Build GRCh38
rs781816329 Chr.12: 6977514-6977514 on Build GRCh38
rs781973336 Chr.12: 6977490-6977490 on Build GRCh38
rs782116568 Chr.12: 6977520-6977520 on Build GRCh38
rs200123453 Chr.12: 6977562-6977562 on Build GRCh38
rs369868584 Chr.12: 6977481-6977481 on Build GRCh38
rs782167922 Chr.12: 6977502-6977502 on Build GRCh38
rs781908140 Chr.12: 6977388-6977388 on Build GRCh38
rs782549253 Chr.12: 6977569-6977569 on Build GRCh38
rs782239619 Chr.12: 6977578-6977578 on Build GRCh38
rs782702564 Chr.12: 6977386-6977386 on Build GRCh38
rs141446248 Chr.12: 6977499-6977499 on Build GRCh38
rs782477008 Chr.12: 6977438-6977438 on Build GRCh38
rs117996263 Chr.12: 6977424-6977424 on Build GRCh38
rs782041747 Chr.12: 6977480-6977480 on Build GRCh38

View Full Product Details