Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62629559-62629816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GANAB
Entrez Gene ID
23193
Gene Name
glucosidase II alpha subunit
Gene Aliases
G2AN, GIIA, GLUII
UniGene
Hs.595071
Gene Chromosome Location
Chr.11: 62624826-62646726 on Build GRCh38
Transcripts
NM_001278192.1 NM_001278193.1 NM_001278194.1 NM_198334.2 NM_198335.3 XM_017017412.1 XM_017017413.1 XM_017017414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6133712 c.1823C>T p.S608F Chr.11: 62629599-62629599 on Build GRCh38
6262845 c.1856C>A p.P619H Chr.11: 62629632-62629632 on Build GRCh38
1241200 c.1786C>T p.R596C Chr.11: 62629636-62629636 on Build GRCh38
4810636 c.1857C>G p.P619P Chr.11: 62629631-62629631 on Build GRCh38
1298316 c.1791C>G p.P597P Chr.11: 62629631-62629631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768271485 Chr.11: 62629676-62629676 on Build GRCh38
rs981906447 Chr.11: 62629788-62629788 on Build GRCh38
rs779582905 Chr.11: 62629793-62629793 on Build GRCh38
rs879142409 Chr.11: 62629661-62629661 on Build GRCh38
rs778158520 Chr.11: 62629732-62629732 on Build GRCh38
rs948683 Chr.11: 62629705-62629705 on Build GRCh38
rs749010632 Chr.11: 62629656-62629656 on Build GRCh38
rs755292031 Chr.11: 62629724-62629724 on Build GRCh38
rs751584036 Chr.11: 62629797-62629798 on Build GRCh38
rs761550860 Chr.11: 62629694-62629694 on Build GRCh38
rs779486964 Chr.11: 62629610-62629610 on Build GRCh38
rs758876878 Chr.11: 62629622-62629622 on Build GRCh38
rs769290554 Chr.11: 62629649-62629649 on Build GRCh38
rs761623163 Chr.11: 62629728-62629735 on Build GRCh38
rs915137149 Chr.11: 62629760-62629760 on Build GRCh38
rs370381689 Chr.11: 62629730-62629730 on Build GRCh38
rs765532287 Chr.11: 62629725-62629725 on Build GRCh38
rs767383584 Chr.11: 62629704-62629704 on Build GRCh38
rs569986944 Chr.11: 62629746-62629746 on Build GRCh38
rs766534942 Chr.11: 62629708-62629708 on Build GRCh38
rs994145126 Chr.11: 62629643-62629643 on Build GRCh38
rs754052468 Chr.11: 62629598-62629598 on Build GRCh38
rs201172482 Chr.11: 62629707-62629707 on Build GRCh38
rs774183839 Chr.11: 62629687-62629687 on Build GRCh38
rs369432448 Chr.11: 62629780-62629780 on Build GRCh38
rs753962181 Chr.11: 62629714-62629714 on Build GRCh38
rs1023507991 Chr.11: 62629747-62629747 on Build GRCh38
rs1018397643 Chr.11: 62629712-62629712 on Build GRCh38
rs760948316 Chr.11: 62629731-62629731 on Build GRCh38
rs774850686 Chr.11: 62629655-62629655 on Build GRCh38
rs76572368 Chr.11: 62629642-62629642 on Build GRCh38
rs772342694 Chr.11: 62629693-62629693 on Build GRCh38
rs778304210 Chr.11: 62629634-62629634 on Build GRCh38
rs369559736 Chr.11: 62629591-62629591 on Build GRCh38
rs764362724 Chr.11: 62629606-62629606 on Build GRCh38
rs879026958 Chr.11: 62629669-62629669 on Build GRCh38
rs77092080 Chr.11: 62629607-62629607 on Build GRCh38
rs748490428 Chr.11: 62629621-62629621 on Build GRCh38
rs373694305 Chr.11: 62629767-62629767 on Build GRCh38

View Full Product Details