Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81354324-81354549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
UniGene
Hs.112569
Gene Chromosome Location
Chr.16: 81314941-81380198 on Build GRCh38
Transcripts
NM_022041.3 XM_017023734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6197023 c.357C>A p.T119T Chr.16: 81354479-81354479 on Build GRCh38
6451959 c.344C>T p.T115M Chr.16: 81354466-81354466 on Build GRCh38
3957865 c.427C>T p.H143Y Chr.16: 81354549-81354549 on Build GRCh38
5039718 c.407G>A p.G136D Chr.16: 81354529-81354529 on Build GRCh38
1380074 c.334C>T p.L112L Chr.16: 81354456-81354456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771287317 Chr.16: 81354355-81354355 on Build GRCh38
rs775270372 Chr.16: 81354368-81354368 on Build GRCh38
rs771660619 Chr.16: 81354444-81354444 on Build GRCh38
rs769725195 Chr.16: 81354367-81354367 on Build GRCh38
rs765503261 Chr.16: 81354453-81354453 on Build GRCh38
rs754860983 Chr.16: 81354513-81354513 on Build GRCh38
rs372022466 Chr.16: 81354470-81354470 on Build GRCh38
rs771476213 Chr.16: 81354527-81354527 on Build GRCh38
rs200045368 Chr.16: 81354452-81354452 on Build GRCh38
rs371984923 Chr.16: 81354385-81354385 on Build GRCh38
rs377019851 Chr.16: 81354382-81354382 on Build GRCh38
rs751690125 Chr.16: 81354407-81354407 on Build GRCh38
rs117009369 Chr.16: 81354365-81354365 on Build GRCh38
rs750608407 Chr.16: 81354478-81354478 on Build GRCh38
rs917427848 Chr.16: 81354371-81354371 on Build GRCh38
rs769446654 Chr.16: 81354424-81354424 on Build GRCh38
rs753122436 Chr.16: 81354455-81354455 on Build GRCh38
rs759028377 Chr.16: 81354395-81354395 on Build GRCh38
rs568951825 Chr.16: 81354449-81354449 on Build GRCh38
rs768322742 Chr.16: 81354467-81354467 on Build GRCh38
rs978580248 Chr.16: 81354364-81354364 on Build GRCh38
rs746154353 Chr.16: 81354417-81354417 on Build GRCh38
rs779968294 Chr.16: 81354426-81354426 on Build GRCh38
rs748121633 Chr.16: 81354525-81354525 on Build GRCh38
rs773910375 Chr.16: 81354374-81354374 on Build GRCh38
rs768151267 Chr.16: 81354373-81354373 on Build GRCh38
rs994187670 Chr.16: 81354492-81354492 on Build GRCh38
rs779767233 Chr.16: 81354485-81354485 on Build GRCh38
rs753249311 Chr.16: 81354394-81354394 on Build GRCh38
rs935670717 Chr.16: 81354435-81354435 on Build GRCh38
rs780979004 Chr.16: 81354360-81354360 on Build GRCh38
rs551360416 Chr.16: 81354465-81354465 on Build GRCh38
rs762300322 Chr.16: 81354481-81354481 on Build GRCh38
rs141396595 Chr.16: 81354477-81354477 on Build GRCh38
rs754182168 Chr.16: 81354390-81354390 on Build GRCh38
rs762920457 Chr.16: 81354370-81354370 on Build GRCh38
rs765672686 Chr.16: 81354393-81354393 on Build GRCh38
rs145789614 Chr.16: 81354521-81354521 on Build GRCh38
rs369700456 Chr.16: 81354391-81354391 on Build GRCh38
rs113655220 Chr.16: 81354480-81354480 on Build GRCh38
rs9936546 Chr.16: 81354353-81354353 on Build GRCh38
rs761334418 Chr.16: 81354436-81354436 on Build GRCh38
rs749087082 Chr.16: 81354501-81354501 on Build GRCh38
rs749211847 Chr.16: 81354428-81354428 on Build GRCh38
rs781296328 Chr.16: 81354410-81354410 on Build GRCh38
rs773783945 Chr.16: 81354433-81354433 on Build GRCh38

View Full Product Details