Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92482681-92482912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1
Entrez Gene ID
2672
Gene Name
growth factor independent 1 transcriptional repressor
Gene Aliases
GFI-1, GFI1A, SCN2, ZNF163
UniGene
Hs.73172
Gene Chromosome Location
Chr.1: 92473043-92486876 on Build GRCh38
Transcripts
NM_001127215.1 NM_001127216.1 NM_005263.3 XM_005270749.3 XM_011541245.2 XM_011541246.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670500 c.288C>T p.S96S Chr.1: 92482874-92482874 on Build GRCh38
7263889 c.279G>A p.P93P Chr.1: 92482883-92482883 on Build GRCh38
6368660 c.272G>T p.R91M Chr.1: 92482890-92482890 on Build GRCh38
4485746 c.296C>T p.P99L Chr.1: 92482866-92482866 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs996280859 Chr.1: 92482710-92482710 on Build GRCh38
rs559490487 Chr.1: 92482751-92482751 on Build GRCh38
rs756930440 Chr.1: 92482856-92482856 on Build GRCh38
rs995056593 Chr.1: 92482793-92482793 on Build GRCh38
rs199514537 Chr.1: 92482818-92482818 on Build GRCh38
rs992126786 Chr.1: 92482881-92482881 on Build GRCh38
rs74101165 Chr.1: 92482827-92482827 on Build GRCh38
rs772457244 Chr.1: 92482819-92482819 on Build GRCh38
rs572617941 Chr.1: 92482771-92482771 on Build GRCh38
rs771263599 Chr.1: 92482823-92482823 on Build GRCh38
rs747215489 Chr.1: 92482802-92482802 on Build GRCh38
rs780103307 Chr.1: 92482884-92482884 on Build GRCh38
rs1052161669 Chr.1: 92482711-92482711 on Build GRCh38
rs752234811 Chr.1: 92482839-92482839 on Build GRCh38
rs778939406 Chr.1: 92482886-92482886 on Build GRCh38
rs760046831 Chr.1: 92482836-92482836 on Build GRCh38
rs542422281 Chr.1: 92482808-92482808 on Build GRCh38
rs191770152 Chr.1: 92482815-92482815 on Build GRCh38
rs762399601 Chr.1: 92482844-92482844 on Build GRCh38
rs763637540 Chr.1: 92482852-92482852 on Build GRCh38
rs144945261 Chr.1: 92482845-92482845 on Build GRCh38
rs751210834 Chr.1: 92482855-92482855 on Build GRCh38
rs1047279810 Chr.1: 92482706-92482706 on Build GRCh38
rs890457074 Chr.1: 92482747-92482747 on Build GRCh38
rs868315713 Chr.1: 92482809-92482809 on Build GRCh38
rs560847313 Chr.1: 92482813-92482813 on Build GRCh38
rs773262174 Chr.1: 92482820-92482820 on Build GRCh38
rs1018695566 Chr.1: 92482737-92482737 on Build GRCh38
rs779533629 Chr.1: 92482763-92482763 on Build GRCh38
rs963289404 Chr.1: 92482783-92482783 on Build GRCh38
rs780907351 Chr.1: 92482857-92482857 on Build GRCh38
rs553996787 Chr.1: 92482716-92482716 on Build GRCh38
rs760933177 Chr.1: 92482822-92482822 on Build GRCh38
rs1021748699 Chr.1: 92482803-92482803 on Build GRCh38
rs1008194643 Chr.1: 92482736-92482736 on Build GRCh38
rs750187013 Chr.1: 92482862-92482862 on Build GRCh38
rs778381648 Chr.1: 92482807-92482807 on Build GRCh38
rs765663437 Chr.1: 92482838-92482838 on Build GRCh38
rs771344042 Chr.1: 92482873-92482873 on Build GRCh38
rs756127881 Chr.1: 92482880-92482880 on Build GRCh38
rs572170195 Chr.1: 92482767-92482767 on Build GRCh38

View Full Product Details