Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6553147-6553404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109540 c.2440T>C p.S814P Chr.9: 6553385-6553385 on Build GRCh38
6395444 c.2446G>T p.A816S Chr.9: 6553379-6553379 on Build GRCh38
3703573 c.2439T>G p.I813M Chr.9: 6553386-6553386 on Build GRCh38
2777831 c.2441C>A p.S814Y Chr.9: 6553384-6553384 on Build GRCh38
1700816 c.2426C>T p.S809F Chr.9: 6553399-6553399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760408603 Chr.9: 6553363-6553363 on Build GRCh38
rs993822316 Chr.9: 6553368-6553368 on Build GRCh38
rs911050625 Chr.9: 6553211-6553211 on Build GRCh38
rs768260628 Chr.9: 6553344-6553344 on Build GRCh38
rs746178235 Chr.9: 6553325-6553325 on Build GRCh38
rs775834004 Chr.9: 6553379-6553379 on Build GRCh38
rs986097214 Chr.9: 6553236-6553236 on Build GRCh38
rs774761384 Chr.9: 6553313-6553313 on Build GRCh38
rs371788223 Chr.9: 6553351-6553351 on Build GRCh38
rs924215580 Chr.9: 6553296-6553296 on Build GRCh38
rs934485623 Chr.9: 6553304-6553304 on Build GRCh38
rs946711526 Chr.9: 6553284-6553284 on Build GRCh38
rs915229392 Chr.9: 6553270-6553270 on Build GRCh38
rs1038098554 Chr.9: 6553335-6553335 on Build GRCh38
rs147308839 Chr.9: 6553360-6553360 on Build GRCh38
rs914288460 Chr.9: 6553314-6553314 on Build GRCh38
rs897976834 Chr.9: 6553203-6553203 on Build GRCh38
rs924491124 Chr.9: 6553300-6553300 on Build GRCh38
rs769440435 Chr.9: 6553221-6553221 on Build GRCh38
rs952062303 Chr.9: 6553223-6553223 on Build GRCh38
rs757838033 Chr.9: 6553321-6553321 on Build GRCh38
rs560943302 Chr.9: 6553195-6553195 on Build GRCh38
rs538324686 Chr.9: 6553275-6553275 on Build GRCh38
rs368399158 Chr.9: 6553324-6553324 on Build GRCh38
rs200347920 Chr.9: 6553319-6553319 on Build GRCh38
rs754241290 Chr.9: 6553347-6553348 on Build GRCh38
rs766061438 Chr.9: 6553374-6553374 on Build GRCh38
rs974818574 Chr.9: 6553254-6553254 on Build GRCh38
rs111262564 Chr.9: 6553262-6553262 on Build GRCh38
rs1056681781 Chr.9: 6553316-6553316 on Build GRCh38
rs1042400387 Chr.9: 6553286-6553286 on Build GRCh38
rs548030253 Chr.9: 6553244-6553244 on Build GRCh38
rs1035163693 Chr.9: 6553202-6553202 on Build GRCh38
rs755854739 Chr.9: 6553327-6553327 on Build GRCh38
rs965189295 Chr.9: 6553213-6553213 on Build GRCh38
rs528124512 Chr.9: 6553209-6553209 on Build GRCh38
rs749155472 Chr.9: 6553260-6553260 on Build GRCh38
rs1012412673 Chr.9: 6553334-6553334 on Build GRCh38
rs939583389 Chr.9: 6553310-6553310 on Build GRCh38
rs771927272 Chr.9: 6553358-6553358 on Build GRCh38
rs183082572 Chr.9: 6553255-6553255 on Build GRCh38
rs546180913 Chr.9: 6553232-6553232 on Build GRCh38
rs779717203 Chr.9: 6553340-6553340 on Build GRCh38
rs768569870 Chr.9: 6553305-6553305 on Build GRCh38

View Full Product Details