Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27374641-27374906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCN3
Entrez Gene ID
8547
Gene Name
ficolin 3
Gene Aliases
FCNH, HAKA1
UniGene
Hs.333383
Gene Chromosome Location
Chr.1: 27369110-27374824 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5750969 c.79C>A p.P27T Chr.1: 27374740-27374740 on Build GRCh38
5380549 c.3G>A p.M1I Chr.1: 27374816-27374816 on Build GRCh38
1602173 c.4G>A p.D2N Chr.1: 27374815-27374815 on Build GRCh38
6321931 c.80C>A p.P27H Chr.1: 27374739-27374739 on Build GRCh38
3487692 c.40C>T p.L14L Chr.1: 27374779-27374779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950099607 Chr.1: 27374750-27374750 on Build GRCh38
rs763786651 Chr.1: 27374862-27374862 on Build GRCh38
rs189227069 Chr.1: 27374694-27374694 on Build GRCh38
rs994388861 Chr.1: 27374667-27374667 on Build GRCh38
rs759497509 Chr.1: 27374835-27374835 on Build GRCh38
rs566891028 Chr.1: 27374675-27374675 on Build GRCh38
rs749956317 Chr.1: 27374689-27374691 on Build GRCh38
rs753381366 Chr.1: 27374748-27374748 on Build GRCh38
rs775288128 Chr.1: 27374841-27374841 on Build GRCh38
rs962181216 Chr.1: 27374666-27374666 on Build GRCh38
rs372596836 Chr.1: 27374825-27374825 on Build GRCh38
rs781329306 Chr.1: 27374820-27374820 on Build GRCh38
rs765301683 Chr.1: 27374767-27374768 on Build GRCh38
rs767380723 Chr.1: 27374720-27374720 on Build GRCh38
rs780342236 Chr.1: 27374808-27374808 on Build GRCh38
rs750964366 Chr.1: 27374792-27374792 on Build GRCh38
rs911139338 Chr.1: 27374687-27374687 on Build GRCh38
rs56088921 Chr.1: 27374768-27374768 on Build GRCh38
rs41366544 Chr.1: 27374785-27374785 on Build GRCh38
rs774481237 Chr.1: 27374710-27374710 on Build GRCh38
rs756772261 Chr.1: 27374766-27374766 on Build GRCh38
rs752456913 Chr.1: 27374736-27374736 on Build GRCh38
rs763657253 Chr.1: 27374747-27374747 on Build GRCh38
rs760316888 Chr.1: 27374743-27374743 on Build GRCh38
rs779366769 Chr.1: 27374782-27374782 on Build GRCh38
rs761923448 Chr.1: 27374756-27374759 on Build GRCh38
rs748239361 Chr.1: 27374821-27374821 on Build GRCh38
rs768920855 Chr.1: 27374819-27374819 on Build GRCh38
rs768658968 Chr.1: 27374741-27374742 on Build GRCh38
rs915380290 Chr.1: 27374860-27374860 on Build GRCh38
rs761868698 Chr.1: 27374705-27374705 on Build GRCh38
rs200720652 Chr.1: 27374827-27374827 on Build GRCh38
rs760452725 Chr.1: 27374843-27374843 on Build GRCh38
rs968060659 Chr.1: 27374665-27374666 on Build GRCh38
rs965391401 Chr.1: 27374709-27374709 on Build GRCh38
rs758930710 Chr.1: 27374800-27374800 on Build GRCh38
rs976456334 Chr.1: 27374793-27374793 on Build GRCh38
rs546584321 Chr.1: 27374674-27374674 on Build GRCh38
rs201979374 Chr.1: 27374681-27374681 on Build GRCh38
rs770980672 Chr.1: 27374707-27374707 on Build GRCh38
rs775655432 Chr.1: 27374680-27374680 on Build GRCh38
rs776483001 Chr.1: 27374746-27374746 on Build GRCh38
rs759374713 Chr.1: 27374714-27374714 on Build GRCh38
rs369159057 Chr.1: 27374685-27374685 on Build GRCh38
rs773555389 Chr.1: 27374768-27374768 on Build GRCh38
rs537358744 Chr.1: 27374837-27374837 on Build GRCh38
rs147142056 Chr.1: 27374810-27374810 on Build GRCh38
rs776709383 Chr.1: 27374703-27374703 on Build GRCh38

View Full Product Details