Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162143239-162143510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCG
Entrez Gene ID
2641
Gene Name
glucagon
Gene Aliases
GLP1, GLP2, GRPP
UniGene
Hs.516494
Gene Chromosome Location
Chr.2: 162142869-162152404 on Build GRCh38
Transcripts
NM_002054.4 NR_110255.1

Gene Symbol
LOC101929532
Entrez Gene ID
101929532
Gene Name
uncharacterized LOC101929532
Gene Aliases
-
UniGene
Hs.639883
Gene Chromosome Location
Chr.2: 162114441-162172739 on Build GRCh38
Transcripts
NM_002054.4 NR_110255.1

SNP Detail

SNP ID Chromosome Location
rs929025979 Chr.2: 162143345-162143345 on Build GRCh38
rs917696060 Chr.2: 162143312-162143312 on Build GRCh38
rs959311376 Chr.2: 162143277-162143277 on Build GRCh38
rs760071818 Chr.2: 162143320-162143320 on Build GRCh38
rs765297547 Chr.2: 162143417-162143417 on Build GRCh38
rs983317050 Chr.2: 162143361-162143361 on Build GRCh38
rs376296040 Chr.2: 162143421-162143421 on Build GRCh38
rs957073338 Chr.2: 162143459-162143459 on Build GRCh38
rs963492250 Chr.2: 162143308-162143308 on Build GRCh38
rs941801169 Chr.2: 162143441-162143441 on Build GRCh38
rs1039368564 Chr.2: 162143467-162143467 on Build GRCh38
rs1004074246 Chr.2: 162143275-162143277 on Build GRCh38
rs1015016803 Chr.2: 162143275-162143275 on Build GRCh38
rs988100713 Chr.2: 162143418-162143418 on Build GRCh38
rs991816474 Chr.2: 162143306-162143306 on Build GRCh38
rs909043060 Chr.2: 162143394-162143394 on Build GRCh38
rs771192295 Chr.2: 162143352-162143352 on Build GRCh38
rs988307717 Chr.2: 162143461-162143461 on Build GRCh38
rs1028959139 Chr.2: 162143317-162143317 on Build GRCh38
rs777387928 Chr.2: 162143386-162143387 on Build GRCh38

View Full Product Details