Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 890947-891180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GET4
Entrez Gene ID
51608
Gene Name
golgi to ER traffic protein 4
Gene Aliases
C7orf20, CEE, CGI-20, TRC35
UniGene
Hs.107387
Gene Chromosome Location
Chr.7: 876554-896434 on Build GRCh38
Transcripts
NM_015949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1208107 c.557G>A p.R186H Chr.7: 891018-891018 on Build GRCh38
6863073 c.560G>C p.G187A Chr.7: 891021-891021 on Build GRCh38
5062673 c.510G>A p.A170A Chr.7: 890971-890971 on Build GRCh38
1093223 c.566G>A p.R189H Chr.7: 891027-891027 on Build GRCh38
6047642 c.522C>T p.G174G Chr.7: 890983-890983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752506322 Chr.7: 891087-891087 on Build GRCh38
rs370621678 Chr.7: 891001-891001 on Build GRCh38
rs146762326 Chr.7: 890974-890974 on Build GRCh38
rs374400171 Chr.7: 891100-891100 on Build GRCh38
rs769687204 Chr.7: 891079-891079 on Build GRCh38
rs755282296 Chr.7: 891114-891114 on Build GRCh38
rs753980165 Chr.7: 891109-891109 on Build GRCh38
rs372478502 Chr.7: 891097-891097 on Build GRCh38
rs906481077 Chr.7: 891157-891157 on Build GRCh38
rs753505111 Chr.7: 891042-891042 on Build GRCh38
rs370235298 Chr.7: 891070-891070 on Build GRCh38
rs546623650 Chr.7: 891145-891145 on Build GRCh38
rs999809216 Chr.7: 891058-891058 on Build GRCh38
rs374735615 Chr.7: 891031-891031 on Build GRCh38
rs756310603 Chr.7: 890976-890976 on Build GRCh38
rs536246178 Chr.7: 891151-891151 on Build GRCh38
rs915033412 Chr.7: 891137-891137 on Build GRCh38
rs1017484399 Chr.7: 891038-891038 on Build GRCh38
rs1054555030 Chr.7: 891130-891130 on Build GRCh38
rs750918390 Chr.7: 891016-891016 on Build GRCh38
rs780137609 Chr.7: 891036-891036 on Build GRCh38
rs780120751 Chr.7: 890983-890983 on Build GRCh38
rs775277936 Chr.7: 891017-891017 on Build GRCh38
rs11764234 Chr.7: 891143-891143 on Build GRCh38
rs770819090 Chr.7: 891012-891012 on Build GRCh38
rs377188160 Chr.7: 890999-890999 on Build GRCh38
rs200742822 Chr.7: 891015-891015 on Build GRCh38
rs150276017 Chr.7: 891013-891013 on Build GRCh38
rs947924434 Chr.7: 891138-891138 on Build GRCh38
rs980146762 Chr.7: 891132-891132 on Build GRCh38
rs1025327434 Chr.7: 891020-891020 on Build GRCh38
rs866868274 Chr.7: 891053-891053 on Build GRCh38
rs779087667 Chr.7: 891117-891117 on Build GRCh38
rs1031339632 Chr.7: 891075-891075 on Build GRCh38
rs376175295 Chr.7: 891093-891093 on Build GRCh38
rs774983520 Chr.7: 891081-891081 on Build GRCh38
rs767817290 Chr.7: 891029-891029 on Build GRCh38
rs756289903 Chr.7: 891035-891035 on Build GRCh38
rs143992969 Chr.7: 891027-891027 on Build GRCh38
rs749335987 Chr.7: 890991-890991 on Build GRCh38
rs762246645 Chr.7: 891095-891095 on Build GRCh38
rs752030449 Chr.7: 891026-891026 on Build GRCh38
rs368471289 Chr.7: 891101-891101 on Build GRCh38
rs762355270 Chr.7: 891018-891018 on Build GRCh38
rs201473801 Chr.7: 891050-891050 on Build GRCh38
rs762147381 Chr.7: 891076-891077 on Build GRCh38
rs556198233 Chr.7: 891152-891152 on Build GRCh38
rs113470707 Chr.7: 891088-891088 on Build GRCh38
rs955965593 Chr.7: 891086-891086 on Build GRCh38
rs920686579 Chr.7: 891136-891136 on Build GRCh38
rs779177640 Chr.7: 891047-891047 on Build GRCh38
rs13171 Chr.7: 891052-891052 on Build GRCh38
rs768849334 Chr.7: 890998-890998 on Build GRCh38
rs754149897 Chr.7: 891046-891046 on Build GRCh38
rs139373772 Chr.7: 891019-891019 on Build GRCh38
rs368451546 Chr.7: 891094-891094 on Build GRCh38
rs936119403 Chr.7: 891113-891113 on Build GRCh38
rs764578617 Chr.7: 890970-890970 on Build GRCh38
rs11543223 Chr.7: 890971-890971 on Build GRCh38
rs766217461 Chr.7: 891112-891116 on Build GRCh38

View Full Product Details