Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120130569-120130842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCN1
Entrez Gene ID
10985
Gene Name
GCN1, eIF2 alpha kinase activator homolog
Gene Aliases
GCN1L, GCN1L1, PRIC295
UniGene
Hs.298716
Gene Chromosome Location
Chr.12: 120127210-120194710 on Build GRCh38
Transcripts
NM_006836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
935852 c.7589G>A p.R2530Q Chr.12: 120130728-120130728 on Build GRCh38
244679 c.7574C>T p.A2525V Chr.12: 120130743-120130743 on Build GRCh38
6135597 c.7659C>A p.S2553R Chr.12: 120130658-120130658 on Build GRCh38
3416547 c.7621G>A p.E2541K Chr.12: 120130696-120130696 on Build GRCh38
244677 c.7588C>T p.R2530W Chr.12: 120130729-120130729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757807251 Chr.12: 120130729-120130729 on Build GRCh38
rs76817269 Chr.12: 120130622-120130622 on Build GRCh38
rs765195519 Chr.12: 120130783-120130783 on Build GRCh38
rs145712872 Chr.12: 120130728-120130728 on Build GRCh38
rs369470290 Chr.12: 120130664-120130664 on Build GRCh38
rs765229532 Chr.12: 120130687-120130687 on Build GRCh38
rs529294048 Chr.12: 120130634-120130635 on Build GRCh38
rs1003447506 Chr.12: 120130764-120130764 on Build GRCh38
rs202191794 Chr.12: 120130663-120130663 on Build GRCh38
rs749753713 Chr.12: 120130633-120130633 on Build GRCh38
rs369317247 Chr.12: 120130727-120130727 on Build GRCh38
rs766499232 Chr.12: 120130668-120130668 on Build GRCh38
rs113960572 Chr.12: 120130645-120130645 on Build GRCh38
rs530651875 Chr.12: 120130628-120130628 on Build GRCh38
rs779611551 Chr.12: 120130733-120130733 on Build GRCh38
rs377763033 Chr.12: 120130779-120130779 on Build GRCh38
rs555079874 Chr.12: 120130749-120130749 on Build GRCh38
rs200903918 Chr.12: 120130651-120130651 on Build GRCh38
rs878967002 Chr.12: 120130595-120130595 on Build GRCh38
rs766116567 Chr.12: 120130697-120130697 on Build GRCh38
rs750157042 Chr.12: 120130688-120130688 on Build GRCh38
rs570700576 Chr.12: 120130662-120130662 on Build GRCh38
rs537293941 Chr.12: 120130790-120130790 on Build GRCh38
rs1013408512 Chr.12: 120130736-120130736 on Build GRCh38
rs768928169 Chr.12: 120130751-120130751 on Build GRCh38
rs375528439 Chr.12: 120130652-120130652 on Build GRCh38
rs762800418 Chr.12: 120130778-120130778 on Build GRCh38
rs757134437 Chr.12: 120130741-120130741 on Build GRCh38
rs576665805 Chr.12: 120130673-120130673 on Build GRCh38
rs372927556 Chr.12: 120130742-120130742 on Build GRCh38
rs373248631 Chr.12: 120130674-120130674 on Build GRCh38
rs780287777 Chr.12: 120130748-120130748 on Build GRCh38
rs370299043 Chr.12: 120130768-120130768 on Build GRCh38
rs372666086 Chr.12: 120130641-120130641 on Build GRCh38
rs61739941 Chr.12: 120130696-120130696 on Build GRCh38
rs750541838 Chr.12: 120130784-120130785 on Build GRCh38
rs952247198 Chr.12: 120130817-120130817 on Build GRCh38
rs199818602 Chr.12: 120130735-120130735 on Build GRCh38
rs535694539 Chr.12: 120130807-120130807 on Build GRCh38
rs751368346 Chr.12: 120130785-120130785 on Build GRCh38
rs771622431 Chr.12: 120130635-120130635 on Build GRCh38
rs201591763 Chr.12: 120130772-120130772 on Build GRCh38
rs751153232 Chr.12: 120130704-120130704 on Build GRCh38
rs370954521 Chr.12: 120130788-120130788 on Build GRCh38
rs935050760 Chr.12: 120130666-120130666 on Build GRCh38
rs200756540 Chr.12: 120130743-120130743 on Build GRCh38
rs767317125 Chr.12: 120130786-120130786 on Build GRCh38
rs376024579 Chr.12: 120130771-120130771 on Build GRCh38
rs754686456 Chr.12: 120130705-120130705 on Build GRCh38

View Full Product Details