Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12897130-12897391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCDH
Entrez Gene ID
2639
Gene Name
glutaryl-CoA dehydrogenase
Gene Aliases
ACAD5, GCD
UniGene
Hs.532699
Gene Chromosome Location
Chr.19: 12891129-12915345 on Build GRCh38
Transcripts
NM_000159.3 NM_013976.3 NR_102316.1 NR_102317.1 XM_006722721.2 XM_011527899.2 XM_011527900.2 XM_017026580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3403795 c.975A>T p.P325P Chr.19: 12897321-12897321 on Build GRCh38
4074527 c.1011A>G p.A337A Chr.19: 12897357-12897357 on Build GRCh38
5659534 c.912C>A p.H304Q Chr.19: 12897390-12897390 on Build GRCh38
4074525 c.1010C>T p.A337V Chr.19: 12897356-12897356 on Build GRCh38
5659533 c.1044C>A p.H348Q Chr.19: 12897390-12897390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2229460 Chr.19: 12897357-12897357 on Build GRCh38
rs907822395 Chr.19: 12897322-12897322 on Build GRCh38
rs956398569 Chr.19: 12897169-12897169 on Build GRCh38
rs878853156 Chr.19: 12897345-12897345 on Build GRCh38
rs1052014827 Chr.19: 12897250-12897250 on Build GRCh38
rs966157693 Chr.19: 12897231-12897231 on Build GRCh38
rs905869770 Chr.19: 12897171-12897171 on Build GRCh38
rs773310063 Chr.19: 12897307-12897307 on Build GRCh38
rs922106457 Chr.19: 12897242-12897242 on Build GRCh38
rs747248999 Chr.19: 12897287-12897287 on Build GRCh38
rs752234195 Chr.19: 12897361-12897361 on Build GRCh38
rs766325734 Chr.19: 12897323-12897323 on Build GRCh38
rs974474993 Chr.19: 12897258-12897258 on Build GRCh38
rs1039155374 Chr.19: 12897229-12897229 on Build GRCh38
rs766974701 Chr.19: 12897263-12897263 on Build GRCh38
rs748270195 Chr.19: 12897297-12897297 on Build GRCh38
rs933535896 Chr.19: 12897244-12897244 on Build GRCh38
rs914981739 Chr.19: 12897190-12897190 on Build GRCh38
rs769694211 Chr.19: 12897298-12897298 on Build GRCh38
rs899224816 Chr.19: 12897342-12897342 on Build GRCh38
rs780352044 Chr.19: 12897283-12897283 on Build GRCh38
rs757764788 Chr.19: 12897271-12897271 on Build GRCh38
rs746308216 Chr.19: 12897278-12897278 on Build GRCh38
rs538544813 Chr.19: 12897238-12897238 on Build GRCh38
rs1033305066 Chr.19: 12897179-12897179 on Build GRCh38
rs923886574 Chr.19: 12897162-12897162 on Build GRCh38
rs776644984 Chr.19: 12897295-12897295 on Build GRCh38
rs149646305 Chr.19: 12897318-12897318 on Build GRCh38
rs752246593 Chr.19: 12897268-12897268 on Build GRCh38
rs555509244 Chr.19: 12897172-12897172 on Build GRCh38
rs1012338390 Chr.19: 12897347-12897348 on Build GRCh38
rs768894245 Chr.19: 12897291-12897291 on Build GRCh38
rs373540530 Chr.19: 12897235-12897235 on Build GRCh38
rs755657008 Chr.19: 12897366-12897366 on Build GRCh38
rs759176685 Chr.19: 12897355-12897355 on Build GRCh38
rs956480788 Chr.19: 12897191-12897191 on Build GRCh38
rs758793900 Chr.19: 12897279-12897279 on Build GRCh38
rs376338666 Chr.19: 12897348-12897348 on Build GRCh38
rs201663026 Chr.19: 12897275-12897275 on Build GRCh38
rs867057161 Chr.19: 12897368-12897368 on Build GRCh38
rs794726972 Chr.19: 12897343-12897343 on Build GRCh38
rs984420800 Chr.19: 12897276-12897276 on Build GRCh38
rs77028101 Chr.19: 12897226-12897226 on Build GRCh38
rs989021687 Chr.19: 12897219-12897219 on Build GRCh38
rs756144778 Chr.19: 12897258-12897259 on Build GRCh38

View Full Product Details