Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109606163-109606408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNAT2
Entrez Gene ID
2780
Gene Name
G protein subunit alpha transducin 2
Gene Aliases
ACHM4, GNATC
UniGene
Hs.36973
Gene Chromosome Location
Chr.1: 109603091-109619733 on Build GRCh38
Transcripts
NM_005272.3 XM_011541264.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687423 c.586T>G p.F196V Chr.1: 109606312-109606312 on Build GRCh38
3470939 c.540A>T p.K180N Chr.1: 109606358-109606358 on Build GRCh38
4832993 c.573C>G p.V191V Chr.1: 109606325-109606325 on Build GRCh38
674454 c.525C>G p.L175L Chr.1: 109606373-109606373 on Build GRCh38
7095123 c.582G>C p.L194F Chr.1: 109606316-109606316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779949808 Chr.1: 109606372-109606372 on Build GRCh38
rs1799940 Chr.1: 109606350-109606350 on Build GRCh38
rs767182806 Chr.1: 109606302-109606302 on Build GRCh38
rs768739908 Chr.1: 109606290-109606290 on Build GRCh38
rs771116688 Chr.1: 109606269-109606269 on Build GRCh38
rs374210835 Chr.1: 109606371-109606371 on Build GRCh38
rs909225562 Chr.1: 109606190-109606190 on Build GRCh38
rs777323795 Chr.1: 109606211-109606211 on Build GRCh38
rs751633351 Chr.1: 109606347-109606347 on Build GRCh38
rs376412882 Chr.1: 109606328-109606328 on Build GRCh38
rs759910267 Chr.1: 109606327-109606327 on Build GRCh38
rs74113953 Chr.1: 109606246-109606246 on Build GRCh38
rs1012677513 Chr.1: 109606286-109606286 on Build GRCh38
rs746117122 Chr.1: 109606353-109606353 on Build GRCh38
rs966699132 Chr.1: 109606242-109606242 on Build GRCh38
rs750089687 Chr.1: 109606318-109606318 on Build GRCh38
rs753083112 Chr.1: 109606332-109606332 on Build GRCh38
rs758844656 Chr.1: 109606333-109606333 on Build GRCh38
rs201798524 Chr.1: 109606348-109606348 on Build GRCh38
rs192670813 Chr.1: 109606265-109606265 on Build GRCh38
rs777780826 Chr.1: 109606262-109606262 on Build GRCh38
rs749110943 Chr.1: 109606373-109606373 on Build GRCh38
rs745663961 Chr.1: 109606278-109606278 on Build GRCh38
rs774099409 Chr.1: 109606363-109606364 on Build GRCh38
rs564109875 Chr.1: 109606189-109606189 on Build GRCh38
rs373111837 Chr.1: 109606271-109606271 on Build GRCh38
rs199969527 Chr.1: 109606272-109606272 on Build GRCh38
rs998557563 Chr.1: 109606317-109606317 on Build GRCh38
rs113610946 Chr.1: 109606283-109606283 on Build GRCh38
rs932086888 Chr.1: 109606186-109606186 on Build GRCh38
rs780043566 Chr.1: 109606198-109606198 on Build GRCh38
rs200962508 Chr.1: 109606291-109606291 on Build GRCh38
rs761237104 Chr.1: 109606299-109606299 on Build GRCh38
rs1799875 Chr.1: 109606352-109606352 on Build GRCh38
rs758934750 Chr.1: 109606257-109606257 on Build GRCh38
rs540640583 Chr.1: 109606187-109606187 on Build GRCh38
rs751935808 Chr.1: 109606188-109606188 on Build GRCh38
rs755321515 Chr.1: 109606207-109606207 on Build GRCh38
rs769526367 Chr.1: 109606280-109606280 on Build GRCh38
rs762948491 Chr.1: 109606288-109606288 on Build GRCh38
rs370869387 Chr.1: 109606344-109606344 on Build GRCh38

View Full Product Details