Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45677002-45677247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPR
Entrez Gene ID
2696
Gene Name
gastric inhibitory polypeptide receptor
Gene Aliases
PGQTL2
UniGene
Hs.251412
Gene Chromosome Location
Chr.19: 45668187-45683724 on Build GRCh38
Transcripts
NM_000164.3 NM_001308418.1 XM_011526709.2 XM_011526710.2 XM_011526711.2 XM_011526713.2 XM_011526714.2 XM_011526715.2 XM_011526716.2 XM_017026584.1 XM_017026585.1 XM_017026586.1 XM_017026587.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6151467 c.709G>T p.E237* Chr.19: 45677024-45677024 on Build GRCh38
4794966 c.746G>A p.G249E Chr.19: 45677061-45677061 on Build GRCh38
311456 c.778C>T p.L260L Chr.19: 45677093-45677093 on Build GRCh38
3104451 c.727A>T p.S243C Chr.19: 45677042-45677042 on Build GRCh38
4079414 c.789C>A p.G263G Chr.19: 45677104-45677104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs544070842 Chr.19: 45677198-45677198 on Build GRCh38
rs975736428 Chr.19: 45677056-45677056 on Build GRCh38
rs34783010 Chr.19: 45677156-45677156 on Build GRCh38
rs779781285 Chr.19: 45677107-45677107 on Build GRCh38
rs200562041 Chr.19: 45677100-45677100 on Build GRCh38
rs758601217 Chr.19: 45677039-45677040 on Build GRCh38
rs572373461 Chr.19: 45677125-45677125 on Build GRCh38
rs192821802 Chr.19: 45677073-45677073 on Build GRCh38
rs767841022 Chr.19: 45677127-45677127 on Build GRCh38
rs746846121 Chr.19: 45677108-45677108 on Build GRCh38
rs988177813 Chr.19: 45677078-45677078 on Build GRCh38
rs766200229 Chr.19: 45677103-45677103 on Build GRCh38
rs746611074 Chr.19: 45677034-45677034 on Build GRCh38
rs776087069 Chr.19: 45677044-45677044 on Build GRCh38
rs924311488 Chr.19: 45677059-45677059 on Build GRCh38
rs575096601 Chr.19: 45677162-45677162 on Build GRCh38
rs768328822 Chr.19: 45677036-45677036 on Build GRCh38
rs536801405 Chr.19: 45677065-45677065 on Build GRCh38
rs771121612 Chr.19: 45677125-45677126 on Build GRCh38
rs760817713 Chr.19: 45677151-45677151 on Build GRCh38
rs763376212 Chr.19: 45677068-45677068 on Build GRCh38
rs76439420 Chr.19: 45677071-45677071 on Build GRCh38
rs769282692 Chr.19: 45677121-45677121 on Build GRCh38
rs763469705 Chr.19: 45677123-45677123 on Build GRCh38
rs144328094 Chr.19: 45677035-45677035 on Build GRCh38
rs752894812 Chr.19: 45677138-45677138 on Build GRCh38
rs375599983 Chr.19: 45677158-45677158 on Build GRCh38
rs751994544 Chr.19: 45677079-45677079 on Build GRCh38
rs756277634 Chr.19: 45677097-45677097 on Build GRCh38
rs369807135 Chr.19: 45677126-45677126 on Build GRCh38
rs767895891 Chr.19: 45677083-45677083 on Build GRCh38
rs775273533 Chr.19: 45677033-45677033 on Build GRCh38
rs747126365 Chr.19: 45677126-45677126 on Build GRCh38
rs778028385 Chr.19: 45677088-45677095 on Build GRCh38
rs995533205 Chr.19: 45677155-45677155 on Build GRCh38
rs73573273 Chr.19: 45677199-45677199 on Build GRCh38
rs761433216 Chr.19: 45677058-45677058 on Build GRCh38
rs993755587 Chr.19: 45677225-45677225 on Build GRCh38
rs755214748 Chr.19: 45677080-45677080 on Build GRCh38
rs752700139 Chr.19: 45677088-45677088 on Build GRCh38
rs375231803 Chr.19: 45677025-45677025 on Build GRCh38
rs768273981 Chr.19: 45677109-45677109 on Build GRCh38
rs5392 Chr.19: 45677099-45677099 on Build GRCh38
rs558718949 Chr.19: 45677116-45677116 on Build GRCh38
rs944629490 Chr.19: 45677043-45677043 on Build GRCh38
rs771597689 Chr.19: 45677124-45677124 on Build GRCh38

View Full Product Details