Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 151959770-151960043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRE
Entrez Gene ID
2564
Gene Name
gamma-aminobutyric acid type A receptor epsilon subunit
Gene Aliases
-
UniGene
Hs.22785
Gene Chromosome Location
Chr.X: 151953124-151974679 on Build GRCh38
Transcripts
NM_004961.3 XM_011531140.2 XM_017029386.1 XM_017029387.1 XM_017029388.1 XM_017029389.1

SNP Detail

SNP ID Chromosome Location
rs78664911 Chr.X: 151959838-151959838 on Build GRCh38
rs755428067 Chr.X: 151959803-151959803 on Build GRCh38
rs867427972 Chr.X: 151959967-151959967 on Build GRCh38
rs769555117 Chr.X: 151959848-151959848 on Build GRCh38
rs757891766 Chr.X: 151960012-151960012 on Build GRCh38
rs746634401 Chr.X: 151959833-151959833 on Build GRCh38
rs866441978 Chr.X: 151959941-151959941 on Build GRCh38
rs781088784 Chr.X: 151959818-151959818 on Build GRCh38
rs773464738 Chr.X: 151959890-151959890 on Build GRCh38
rs868233464 Chr.X: 151959852-151959852 on Build GRCh38
rs745815848 Chr.X: 151959846-151959846 on Build GRCh38
rs138020133 Chr.X: 151959927-151959927 on Build GRCh38
rs373796944 Chr.X: 151959817-151959817 on Build GRCh38
rs763625359 Chr.X: 151960014-151960014 on Build GRCh38
rs867066503 Chr.X: 151959883-151959883 on Build GRCh38
rs1026207034 Chr.X: 151959823-151959823 on Build GRCh38
rs61740941 Chr.X: 151959945-151959945 on Build GRCh38
rs376183256 Chr.X: 151959855-151959855 on Build GRCh38
rs781092437 Chr.X: 151959843-151959843 on Build GRCh38
rs374796522 Chr.X: 151959959-151959959 on Build GRCh38
rs2856386 Chr.X: 151959888-151959888 on Build GRCh38
rs760038678 Chr.X: 151959956-151959956 on Build GRCh38
rs753230065 Chr.X: 151959963-151959963 on Build GRCh38
rs775496761 Chr.X: 151959853-151959853 on Build GRCh38
rs745631017 Chr.X: 151959832-151959832 on Build GRCh38
rs186167333 Chr.X: 151959813-151959813 on Build GRCh38
rs769529647 Chr.X: 151959875-151959875 on Build GRCh38
rs751285275 Chr.X: 151960018-151960018 on Build GRCh38
rs757155375 Chr.X: 151959841-151959841 on Build GRCh38
rs762535985 Chr.X: 151959926-151959926 on Build GRCh38
rs760847906 Chr.X: 151959915-151959915 on Build GRCh38
rs370193189 Chr.X: 151959925-151959925 on Build GRCh38

View Full Product Details